Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Designed specifically to aid in cell culture growth and propagation, cell and tissue culture dishes are offered in a myriad of formats, materials, and surface modifications. Each dish is designed to promote healthy cells and reproducibility.
Thermo Scientific Nunc EasYDish cell culture dishes are next-generation cell culture dishes designed to improve handling, stacking, and transporting of cell cultures in a lab. Treated with Nunclon Delta-Supra, Sphera, Biolite or not with non-treated, every EasYDish lot is tested with trusted Gibco media to confirm consistent cell growth across multiple cell lines.
We are proud to provide a new product line surface treatment that will support improved cell yield and morphology as well enabling you to do xeno-free or low-serum applications.
See how our surfaces perform with 39 cell types and cell lines at thermofisher.com/supradata
Nunc 35 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150460 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
150318 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 500 | Order |
153066 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
174926 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 20 | 500 | Order |
150470 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 500 | Order |
130180 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 500 | Order |
172931 | Nunc Cell Culture/Petri Dishes | Non-treated | No | Yes, vented | Yes | Sleeve of 20 | 500 | Order |
174904 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
150682 | Nunc Glass Bottom Dishes | Nunclon Delta | No | Yes | Yes | Individually Packaged | 20 | Order |
150680 | Nunc Glass Bottom Dishes | Nunclon Delta | No | Yes | Yes | Individually Packaged | 20 | Order |
174943 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 60mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150462 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 280 | Order |
174888 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 400 | Order |
150326 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
150288 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 400 | Order |
150472 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 280 | Order |
130181 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 500 | Order |
169558 | Nunc Cell Culture/Petri Dishes | Non-treated | Yes | Yes, vented | Yes | Sleeve of 20 | 400 | Order |
174903 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
174906 | Nunc Dishes with UpCell Surface | UpCell | Yes | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
174944 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 100 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150464 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 150 | Order |
150466 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 240 | Order |
172931 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 240 | Order |
150350 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 150 | Order |
150474 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 150 | Order |
150476 | Nunc Cell Culture/PetriDishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 240 | Order |
130182 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 200 | Order |
174902 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Individually Packaged | 6 | Order |
174905 | Nunc Dishes with UpCell Surface | UpCell | Yes | Yes, vented | Yes | Individually Packaged | 6 | Order |
174945 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 150 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150468 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 80 | Order |
168381 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 80 | Order |
157150 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 120 | Order |
150478 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 80 | Order |
130183 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 120 | Order |
Nunc 35 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150460 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
150318 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 500 | Order |
153066 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
174926 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 20 | 500 | Order |
150470 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 500 | Order |
130180 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 500 | Order |
172931 | Nunc Cell Culture/Petri Dishes | Non-treated | No | Yes, vented | Yes | Sleeve of 20 | 500 | Order |
174904 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
150682 | Nunc Glass Bottom Dishes | Nunclon Delta | No | Yes | Yes | Individually Packaged | 20 | Order |
150680 | Nunc Glass Bottom Dishes | Nunclon Delta | No | Yes | Yes | Individually Packaged | 20 | Order |
174943 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 60mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150462 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 280 | Order |
174888 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 400 | Order |
150326 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 500 | Order |
150288 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 400 | Order |
150472 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 280 | Order |
130181 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 500 | Order |
169558 | Nunc Cell Culture/Petri Dishes | Non-treated | Yes | Yes, vented | Yes | Sleeve of 20 | 400 | Order |
174903 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
174906 | Nunc Dishes with UpCell Surface | UpCell | Yes | Yes, vented | Yes | Sleeve of 5 | 30 | Order |
174944 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 100 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150464 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 150 | Order |
150466 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 240 | Order |
172931 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 240 | Order |
150350 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 150 | Order |
150474 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 150 | Order |
150476 | Nunc Cell Culture/PetriDishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 240 | Order |
130182 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 200 | Order |
174902 | Nunc Dishes with UpCell Surface | UpCell | No | Yes, vented | Yes | Individually Packaged | 6 | Order |
174905 | Nunc Dishes with UpCell Surface | UpCell | Yes | Yes, vented | Yes | Individually Packaged | 6 | Order |
174945 | Nunclon Sphera Dishes | Nunclon Sphera | No | Yes | Yes | Sleeve of 5 | 20 | Order |
Nunc 150 mm tissue culture dishes feature:
Cat. No. | Description | Surface | Gridded | Lid | Sterile | Pack size | Case size | |
---|---|---|---|---|---|---|---|---|
150468 | Nunc EasYDish Dishes | Nunclon Delta | No | Yes | Yes | Sleeve of 10 | 80 | Order |
168381 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 80 | Order |
157150 | Nunc Cell Culture/Petri Dishes | Nunclon Delta | No | Yes, vented | Yes | Sleeve of 10 | 120 | Order |
150478 | Nunc EasYDish Dishes | Nunclon Supra | No | Yes | Yes | Sleeve of 10 | 80 | Order |
130183 | BioLite Cell Culture Treated Dishes | BioLite | No | Yes | Yes | Sleeve of 10 | 120 | Order |
Non-treated Thermo Scientific Nunc Rectangular Dishes with ANSI™ standard dimensions are optimized for molecular biology applications using robotic instruments.
Perform high quality imaging in the ease of a 35 mm dish. The Thermo Scientific Nunc Glass Bottom Dish combines the convenience of a standard 35 mm dish with the imaging benefits of cover glass to provide the optimal optical characteristics required for high-magnification microscopy and confocal image analysis. The interior surface of the dish bottom features Nunclon Delta surface treatment.
With more than 100 combinations of format and surface, our polystyrene cell culture plates help promote healthy cells and generate reliable results.
Adjustable blades for optimal application flexibility. Reduce cell damage during the harvesting process with our sterile, nonpyrogenic cell scrapers.
For Research Use Only. Not for use in diagnostic procedures.