Autoclaves
Reliability you can count on with Tuttnauer
Autoclaves trusted by hospitals, research institutes, clinics and laboratories
Thermo Fisher Scientific has strategically partnered with industry-leading autoclave supplier, Tuttnauer for Australia and New Zealand. For over 90 years, Tuttnauer has been meeting customer expectations with top-quality and high-performance products. Meeting the highest standards for quality, safety and performance.
Maintain lab safety and infection control with the Tuttnauer range of small and large autoclaves for cleaning and sterilisation purposes. The range includes systems to suit a wide range of applications and throughput needs.
Small laboratory autoclaves
Steam autoclave sterilisers
Steam autoclave sterilisers with advanced technology in a range of chamber sizes from 23 to 160 L. Designed for laboratory applications used in research institutes, universities, and the medical, pharmaceutical, biotechnology, food, and chemical industries. Both benchtop and floor standing options available.
Large laboratory autoclaves
Large capacity horizontal lab sterilisers
Tuttnauer offers sterilisers that provide high quality repeatable performance and accountability for a wide range of lab applications, including liquids, glassware, instruments, porous loads and other laboratory items. Chamber sizes ranging from 310 to 8840 liters and options for single or double doors. All manufactured from long lasting 316L Stainless Steel. Customised solutions are available including unique chamber sizes and advanced sterilisation cycles.
Biohazard autoclaves
Bio Safety Level (BSL) biohazard autoclaves
Tuttnauer BSL biohazard autoclaves are designed to meet stringent Bio Safety Level requirements for BSL3 and BSL4 facilities. These include a hermetically sealed barrier between the different risk level zones and effective sterilisation and disposal of all effluent and gases.
Medical autoclaves
Medical sterilisation solutions
As a global leader in sterilisation and infection control, Tuttnauer provides autoclaves that satisfy the needs of any hospital operating room, central sterile services department or medical clinic.
Resources
Videos
Tuttnauer laboratory solutions
Agar preparation with Tuttnauer
Documents