Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Thermo Fisher Scientific is the authorised distributor for the Pinnacle range of laboratory freezers and refrigerators in Australia and New Zealand.
There are a number of common features including:
The Pinnacle purpose-built pharmacy refrigerators and freezers have been designed in Australia to meet vaccine storage guidelines. Ideal for pharmacies and hospitals, they provide reliable temperature control and are easy to use.
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPRS361GD/SD | 36 L capacity, glass / solid door option | 450 x 490 x 535 | 27 kg |
TMOPRS661GD/SD | 66 L capacity, glass / solid door option | 450 x 490 x 730 | 29.5 kg |
TMOPRS901GD/SD | 90 L capacity, glass / solid door option | 490 x 520 x 820 | 35 kg |
TMOPRS1451GD/SD | 145 L capacity, glass / solid door option | 610 x 610 x 820 | 43 kg |
TMOPRS3001GD/SD | 300 L capacity, glass / solid door option | 610 x 655 x 1500 | 71 kg |
TMOPRS4301GD/SD | 430 L capacity, glass / solid door option | 610 x 700 x 1870 | 84 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPFS20661SD | 66 L capacity, (-20°C) solid door only | 490 x 520 x 820 | 35 kg |
TMOPFS201151SD | 115 L capacity, (-20°C) solid door only | 610 x 610 x 820 | 41 kg |
TMOPFS204001SDS | 400 L capacity, (-20°C) solid door only, static | 600 x 620 x 1880 | 77 kg |
TMOPFS206001SDS | 600 L capacity, (-20°C) solid door only, static | 775 x 750 x 1900 | 99 kg |
TMOPFS401051SD | 105 L capacity, (-40°C) solid door only | 595 x 595 x 815 | 41 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPSPRS1451SD | Spark proof, solid door only | 610 x 610 x 820 | 43 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPSPFS1151SD | 115 L, spark proof (-20°C) solid door only | 610 x 610 x 820 | 43 kg |
TMOPSPFS204001SD | 400 L, spark proof (-20°C) solid door only | 600 x 620 x 1880 | 77 kg |
TMOPSPFS206001SD | 600 L, spark proof (-20°C) solid door only | 775 x 750 x 1900 | 99 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPRFS200/1001 | 200 L glass door refrigerator (-20°C) 100 L solid door freezer | 600 x 610 x 1750 | 89 kg |
Pinnacle laboratory refrigerators are designed to safely store samples, reagents, kits and scientific research substances safely between 2°C and 10°C.
Pinnacle laboratory freezers come in a range of sizes, from smaller models that you can comfortably fit under a bench to larger ones that can store large quantities of samples. Temperatures range from -20°C to -40°C.
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPRP6301GD/SD | 630 L capacity, full length glass/solid door option | 790 x 825 x 1965 | 150 kg |
TMOPRP11502GD/SD | 1,150 L capacity, 2 door, full length glass/solid door option | 1310 x 825 x 1965 | 226 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
TMOPFP206301SD | 630 L capacity, (-20°C) solid door only | 790 x 825 x 1965 | 150 kg |
TMOPFP206301SDS | 630 L capacity (-20°C) solid door only, static | 790 x 825 x 1965 | 150 kg |
TMOPFP306301SD | 630 L capacity, (-30°C) solid door only | 790 x 825 x 1965 | 150 kg |
TMOPFP2011502SD | 1,150 L capacity, 2 door, (-20°C) solid door only | 1310 x 825 x 1965 | 230 kg |
Part number | Description | External dimensions WxDxH mm | Unit weight |
---|---|---|---|
PSPF-P-20-630-1-SD | 630 L, spark proof (-20°C) solid door only | 790 x 825 x 1965 | 150 kg |