Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We offer a comprehensive range of controlled-atmosphere glove boxes and chambers designed to fit your laboratory needs. The Coy Lab range includes vinyl anaerobic chambers for microbiology/biochemistry studies, hypoxic chambers for cancer research, cell biology, and ischemia research, and basic glove boxes for general basic applications.
Anaerobic chambers are ideal for use in microbiology research, clinical microbiology, biochemistry, cell culture, human microbiome studies, biofuels, and more. Coy Anaerobic Chambers eliminate oxygen to 0–5 parts per million (ppm) using a hydrogen gas mix (5% or less) reacting with a palladium catalyst to remove oxygen by forming a water molecule. A vacuum airlock is used to reduce oxygen levels prior to transfer of samples and tools in and out of the glove box. Access ports allow sample manipulation without ever exposing your samples to ambient conditions.
Temperature incubation options include heating the entire glove box to be your incubator or placing a specially designed Coy incubator within the workstation interior for a more comfortable working atmosphere for the users. Large ports are included for large equipment transfer and smaller feed-through ports are available on every unit to pass cords/tubing in and out of the chamber in a sealed fashion.
Vinyl
Aluminium
Polymer
For applications that require incubation with temperature control, Coy has several solutions. The advantages and disadvantages of these options should be factored into your selection.
With this option, the entire chamber becomes an incubator. Incubation capacity is limited only by the size of the chamber. Disadvantages include larger temperature variations and a less comfortable work area when compared to an unheated chamber with incubator option. It is a less expensive solution than purchasing an unheated chamber with an incubator.
An unheated chamber with an incubator has the advantage of incubation with more precise temperature control when compared to the same process with just a heater in the chamber. The chamber is more comfortable to work in than a heated chamber. The disadvantage is the limited incubation capacity, and odd-size containers may not fit inside.
The advantage with this option is that the heater can be kept turned off until the incubator reaches overflow capacity, at which point it can be turned on. While this is a best-of-both-worlds choice, it is more expensive.
Only Coy Vinyl Anaerobic Chambers can be retrofitted in the field with an incubator. All three types of anaerobic chambers can have a heater specified at initial purchase or later.
Coy vacuum airlocks (manual or automatic) remove O2 from ambient conditions to achieve acceptable levels of O2 prior to transfer to/from the chamber. The airlock reaches the low O2 level through a multiple vacuum/purge procedure. The standard factory procedure is to pull a vacuum to 20” of mercury (Hg), then purge back to 1” of Hg using an inert background gas. Another cycle is repeated with the inert gas. A third cycle is performed with the H2 gas mix used for anaerobic work. On the third and final purge, the vacuum level is brought back to ambient with just a slight vacuum left to hold the seal. The seal is easily broken by the user when opening the door.
Automatic airlocks are equipped with an advanced electronic program that allows adjustments in vacuum levels, number of cycles, calibration of pressure sensors, and programmable profiles, depending on the type of work being done in the chamber. All vacuum pumps are equipped with moisture traps to prevent excess moisture from the chamber or gas tanks from entering the pump. This helps prevent pump vane rusting, which can significantly damage the pump. Manual airlocks operate with the user turning ball valves for the gas and pump while monitoring the vacuum levels. Automatic airlocks operate with the touch of a button.
This update to the airlock door has a spring-loaded corner pivot which allows the door to swing up while parallel to the airlock, saving valuable space. Older Coy airlocks may be retrofitted in the field with the new style door and updated digital electronics.
The purge-only units operate by flushing background gas into the airlock to push out excess oxygen prior to opening the interior door. Ideal purge times are provided by Coy for various O2 levels. Automatic units allow the user to preset a specific time to purge the airlock with a simple touch of a button. With manual units, the user operates ball valves and times the purge. Purge units, while initially less expensive to purchase than vacuum airlocks, generally have a higher operational cost and a longer transfer time.
When your research requires consistent atmospheric control to maintain hypoxia, normoxia, or hyperoxia, you can count on Coy’s Hypoxic Chambers. Easy to use and easy to customise, Coy’s complete line of glove boxes and cabinets allows you to maintain desired levels of O2, CO2, temperature, and humidity in either cell and tissue culture or animal applications.
The feedback loop system uses an O2 sensor mounted inside the glove box/cabinet to monitor oxygen levels. Based on these sensor readings and the user-adjustable set points you establish, the system flushes the chamber with N2, displacing O2, and returning the chamber to ideal experimental conditions. For hyperoxic conditions, an O2 source is added.
Perform incubation, culture maintenance, and analysis in the same environment with Coy’s O2 Control Glove Box for Tissue Culture. CO2 and O2 concentrations will need to be adjusted from their normal atmospheric air concentrations within the experimental chamber or the glove box.
N2 is used as a flush gas to displace the atmospheric gas mix to the experimental presets. The controllers, combined with the sealed glove box and airlock, provide a more accurate and uninterrupted environment than an incubator or cabinet system by eliminating exposure to non-experimental conditions.
The Coy O2 Control Glove Box for Cell and Tissue Culture has temperature control up to forty degrees Celsius. For long-term incubation, users will want to maintain high humidity levels to prevent sample from drying out.
Coy offers a small humidified incubation box that allows samples to be incubated at levels of moisture at or near saturation while minimizing the amount of moisture that escapes into the glove box. Coy provides two solutions for controlling and removing moisture from this hypoxic chamber — a desiccant-based system or an automatic dehumidifier, which is recommended.
Coy O2 Control Glove Boxes for tissue culture offer the following optional features:
All O2 Control Glove Boxes for tissue culture are supplied with the following:
Coy O2 Control Glove Boxes are equipped with a purge-only airlock, which is a transfer chamber that equilibrates O2 levels by purging excess O2 prior to opening the door into the actual glove box and placing items inside. Automatic units use a specific preset time for purges based on protocol and desired glove box O2 levels. Once preset, the airlock is operated with the touch of a button by lab personnel. With manual units, the user operates a valve and times the purge.
The Coy Humidified Incubation Box for Cell and Tissue Culture is a separate unit that sits inside the glove box. It allows cultures to be humidified with the same atmosphere content (gas and temperature) without immediately increasing the humidity of the rest of the box.
Coy O2 Control Glove Boxes for InVitro Studies are available in polycarbonate (three standard sizes) and aluminum (two standard sizes). The choice of material depends on your research needs and budget. Aluminum is generally more robust and, therefore, offers more service years than polycarbonate. Polycarbonate is a less expensive option and is easily customisable.
Coy 1 Person O2 Control Glove Box |
Coy 2 Person O2 Control Glove Box |
||
Construction material | Polycarbonate plastic and aluminum | ||
Total glove box footprint | 137 L x 61 D (H=72) cm |
152 L x 61 D (H=72) cm |
|
Interior work area | 104 L x 59 D cm | 188 L x 59 D cm | |
Number of glove ports | 2 | 4 | |
Glove port style | Gloveless sleeve standard Vacuum/purge sleeves or gloved options available |
||
Pressure regulation | Automatic pressure relief valve and patented diaphragm top to compensate for hands entering through the gloves | ||
Moisture control | Glove box dehumidifier to maintain non-condensing moisture levels | ||
Glove box temperature control, and range | +3 ambient to 45°C Note: Not used if using internal incubator(s) |
||
Glove box working incubation capacity (# of 100mm Petri dishes) | Up to 450 x 90 mm Petri dishes | Up to 600 x 90 mm Petri dishes | |
Airlock capacity | 30 x 100 mm plates | ||
Airlock operation | Automatic timer | ||
Airlock transfer time | 10–190 seconds user adjustable | ||
Interior power supply | 5 outlet 230 V | ||
Power outlets needed | 4 for standard operation Note certain options will require additional outlets |
||
Glove material | Neoprene sleeve with latex cuff | ||
Large side door | Standard options for removable front panel |
||
Gas requirements | N2, CO2 and Oxygen gas sources | ||
Oxygen control | 0–100% +/- 0.1% resolution | ||
CO2 control | 0–20% +/- 0.1% resolution | ||
Humidified incubation box | Separate internal incubator designed to minimise media loss |
Basic glove boxes are an economical way to perform and maintain a wide range of atmospheres and functions, including use as a dry glove box, inert gas or nitrogen glove box. The base unit is a manual system, however over 20 different options are available to configure the unit for your specific glove box needs. Equipped with a purge valve, automatic pressure relief valves, large side door, interior power supply, and hermetically sealed glove ports, these glove boxes can provide your desired environment for critical operations in industries such as semiconductor and pharmaceutical testing and manufacturing.
All glove boxes are fabricated to meet rigorous standards for cleanliness and durability. Solvent welding ensures a strong, airtight structure that won’t leak. The gaskets on the side doors provide a safe, positive seal without interfering with full access to the internal chamber.
Vinyl glove boxes
Coy vinyl glove boxes are ideal for simple containment and superior operator comfort. Flexible vinyl chambers are a simple, low-cost alternative to rigid glove boxes for sealed environments or simple containment applications. They come equipped with gas inlet and automatic pressure relief system.
Aluminium glove boxes
Coy aluminium glove boxes are designed for durable and harsh chemical operations. With a gas inlet valve and an automatic pressure relief valve, the Coy aluminium glove boxes are ideal for simple containment or dry gas purge operations.
Polymer glove boxes
Coy polymer glove boxes are flexible to design and easy to operate, ideal for most simple purge glove box applications. Rigid polymer glove boxes are an economical way to perform and maintain a wide range of atmospheres and functions. The modular design lets you select the options you need for your operation.
Humidity and dry polymer glove boxes | Humidity and dry aluminum glove boxes | Basic polymer glove boxes | Basic aluminum glove boxes | Basic vinyl glove boxes | ||
---|---|---|---|---|---|---|
Atmosphere filtration system | ✓ | ✓ | ✓ | ✓ | ✓ | |
Feed-thru adaptor | ✓ | ✓ | ✓ | ✓ | ✓ | |
Microscope view port | ✓ | ✓ | ✓ | ✓ | ✓ | |
Extra gloves/ports | CC | CC | CC | CC | ✓ | |
Fluorescent lights | ✓ | ✓ | ✓ | ✓ | ✓ | |
Removable rear panel | ✓ | CC | ✓ | CC | ||
Stainless steel pan | CC | CC | CC | CC | ✓ | |
Shelf assemblies | ✓ | SA | ✓ | SA | ✓ | |
Gas leak detector | ✓ | ✓ | ✓ | ✓ | ✓ | |
Support tables | ✓ | ✓ | ✓ | ✓ | ✓ | |
UV lights | ✓ | CC | ✓ | CC | ||
Added capacity shelf (external) | ✓ | ✓ | ✓ | ✓ | ✓ | |
Extra gas inlets and flow meters | ✓ | ✓ | ✓ | ✓ | ✓ | |
Temperature control options for humidity/dry glove boxes | ||||||
Heaters (digital PID) | ✓ | ✓ | ✓ | ✓ | ✓ | |
Chiller | ✓ | ✓ | ✓ | ✓ | ✓ | |
Added capacity dryer (assumes desiccant drying system) | ✓ | ✓ | ✓ | ✓ | ✓ | |
Dry gas purge system (can be substituted for desiccant system) | ✓ | ✓ | ✓ | ✓ | ✓ | |
Chemical resistant upgrades | ||||||
Aluminum glove port panel | ✓ | |||||
Tempered glass viewing screen | ✓ | |||||
Tempered glass top | ✓ | |||||
Airlocks | ||||||
Pass-through airlock | ✓ | ✓ | ||||
Manual purge airlock | ✓ | ✓ | ||||
Automatic purge airlock | ✓ | ✓ | ||||
Vacuum airlocks | CC | CC | CC | CC | CC | |
Sliding shelf for the airlock | ✓ | ✓ | ✓ | ✓ | ✓ | |
Large side door (1 is standard but 2nd one can be added to provide access on glove boxes.) | ✓ | ✓ | ||||
Humidification system | ✓ | ✓ | CC | |||
Dehumidification system manual | ✓ | ✓ | CC | |||
Humidity controller | ✓ | ✓ | ✓ | |||
Vinyl tubing | ✓ | ✓ | ✓ | ✓ | ✓ | |
Circulation fans | ✓ | ✓ | ✓ | ✓ | ✓ | |
Gloveless sleeve upgrade | SE | SE | ✓ | ✓ | ||
Arm port plugs | SE | SE | ✓ | ✓ | ||
Gas regulators N2, CO2, O2 available | ✓ | ✓ | ✓ | ✓ | ✓ | |
Digital hygrometer (hand held) | ✓ | ✓ | ✓ | ✓ | ✓ | |
Ionizer, workstation | ✓ | ✓ | ✓ | ✓ | ✓ | |
Iris ports | ✓ | ✓ | ✓ | ✓ | ✓ | |
Static dissipative mat | ✓ | ✓ | ✓ | ✓ | ✓ |
Key:
✓: Available option/accessory CC: Common Customisation
SA: Standard Accessory SE: Standard Equipment