Simple, flexible, and reliable
   CO2 resistant shakers
Use your existing CO2 incubator to investigate suspension culture and scale-up. Save space and achieve reliable around-the-clock operation with CO2 resistant shakers ideally suited to maximize cell health within your existing CO2 incubator.
Choose shaker    Choose accessories
Reliability
reliable co2 resistant shakers
Sealed electronics and proven mechanism technologies provide minimal heat dissipation to ensure conditions within the CO2 chamber are not affected, and cells can thrive.
Simplicity
easy-to-use co2 resistant shakers
CO2 resistant shakers can be easily removed from the incubator to facilitate cleaning and disinfection, protecting your cultures from unwanted contamination, providing ease of use for multi-tasking cell culture labs.
Flexibility
flexible co2 resistant shakers
Flexibility allows it to go from the CO2 incubator to the benchtop to the microbiological incubator for culturing mammalian suspension cells, insect cells, bacteria, fungi or other cell types for maximum value and versatility.
Choose a CO2 resistant shaker
 CO2 Resistant Shaker 
3mm Orbit
CO2 Resistant Shaker
19mm Orbit
MaxQ 416 Shaker
25mm Orbit
co2 resistant shakermaxq 416 shaker
 Learn moreLearn moreLearn more
Maximum vessel size5L Erlenmeyer flask5L Erlenmeyer flask6L Erlenmeyer flask
Available platforms14.1 x 11.8 in.
18 x  18 in.
14.1 x 11.8 in.
18 x  18 in.
18 x 18 in.
18 x 24 in.
Speed30 to 600 rpm30 to 300 rpm25 to 525 rpm
Environmental
temperature
range
4° to 60°C4° to 60°C4° to 40°C
Humidity limits20% to 95%20% to 95%50% to 80%
Exterior dimensions
(H x W x D)
3.8 x 14.2 x 11.8 in.
(9.7 x 36 x 30 cm)
3.8 x 14.2 x 11.8 in.
(9.7 x 36 x 30 cm)
7.8 x 30.5 x 26 in.
(19.8 x 77.5 x 66 cm)
Orbit.118 (0.3 cm).75 in. (1.9 cm)1 in. (2.5 cm)
Weight capacity22 lbs (10 kg)*13 lb. (6 kg)71 lb. (32 kg)
Data output4-20 mA4-20 mANA
AccessoriesPlatforms & Clamps ›
Test Tube Racks ›
Plugs & Accessories ›
Platforms & Clamps ›
Test Tube Racks ›
Plugs & Accessories ›
Platforms & Clamps ›
Test Tube Racks ›
Accessories ›
*Maximum load when <150 rpm
Optimize your CO2 resistant shaker
co2 resistant shaker platforms
Maximize efficiency and productivity by mixing and matching labware on various platform and holder types and truly customize your shaker to fit your unique needs. Go ›
co2 resistant shaker racks
Racks and trays will outfit your shaker to accommodate a wide variety of applications and objectives, and to keep conditions clean and safe. Go ›
co2 resistant shaker plugs
Plugs and miscellaneous accessories ensure you have the correct configuration for your geographic region plus parts to keep your shaker performing at its best. Go ›
CO2 Platforms
MaxQ 416 HP Shaker Platforms & Clamps
CO2 Shaker Tube Racks
MaxQ 416 HP Shaker Test Tube Racks & Trays
CO2 Shaker Plugs & Accessories
MaxQ 416 HP Shaker Accessories
Maximum shaker capacity inside our CO2 Incubators
Thermo Scientific CO2 incubators and environmental chambers are ideal to pair with your Thermo Scientific CO2 resistant shakers for the flexibility to scale up suspension cultures while continuing stationary cultures in one system.
Capacity with CO2 Resistant Shaker (14.1 x 11.8 in platform)
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Forma 310 Direct Heat CO2 Incubator19470 x 470mm16kg slide in and out
Forma Steri-Cycle CO2 Incubator19470 x 470mm16kg slide in and out
Forma Series II 3110 Water-Jacketed CO2 Incubator19470 x 470mm13.6kg slide in and out
Large-Capacity Reach-In CO2 Incubator8 units, 4 on each shelf72777 X 655mm90kg fixed reinforced shelves* (1900005)
Heracell 150i CO2 Incubators -Stainless19423 x 465mm10kg
Heracell 150i CO2 Incubators -Copper19423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Stainless19423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Copper19423 x 465m10kg
Heracell 240i CO2incubator-Stainless118560 x 500mm10kg
Heracell 240i CO2incubator-Copper118560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Stainless118560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Copper118560 x 500mm10kg
Thermo Scientific Forma 3940 Series Environmental Chambers348656 x 777mm90kg fixed reinforced shelves* (1900005)
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Capacity with CO2 Resistant Shaker (18 x 18 in platform):
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Forma 310 Direct Heat CO2 Incubator00470 x 470mm16kg slide in and out
Forma Steri-Cycle CO2 Incubator00470 x 470mm16kg slide in and out
Forma Series II 3110 Water-Jacketed CO2 Incubator00470 x 470mm13.6kg slide in and out
Large-Capacity Reach-In CO2 Incubator8 units, 4 on each shelf80777 X 655mm90kg fixed reinforced shelves* (1900005)
Heracell 150i CO2 Incubators -Stainless00423 x 465mm10kg
Heracell 150i CO2 Incubators -Copper00423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Stainless00423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Copper00423 x 465m10kg
Heracell 240i CO2incubator-Stainless110560 x 500mm10kg
Heracell 240i CO2incubator-Copper110560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Stainless110560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Copper110560 x 500mm10kg
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Capacity with MaxQ 416 HP Shaker
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Thermo Scientific Large-Capacity Reach-In CO2 Incubator248777 X 655mm90kg fixed reinforced shelves* (1900005)
Thermo Scientific Forma 3960 Series Environmental Chambers348656 x 777mm90kg fixed reinforced shelves* (1900005)
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Capacity with CO2 Resistant Shaker (14.1 x 11.8 in platform)
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Forma 310 Direct Heat CO2 Incubator19470 x 470mm16kg slide in and out
Forma Steri-Cycle CO2 Incubator19470 x 470mm16kg slide in and out
Forma Series II 3110 Water-Jacketed CO2 Incubator19470 x 470mm13.6kg slide in and out
Large-Capacity Reach-In CO2 Incubator8 units, 4 on each shelf72777 X 655mm90kg fixed reinforced shelves* (1900005)
Heracell 150i CO2 Incubators -Stainless19423 x 465mm10kg
Heracell 150i CO2 Incubators -Copper19423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Stainless19423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Copper19423 x 465m10kg
Heracell 240i CO2incubator-Stainless118560 x 500mm10kg
Heracell 240i CO2incubator-Copper118560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Stainless118560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Copper118560 x 500mm10kg
Thermo Scientific Forma 3940 Series Environmental Chambers348656 x 777mm90kg fixed reinforced shelves* (1900005)
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Capacity with CO2 Resistant Shaker (18 x 18 in platform):
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Forma 310 Direct Heat CO2 Incubator00470 x 470mm16kg slide in and out
Forma Steri-Cycle CO2 Incubator00470 x 470mm16kg slide in and out
Forma Series II 3110 Water-Jacketed CO2 Incubator00470 x 470mm13.6kg slide in and out
Large-Capacity Reach-In CO2 Incubator8 units, 4 on each shelf80777 X 655mm90kg fixed reinforced shelves* (1900005)
Heracell 150i CO2 Incubators -Stainless00423 x 465mm10kg
Heracell 150i CO2 Incubators -Copper00423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Stainless00423 x 465mm10kg
Heracell VIOS 160i CO2incubator-Copper00423 x 465m10kg
Heracell 240i CO2incubator-Stainless110560 x 500mm10kg
Heracell 240i CO2incubator-Copper110560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Stainless110560 x 500mm10kg
Heracell VIOS 250i CO2incubator-Copper110560 x 500mm10kg
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Capacity with MaxQ 416 HP Shaker
CO2 IncubatorMaximum ShakersMaximum 500mL FlasksShelf DimensionLoad Limit per Shelf
Thermo Scientific Large-Capacity Reach-In CO2 Incubator248777 X 655mm90kg fixed reinforced shelves* (1900005)
Thermo Scientific Forma 3960 Series Environmental Chambers348656 x 777mm90kg fixed reinforced shelves* (1900005)
*Some incubators and environmental chambers will require reinforced shelving to support the weight of a shaker. Please refer to the loading limit per shelf indicated for your incubator or contact your sales representative for more information.
Related products