Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
In New Zealand, in partnership with Merck, we offer the Milli-Q® Lab Water Solutions portfolio which offers a broad range of pure and ultrapure water purification systems and services.
This portfolio is designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments.
Whether you’re after Ultrapure (Type 1) or pure (Type 2, Type 3/RO) water; whether your feed water is from a tap or pre-treated water; whether you’re needing a benchtop polisher system or to supply lab water for your entire building, we have a solution for you.
Norms define different laboratory water grades for both technical and economical reasons. The purpose of these norms is to ensure that the right water quality is used for your applications, while limiting laboratory operating costs. Type 1 water has the highest purity but is also the most expensive to generate.
If your application requires different types of water, let our team assist with designing a water purification system specific to meet your needs.
We offer complete solutions designed to meet your laboratory water needs. Our Lab Water experts will take the time required to evaluate your unique situation and to propose the best-adapted lab water purification solution.
A pre-filtration setup before the system is recommended with potable feedwater source. Will always need a tank/reservoir to store the product water.
Water production rate | Systems |
---|---|
Up to 13,000 L/day | Milli-Q™ HR 7000 series |
Up to 480 L/day | RIOs™ Essential |
Up to 150 L/day | RIOs™ |
A pre-filtration setup before the system is recommended with potable feedwater source. Will always need a tank/reservoir to store the product water.
Water production rate | Systems | |
---|---|---|
Up to 9,000 L/day | Milli-Q™ HX 7000 series | |
Up to 3,000 L/day | Milli-Q™ HX 7000 SD series | |
Up to 300 L/day | Milli-Q™ IX 7003/05/10/15 | Elix™ Essential |
Requires pre-treated water (RO or Type II water quality) to ‘feed’ the Type I water system.
Water production rate | Systems | |
---|---|---|
Up to 2 L/min | Milli-Q™ IQ 7000 | Milli-Q™ EQ 7000 |
Up to 1.5 L/min | Synergy™ | |
Up to 0.5 L/min | Simplicity™ |
A pre-filtration setup before the system is recommended with potable feedwater source. Unit will produce ‘pure’ water and requires storage reservoir; Type I water produced on demand.
Milli-Q™ IQ 70XX series produces Type II and Type I product water.
Water production rate | Systems | |
---|---|---|
Up to 2 L/min | Milli-Q™ IQ 70003/05/10/15 | Milli-Q™ EQ 7008/7016 |
Up to 0.5 L/min | Direct-Q™ |
Please note: The Millipore range of water purification systems are only available in New Zealand. If you are in Australia, please view our range available here.