RephiLe Laboratory Water Purification Systems
A revolution in filtration
Explore RephiLe's water purification solutions
Innovative and high-quality laboratory water systems
Thermo Fisher Scientific has partnered with RephiLe to provide cutting-edge purification solutions for biotechnology and life science labs. The product range encompasses pure water systems and consumables. The primary objective is to equip our customers with reliable tools that guarantee the quality of their research or testing results, all while maintaining an affordable price point.
Control management console
RephiLe's water technology platforms incorporate a range of advanced purification technologies, including continuous electro-deionization (CEDI) technology and reverse osmosis, to cater to the diverse requirements of laboratories. Alongside optimised flow paths, each water system is equipped with a sophisticated process control management console. This console enables continuous monitoring of purity levels following each purification step, ensuring consistent and reliable performance.
Key features of RephiLe water systems
Key features
  • RFID tracking of cartridges and accessories
  • Real time information and alarm notifications on a smartphone, tablet or laptop (Rephilink)
  • Automatic data backup for up to 2 years
  • A tank sanitisation module with a mercury-free UV lamp is included in all tank kits
  • Inline TOC monitoring available in selected units
  • A full range of ultrapure cartridges including ultra-low TOC, low Mg, low boron, ICP and DI type
  • All water products generated by the system can be dispensed directly from the main system
  • Automatic system shut off upon detecting any water leakage
Explore the RephiLe range
NuZar benchtop water systems
NuZar benchtop water systems deliver powerful performance in a compact size.
  • Plug n Play. Simple to install and maintain
  • Integrated 10L tank which can be upgraded to 30L to 350L tank
  • System, tank and dispenser are all wall mountable
  • Reuse of RO concentrate to reduce wastewater and increase yield
Genie benchtop water systems
Cutting-edge technologies and sophisticated design come together to deliver an exceptional user experience.
  • One system can connect up to 10 dispensers
  • Drop by drop dispensing
  • Tank recirculation model guarantees water quality in the tank
  • Built in continuous liquid level sensor in the storage tank
  • System, tank and dispenser are all wall mountable
  • Reuse of RO concentrate to reduce wastewater and increase yield
Large Genie benchtop water systems
This compact system efficiently utilizes your laboratory space while enabling a daily production capacity of up to 1,900L.
  • The automatic bypass diverts the RO or EDI water to the drain if it does not meet preset quality standards
  • Reuse of RO concentrate to reduce wastewater and increase yield
  • System, tank and dispenser are all wall mountable
  • External prefiltration, feed tank, booster pump, distribution pump, tank sanitisation and leak protector are all controlled from the main system console
Super-Genie centralised workstations
Centralised workstations with daily pure water production up to 14,000L from tap feed water.
  • The automatic bypass diverts the RO or EDI water to the drain if it does not meet preset quality standards
  • Reuse of RO concentrate to reduce wastewater and increase yield
  • Built in continuous liquid level sensor in the storage tank
  • Integrated 100L reservoir which is upgradable to suit volume requirements
Genie De-Ion ultrapure water purification kit
A water purification and dispensing kit for trace and ultra-trace element analysis.
The rapid increase of detection capabilities of modern analytical techniques, including ICP-MS, nano UHPLC and nano LC-MS, greatly simplifies the quantification of trace and ultra-trace elements in diversified scientific and industrial fields. With their extreme sensitivity, low detection limits, high throughput and lots of other advantages, these analytical techniques project rigorous requirements for the control of contamination, in which, purity of the water substantially used during the analytical process is a key to a meaningful analysis.
Resources
Videos
Genie lab water system