The presence of contaminants in water can lead to a number of health issues, so accurately checking water quality is essential in many industries.
LabServ manufactures and supplies a comprehensive range of disposable and reusable laboratory plasticware, including PET water sampling containers and bottles, as well as general purpose HDPE bottles, which are ideal for water testing applications.
Choose a greener option
Choosing our PET and HDPE options ensures you are selecting the most recyclable materials, making it a sustainable choice for all your laboratory applications. PET and HDPE are widely used due to their excellent durability, recyclability and multitude of uses.
Our PET range of square containers and bottles are the perfect solution for collecting, storage and transport of water samples to the laboratory. Each product is sprayed with a pre-dosed concentration of sodium thiosulphate based on the volume of the bottle. Pair the sampling containers with our HDPE bottles for additional support within your workflow.
LabServ PET containers and bottles for water sampling
LabServ PET containers and bottles
  • Square containers available in 200mL to 1000mL
  • Bottles available in 250mL and 500mL
  • Gamma sterilised
  • Pre-dosed with sodium thiosulphate
  • Sodium thiosulphate labelled
  • CoA and CoC available
  • Blue wadded cap for square containers
  • Bottles come with HDPE screw caps
Browse PET containers     Browse PET bottles
LabServ HDPE general purpose bottles
LabServ HDPE bottles
  • Round bottles available from 60mL to 500mL
  • Narrow mouth – Neck finish 28mm
  • Opaque
  • Bottle only and bottle and cap available
  • Screw cap – available separately
  • All bottles manufactured in a controlled environment to minimize risk of contamination
  • HDPE raw material is selected to meet rigorous requirements of analytical laboratories
Browse HDPE bottles
LabServ PET containers and bottles for water sampling
LabServ PET containers and bottles
  • Square containers available in 200mL to 1000mL
  • Bottles available in 250mL and 500mL
  • Gamma sterilised
  • Pre-dosed with sodium thiosulphate
  • Sodium thiosulphate labelled
  • CoA and CoC available
  • Blue wadded cap for square containers
  • Bottles come with HDPE screw caps
Browse PET containers     Browse PET bottles
LabServ HDPE general purpose bottles
LabServ HDPE bottles
  • Round bottles available from 60mL to 500mL
  • Narrow mouth – Neck finish 28mm
  • Opaque
  • Bottle only and bottle and cap available
  • Screw cap – available separately
  • All bottles manufactured in a controlled environment to minimize risk of contamination
  • HDPE raw material is selected to meet rigorous requirements of analytical laboratories
Browse HDPE bottles
New LabServ plasticware digital catalogue out now
This catalogue covers a wide range of LabServ specialised disposable and reusable plasticware including bottles, vials, tubes, containers, water collection, microbiology, and general labware. Get your digital copy today.
Download catalogue
LabServ Plasticware
LabServ custom laboratory plasticware
LabServ custom laboratory plasticware
Whether you have a one-time project or ongoing production needs, talk to our team about our custom plastic product capabilities. We have options to customise your cap colours, labels, and specifications*.
Let LabServ be your success, helping you to design and produce everything you need to develop your custom product into reality.
*Custom manufacturing dependent on volume quantities
Request custom quote
Recycling LabServ products and packaging
The recycling process involves sorting plastic products by their resin type for recovery and then processing them to produce recycled resins for use as raw materials within other industries.
To make this easy, an international system has been developed to identify the types of plastics that can be recycled.
Our PET and HDPE containers and bottles fall within this system.*
Recycling
SymbolType of PlasticProperties
PET recycle icon
PET, PETE: Polyethylene Terephthalate
Clear, tough solvent resistant, barrier to gas and moisture, softens at 80°C
HDPE recycle icon
HDPE, PE HD: High Density Polyethlene
Hard to semi flexible, resistant to chemicals and moisture, waxy surface, opaque, softens at 75°C, easily coloured, processed and formed
*CAUTION: The recycling symbol does not imply that the product may be automatically recycled or disposed of in the general waste stream after use. Be sure to follow appropriate decontamination and disposal procedures whenever a product has been in contact with hazardous and/or infectious materials.