Sample Convenience: One Bag for Every Step
September 2022
Whether it’s for testing food, pharmaceuticals, soil or stormwater, taking samples is key to various industrial and public health safety workflows.
The challenge with samples is that once collected, they need to remain sterile and uncompromised on their way to processing. Additionally, they often need to be stored safely, which can be an issue when there’s hundreds of sample bottles in stow.
As part of Thermo Fisher Scientific’s commitment to providing complete end-to-end workflow solutions, having a sampling bag solution that can be used throughout the entire sampling process has been an important inclusion in their supplier partner product portfolio.
The Whirl-Pak™ sterilised sample bags address challenges around sample protection by safeguarding samples from the point of collection, through transit, processing and storage. The bags take their name from the unique leak-proof closure which allows the user to ‘whirl’ their sample and keep it sealed in the same way as a plastic container.
Since the brand’s inception over 60 years ago, Whirl-Pak have been trusted by professionals in the scientific community, offering guaranteed sterility and manufactured to meet the quality management systems standard ISO9001:2015.
The Whirl-Pak sample bags are not only leak-proof but have puncture-proof closure tabs and are made with high strength plastic film. Importantly, one single bag can be used in every step of the process, reducing the risk of sample damage or loss.
In comparison to other sample containers the Whirl-Pak require little storage space. For example, 500 Whirl-Pak bags take up only a fraction of the space that 500 sample bottles would in a lab or specialised storage unit. The bags can also be frozen to liquid nitrogen temperatures as low as -210°C.
Whirl-Pak sample bag features and benefits: 
  • Easy to open: They have Tyvek® non-slip pull tabs for easy opening which also prevents contamination from finger contact when opening.
  • Leak-proof closure: Whirling or folding the bag over three times secures the closure and creates a leak-proof sampling container.
  • Reduced storage and shipping: Due to their slim size and durability, Whirl-Pak use less shelf space and can save on both shipping and storage costs in comparison with other sample containers.
  • Guaranteed, traceable sterility: All Whirl-Pak bags and sealed cartons are sterilised (+1) after manufacturing at an independent ISO-certified facility. Sterility certificates are available using the lot number and master number on each box.
  • Puncture-proof closure tabs: The tape extends beyond the wire to eliminate sharp points that could puncture and compromise the sample bag. This also enables the users to keep a sturdy grip while the wire tabs keep the bag open for easy access.
  • High strength film: Uses blended, virgin polyethylene film that’s exceptionally clear and strong.
  • Convenient write-on label strip: Ballpoint pens or pencils can be used to write on the strip for clear labelling and easy sample identification.
  • Continuous perimeter seal: Amplifies protection against leaks.
For more information or to order Whirl-Pak sample bags for your facility, please visit Whirl-Pak™ Standard Sample Bags.
Style Sheet for Global Design System