Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Cerillo™ Microplate Platform is transforming the way you collect your data. The platform is made up of the Stratus Kinetic Microplate Reader, the Canopy, a miniaturised portable wireless data transmitter, and the Labrador Software, creates a complete, portable, and user-friendly system. With automated data-point collection capabilities, this enables the creation of high-resolution microbial growth curves to effectively address analysis and testing needs.
Cerillo Stratus Microplate reader is filled with features, starting with its size; taking up less bench space to maximise your available bench space for other resources. The device is portable, creating a testing-environment adaptable solution, allowing you to take the Stratus outside of the laboratory setting if required. The ability to operate in anaerobic conditions, with heat resistance up to 50°C and low oxygen environments makes this a dynamic solution. Further, it allows for automated data collection with minimal manual user input when sampling data – allowing for more accurate results. Available to purchase with the Stratus is an adaptor, which allows the Stratus to become compatible with shakers.
Using the Labrador software, high-resolution bacterial growth curves are generated, allowing for experiments with minimal user input, longer testing run times (day/night experiments) and a more accurate results curves – providing an expanded scope of results for analysis. Transfer your results directly to your computer via the purchasable accessory, Canopy, or access stored results with the SD card. Take your research to the next level with this advanced device.
This comprehensive range provides a complete offering with the ability to generate research that is more comprehensive and affordable. This novel and innovative range allows you to save space without compromising on quality.