Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Macrophages cultured on a polymer surface that have fused to form a foreign-body giant cell following treatment with interleukin-4. Cells were fixed with 3.7% formaldehyde, treated with RNase A and stained with rhodamine phalloidin (Cat. no. R415) to visualize F-actin, and with YO-PRO®-1 iodide (Cat. no. Y3603) to visualize cell nuclei. Cells were imaged with a Bio-Rad MRC600 confocal laser-scanning microscope. The image is a composite of optical sections taken through the Z-axis of the cell. F-actin (red) is restricted to the periphery of the multinucleated cell and surrounds a cluster of nuclei (green). Image contributed by Kristin DeFife and James M. Anderson, Institute of Pathology, Case Western Reserve University.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›