Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Confocal micrograph illustrating sexual reproduction of the ciliate protist, Tetrahymena thermophila, six hours after mating. After fixation and permeabilization, the cytoskeleton was labeled with an anti-tubulin antibody and subsequently visualized with Texas Red®-X goat anti–mouse IgG antibody (Cat. No. T6390). The macro- and micronuclei were stained with SYTOX® Green nucleic acid stain (Cat. No. S7020). Image contributed by David Asai and Amy Walanski, Purdue University.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›