Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Live bovine pulmonary artery endothelial cells (BPAEC) loaded with red-fluorescent MitoTracker Red CMXRos (M7512), fixed and permeabilized (middle). After fixation and permeabilization, the cells were treated with a cocktail containing two mouse monoclonal antibodies to cytochrome oxidase (the anti–OxPhos Complex IV subunit VIc (A6401) and anti–OxPhos Complex IV subunit I (A6403) antibodies), which were then detected using blue-fluorescent Alexa Fluor 350 goat anti–mouse IgG antibody (A11045) (left). The image in the right panel is an overlay of the first two images.
A prometaphase muntjac skin fibroblast stained with Alexa Fluor® 350 phalloidin, an anti–a-tubulin antibody and an anti–cdc6 peptide antibody. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›