Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your analyses needs.
View the relevant questions below:
Having problems with your experiment? Visit
Quadrupole isolation is faster because ions must travel through the quadrupole analyzer regardless of the analysis type chosen. As a result, there is little to no time penalty when using the quadrupole for MS2isolation. In such a case, quadrupole transmission efficiency will depend on isolation width, m/z, and the cleanliness of the analyzer. Ion trap isolation is best suited for MS3 experiments, including those performed using multinotch isolation for TMT analysis; these types of isolation events are not possible with the quadrupole analyzer.
Yes. The ion routing multipole has multiple functions. The IRM not only functions as a collision cell for HCD fragmentation, but also as an ion storage-routing device so ions can be routed to the linear ion trap and back to the Orbitrap via the IRM cell.
Yes. The fluoranthene radical anion will need the calibrated ion optic and ion trap parameters for effective transmission and trapping for ETD.
Start by checking each group of calibrations, one at a time. If they fail the check repeatedly, perform calibration.
If any coarse calibrations are performed, the main calibrations must be performed as well.
Upon instrument install, the instrument needs to be calibrated.
Venting/replacing hardware/cleaning, the instrument needs to be calibrated.
Yes. The calibrations are designed to start with the ion optics and conclude at the ion trap. Subsequent calibrations rely on the previous ones being calibrated correctly. In particular, if anything other than mass is recalibrated, pAGC will have to be recalibrated. If eFT is calibrated, then mass must also be recalibrated.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.