Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
We are dedicated to your success. View our expert recommendations for commonly encountered inquiries.
View the relevant questions below:
Beginning your experiment? Visit
AcquireX is available on the Thermo Scientific Orbitrap Ascend, Orbitrap Eclipse, Orbitrap IQ-X, Orbitrap Fusion Lumos, Orbitrap Fusion and Orbitrap ID-X Tribrid MS instruments. In regards to the Orbitrap Fusion and Fusion Lumos instruments, you will need to upgrade to Instrument Control Software Version 3.1 and XCalibur Version 4.2.
No, AcquireX is used for small molecule (ions with a charge state of 2 or less) applications specifically.
AcquireX only works for LC experiments because the algorithm looks for things that elute as peaks specifically.
Speed is impacted by the cleanliness and complexity of your samples as well as the processing speed of the computer.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.