Search Thermo Fisher Scientific
Thermo Scientific Samco liquid handling solutions bring you safety and innovation. Bio-Tite specimen containers, 24-hour urine containers, operating room containers, pathology and general purpose sample containers, analyzer cups, sample cups, test tubes, disposable culture tubes, and centrifuge tubes are also available in a wide variety of standard sizes for clinical, research, and industrial laboratories.
An all plastic cap and vial transport systems for the collection of urine, formalin or fixative fills for histology applications. Popular sizes include 20mL/35mm, 40mL/48mm, 60mL/48mm,90mL/48mm, 90mL/53mm, and 120mL/53mm.
Precision molded and manufactured from premium grade materials, the plastic tubes are chemical-free and biologically inert.
Geared at alleviating mess and exposure, the amber tinted containers are inert tor normal preservatives.
These cups are the perfect choice for autosamplers, serum dilutions, microsampling, and other laboratory applications.
These fine tip pipettes are designed to easily grip with latex gloves, yielding repeatable results that give you the confidence you need.
Made of virgin polypropylene, these containers resist cracking and splitting and are inert to most chemicals, including formaldehyde.