System overview
SureTect Food Safety PCR System
Visibly simple food pathogen and quality testing—simple workflows designed to get results.
Rapidly and accurately detect foodborne pathogens, and other important food safety and quality targets with the Thermo Scientific™ SureTect Food Safety PCR System.
Empower your operations with simple, automatable workflows and rapid reporting to enable faster product release and interventions. Our food safety PCR assays offer detection of the major pathogens and other important targets in food, pet food and animal feed, primary production samples, and samples from environmental manufacturing.
Whether your laboratory is looking to introduce PCR technology for the first time, or you want to expand the PCR testing services you offer, we can help you find the right solution.
Streamlined workflow
Enrichment
 Report/Confirm
Single enrichment step
Sample preparation and lysis
Run, read and report
Confirm PCR positives
8-24 hours
Single enrichment step of 8-24 hours (depending on target and matrix).
20-40 minutes
Fully automated using the SureTect™ Automation Workflow, or manual preparation with direct lysis in the Applied Biosystems™ SimpliAmp™ Thermal Cycler.
~80 minutes
Perform PCR with the Applied Biosystems QuantStudio™ Instrument using the RapidFinder Analysis Software. Report PCR-negative results immediately (results are automatically interpreted).
24 hours
Confirm PCR-positive results, after overnight incubation, using rapid chromogenic culture media methods.
See how we have innovated to improve workflow & handling:
Learn how you can reduce hands-on time whilst increasing accuracy with the SureTect Automation Workflow:
SureTect PCR Assays
Validated for a broad range of targets and matrices according to ISO 16140-2:2016 (AFNOR validation) and AOAC certification requirements, the SureTect PCR Assays provide the ultimate solution for all your foodborne pathogen detection requirements.
  • Pre-dispensed lysis and PCR reagents, reducing handling steps and risk of operational error
  • NEW color-coded frames for lysis and PCR stripe well plates for easy assay target identification
  • NEW clearly oriented lysis and assay plates reducing human error
  • NEW color indicator in Proteinase K Reagent enabling tracking of pipetted wells
  • Thin-walled polypropylene wells ensure superior thermal cycling performance for consistent results run after run
  • Internal Positive Control (IPC) in every well providing confidence in every result
  • NEW pierceable seals to reduce handling steps
  • All assays compatible with both manual and automated workflows
Suretect PCR assays kit
New features of the upgraded SureTect Assays include:
Color-coded assay strip plates
 Color-coded Assay Strip Plates
Easy, safe separation into 8-well strips
Easy, safe separation of Lysis and Assay plates into 8-well strips
Cut strips to required number of tubes
Cut strips to required number of tubes
Color-indicator in Proteinase K Reagent
Color-indicator in Proteinase K Reagent
Efficient plate sealing
Efficient plate sealing
Pierceable lysis tube seals
Pierceable lysis tube seals
SureTect assay list
View the full range of SureTect PCR Assays:
Featured Product
SureCount Salmonella
Salmonella control you can count on
Simultaneously quantify & type with the innovative Thermo Scientific™ SureCount™ Salmonella species, Typhimurium and Enteritidis PCR Kit method. A simple, all-in-one solution designed to give you quantification and serotype results for the most common pathogenic Salmonella strains associated with meat and poultry matrices.
Visit page
Instruments & software
MBD-SureTecct-landing-page-CyBio-FeliX-650x600
Thermo Scientific™ SureTect™ Automation Workflow (using the CyBio™ FeliX)
With the Thermo Scientific SureTect Automation Workflow you can take your food safety testing to the next level with automated lysis and PCR setup. Utilizing the CyBio FeliX Instrument, the automated workflow allows you to reduce hands-on time whilst increasing the accuracy and reproducibility of results.
Fully validated by AFNOR and AOAC for use with all SureTect Assays, the instrument's flexible programs, user-friendly interface and rapid yet precise setup ensure maximum efficiency, whilst the compact design saves valuable bench space.
Time saving
  • Fully automated lysis, PCR setup or complete workflow
  • Rapid 5-minute setup for loading reagents
Intuitive software
  • Pre-programmed runs for quick setup
  • User-friendly interface with no specialist training required
Flexible
  • Up to 6 different assays/96 samples on the same run
  • Integrated heating and cooling block
  • Choose between the CHOICE head for simultaneous 8-sample pipetting or the Head R 96
    for 96-sample pipetting
  • Choose from full or partial plate processing
  • Compact design: 58kg and 650x450x700mm
Accurate and traceable
  • Barcoded sample entry for full traceability
  • Precise pipetting
  • Robust and factory calibrated instrument
Order the SureTect Automation Workflow
Applied BioSystems™ QuantStudio™ 5 Food Safety Real-Time PCR System
The SureTect PCR Assays are validated to run on the QuantStudio 5 Food Safety PCR system using Thermo Scientific RapidFinder Analysis Software for easy test set-up, results interpretation and data analysis.
The QuantStudio 5 Food Safety PCR System is a high-performance benchtop platform, combining ease-of-use and system connectivity with powerful options that enable maximum data utility and control. Benefit from the flexibility of a 6-channel, 96-well, cloud-enabled open platform suitable for running our full range of PCR solutions for food safety, authenticity, and quality testing.
Intuitive and easy-to-use for all levels of experience with touchscreen usability, allowing you to stay connected to your data easily.
  • Multiplex with ease—up to six excitation and six emission filters offer 21 different color combinations, allowing a broad range of detection chemistries and maximum multiplexing
  • Get up and running quickly—factory-calibrated for accuracy, quick installation, and immediate use; pre-optimized protocol templates help minimize training for new users
  • Maximize benchtop space—compact instrument can be configured as a stand-alone or with a computer
  • Open instrument—suitable for all our food safety, authenticity, and quality PCR tests
  • Intuitive, user-friendly software, avoiding subjective interpretation
  • Cloud-enabled, offering the potential for data sharing, analysis, and trending
  • Capacity for processing several hundred samples a day
Order the QuantStudio 5 Food Safety PCR System
Thermo Scientific RapidFinder Analysis Software
An integral part of the SureTect workflow, the RapidFinder Analysis Software is a graphical, easy-to-use tool for food and environmental testing labs to optimize their pathogen detection workflow.
Simple set-up
  • Intuitive sample set up
  • Flexible plate layout including automatic or customisable options
  • Pre-set, optimized thermal cycling conditions per target
Automatic data interpretation
  • Fully automated amplification, detection, data collection, and data analysis
  • Quick interpretation of data with flags, notifications, and prompts
  • Detailed report generation for printing or export
  • Interoperability with LIMS or similar data management tool for importing plate set-up and exporting assay results
  • Automatically generates results (presence or absence of the target)
  • Offers interrogation of results for the whole plate, grouped by sample, or for individual PCR tubes
Order the RapidFinder Analysis Software
Handling tools
Easy-to-use tools to simplify your workflow: designed specifically to enhance each step of the SureTect PCR Assay workflow, the new Thermo Scientific™ SureTect Tools enable ease of handling, reducing hands-on time and human error, providing a superior user handling experience.
 ProductDescriptionCat. No.
PCR Strip Cutting Tool
Cut Lysis and PCR Assay Strips when fewer than 8 tubes are needed.
A55329
PCR Lysis Tube Uncapping Tool
For removal of Domed Strip Caps from Lysis Strips.
A55328
PCR Capping Tool
For capping Lysis and PCR Assay Plate strips.
A55327
96-well PCR Base
96-well base for Lysis and PCR Assay Strip Plates. Use in Lysis and PCR steps.
A55330
MicroAmp adhesive film applicator
Pack of 5; for ease of applying the Pierceable Film to Lysis Plate strips
4333183
PCR Strip Cutting Tool:
Capping Tool with Domed Lysis Tube Caps:
Uncapping Tool with Domed Lysis Tube Caps:
SureTect PCR Workflow video:
SureTect Trays
ProductDescriptionCat. No.
SimpliAmp Strip-Plate Tray
For compatibility and ease of loading SureTect Lysis Tubes into the SimpliAmp Thermal Cycler
A55333
QuantStudio 5 Strip-Plate Tray
For compatibility and ease of loading SureTect PCR Assay Tubes into the QuantStudio 5 PCR instrument
A55332
ABI 7500 Fast Precision Plate Adapter
For compatibility and ease of loading SureTect PCR Assay Tubes into the 7500 FAST PCR instrument
A46347
SimpliAmp Strip Plate Tray Cover
Pack of 10 For covering and securing the Lysis Tubes before pipetting
A55971
Accessories
Each SureTect PCR Assay comes with all the reagents and PCR accessories needed to set up your SureTect PCR Assay tests to run on the QuantStudio 5 Food Safety PCR System. Additional SureTect Accessories are available to order separately, including Armadillo low-profile PCR strip plates for balancing when using the Applied Biosystems 7500 Fast Food Safety PCR System and domed lysis tube caps for use if preferred in place of the pierceable lysis seals.
ProductDescriptionCat. No.
Pierceable lysis seal
Pack of 100 seals
  • 2 included with each kit
  • Alternative option to lysis caps
  • Printings around each well for positioning and orientation
  • Strip perforations
A55331
Lysis tube caps, domed, pack of 120 strips
Pack of 120 strips: Optional domed caps that can be used in place of pierceable seals for the Lysis Tubes
A56895
Armadillo Low-Profile PCR Strip Plate, 96 well, clear
For balancing of 7500FAST plates
AB2696
Ultra clear qPCR caps
Additional PCR caps, if required
AB0866
SureTect workflow improvement starter pack
Get up and running quickly and easily with the new, improved SureTect PCR Assay workflow with the convenient SureTect Workflow Starter Pack containing all of the tools and accessories you need.
ProductDescriptionCat. No.
SureTect Workflow Starter Pack
Contains: 1x PCR Strip Cutting Tool, 1x PCR Strip Capping Tool, 1x PCR Strip Uncapping Tool, 2x 96-well PCR Base, 1x SimpliAmp Strip Plate Tray, 1x QuantStudio 5 Strip Plate Tray, 1x ABI 7500 Fast Precision Plate Adapter, 1x SimpliAmp strip plate tray cover, 1x MicroAmp Adhesive Film Applicator and 120x Lysis tube caps–domed
A56898
Validation & certification
SureTect PCR Assay validated workflows
SureTect PCR Assay workflows are validated for a broad range of matrices according to ISO 16140-2:2016 (AFNOR VALIDATION) and AOAC and fully supported by our dedicated and experienced technical teams.
View certifications
A guide to efficiently implementing a new, alternative food testing method in accordance with ISO 16140 - Part 3
As the global food testing market continues to grow, businesses need faster, more efficient, and cost-effective solutions. That’s where alternative food testing approaches come in, offering rapid, reliable results that align with modern industry demands.
Our comprehensive eBook, "A guide to efficiently implementing a new, alternative food testing method in accordance with ISO 16140 – Part 3" is your roadmap to navigating, selecting and verifying alternative workflows inline with an internationally recognised standard.
Download eBook
Resources
SureTect workflow video
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet for Cards
Style Sheet for Fonts
Style Sheet for Support and Service footer