Search Thermo Fisher Scientific
Complete elemental analysis from sub-ppt to ppm levels, allowing high sample throughput in both applied testing and industrial laboratories.
Advanced interference removal for best-in-class detection limits and accuracy built into a compact benchtop instrument.
Offers isotope ratio analysis with precision, sensitivity, dynamic range and stability for the fields of earth sciences, nuclear sciences, and other disciplines.
Class-leading elimination of interferences for ultra-sensitive, reliable multi-element analyses at trace levels, even without sample preparation.
We offer an intuitive, user-friendly software platform designed to simplify trace elemental analysis workflows and maximize efficiency.
SQ-ICP-MS | TQ-ICP-MS | HR-ICP-MS | MC-ICP-MS | |
---|---|---|---|---|
Features | Combines best-in-class interference removal with speed of analysis for fast and reliable trace elemental analysis | Advanced interference removal and an additional quadrupole to suppress unwanted side reactions | Elimination of interferences for ultra sensitive, reliable multi-element analyses at trace levels, without sample preparation | Combines high-resolution, high-sensitivity and multicollection with innovative amplifier technology for multi-isotope analysis down to trace levels |
Key Technology | Single quadrupole + flatapole design QCell, automatically adjusted low mass cut off Argon Gas Dilution for analysis of high matrix samples | Triple quadrupole + flatapole design QCell Reaction Finder Method Development Assistant for easy method development | Double-focusing magnetic sector field available with either discrete dynode detector or with additional single Faraday detector | Double-focusing magnetic sector field with moveable Faraday collectors; also available with multiple ion-counting devices |
Interference Removal | Kinetic Energy Discrimination (KED), optional reactive gases | Kinetic Energy Discrimination (KED), different reactive gases | Physical interference removal using magnetic and electrostatic fields | |
Applications | High quality, high-throughput analysis in applied testing for environmental, food safety, industrial or pharmaceutical applications; challenging matrices | For applied testing applications or cutting-edge research analyses; suitable for high-matrix samples | Semiconductor, geological and material science laboratories, organic and inorganic solutions or solid matrices | Earth and nuclear sciences, biochemistry, food, forensics and medical sciences that require high-precision isotope |
Size | Benchtop | Benchtop | Floor-standing | Floor-standing |
Building on over 50 years of serving science, the Unity Lab Services team delivers consistent, high-quality service and OEM expertise whenever and wherever you need it. We’ve got your back.
Get superior instrument services and support to help keep your lab up and running—so you can focus on your work. Unity Lab Service plans help maximize your uptime and predict your expenditure. We’ve got your back.
Reduce your lab’s environmental impact with the iCAP RQplus ICP-MS and iSC-65 Autosampler, both recipients of the ACT label from My Green Lab. The ACT label, the standard for determining the environmental impact of laboratory products, helps labs make informed, sustainable purchasing decisions.
Download our eBook for recommendations on getting started, best practices, and overcoming analytical challenges.
* Required field