Search Thermo Fisher Scientific
Thermo Scientific Nalgene Bottle Safety Carriers and Totes help you shuttle chemical bottles around the lab. Safety carriers hold glass reagent bottles securely in place and cushion them from bumps as they are carried around and between labs within your facility. The 4-in-1 Easy Tote and Multi-Bottle Rack hold multiple bottles for easy and efficient toting between work stations.
Safety Bottle Carrier, 2.5L | Safety Bottle Carrier, 4L | Safety Bottle Carrier, 500mL | Multi-Bottle Rack, 500mL | 4-in-1 Easy Tote |
Safely hold; cushions bottle and contains spills | Safely hold; cushions bottle and contains spills | Safely hold; cushions bottle and contains spills | Bottle rack for up to eight 500mL bottles | Open tote for multiple-sized bottles |
![]() | ![]() | ![]() | ![]() | ![]() |
6501-2500 | 6501-4000 | 6505-0010 | DS5996-0871 | 6565-0001 |
Provides some impact protection | Provides some impact protection | Provides some impact protection | No impact protection | Little impact protection |
Bottle completely contained within carrier | Bottle completely contained within carrier | Bottles completely contained within carrier | Open-top rack | Open-top tote |
Holds a 2.5L glass chemical bottle | Holds a 4L glass chemical bottle | Holds 6 500mL glass reagent bottles | Holds 8 500mL reagent bottles | Configurable to hold multiple-sized bottles |
Not autoclavable | Not autoclavable | Not autoclavable | Autoclavable | Not autoclavable |
For assistance choosing products appropriate for your application, please speak with a Nalgene Technical Support Representative team by phone at +1.585.586.8800 or (1.800.625.4327 US toll free), or email your request to
technicalsupport@thermofisher.com.
In Austria, France, Germany, Ireland, Switzerland, and the United Kingdom please contact technical support by phone at +800 1234 9696 (toll free) or +49.6184.90.6321, or email your request to techsupport.labproducts.eu@thermofisher.com.
Regulatory support: For regulatory documentation of product or material claims, please contact Nalgene Regulatory Support at RocRegSupport@thermofisher.com