Search Thermo Fisher Scientific
FluoCells® prepared slide #3 (Cat. no. F24630) contains a mouse kidney section stained with a combination of fluorescent dyes. Alexa Fluor® 488 wheat germ agglutinin (Cat. no. W11261) is a green-fluorescent lectin that was used to label elements of the glomeruli and convoluted tubules. The filamentous actin prevalent in glomeruli and the brush border were stained with red-orange–fluorescent Alexa Fluor® 568 phalloidin (Cat. no. A12380). Finally, the nuclei were stained with the blue-fluorescent DNA stain DAPI (Cat. no. D1306, D3571, D21490). This pseudocolored image was acquired on a Zeiss confocal microscope located at the Institute of Neuroscience, University of Oregon.
1% Agarose gel containing 16S and 23S ribosomal RNA (rRNA). SYBR® Green II RNA gel stain. Go ›
Bovine pulmonary artery endothelial cells (BPAEC). MitoTracker® Red CMXRos, SYTOX® Green nucleic acid stain, biotin-XX goat anti–mouse IgG antibody and Cascade Blue® NeutrAvidin biotin-binding protein. Go ›
Endothelial cells. Go ›
Mouse Anti-Alpha Tubulin Monoclonal Antibody (Cat. No. A11126) Go ›
Simultaneous detection of expression of five genes in a whole mount Drosophila embryo by fluorescence in situ hybridization (FISH) with five RNA probes Go ›
Multicolor fluorescence analysis of muntjac fibroblasts. Go ›