Search Thermo Fisher Scientific
A mixture of live and ethanol-killed bovine pulmonary artery epithelial cells stained with the reagents in our LIVE/DEAD® Cell Viability/Cytotoxicity Assay Kit (L3224). Live cells fluoresce bright green, whereas dead cells with compromised membranes fluoresce red-orange.
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
BPAE cells fixed and permeabilized using the Image-iT® Fixation/Permeabilization Kit. Go ›
Live cell imaging with CellLight™ reagents. Go ›
Live cells transduced with Organelle Lights™ or Cellular Lights™ reagents. Go ›