The SpeciMAX Dx Stabilized Saliva Collection Kit is intended for the collection and preservation of saliva specimens as an aid in nucleic acid-based Polymerase Chain Reaction (PCR) for In Vitro Diagnostic purposes. The SpeciMAX Dx Stabilized Saliva Collection Kit is intended for use in the presence of healthcare professionals to collect patient saliva samples.
PLEASE NOTE: This product is only available in select countries including UK, Ireland, Malta, Germany, Switzerland, Belgium, Austria, Luxembourg, Spain, Italy, France and Poland. Regulatory requirements vary by country; products may not be available in your geographic area.
Key features of the SpeciMAX Dx Stabilized Saliva Collection Kit
Helps keep staff safe
Virus-inactivating and nucleic acid–stabilizing solution for common enveloped respiratory viruses
Nonhazardous stabilization solution compatible with all disinfection protocols—no reaction with bleach (no guanidinium or ethanol)
Standardized processing
6 mL standardized tubes and caps are compatible with automation racks and decappers for downstream workflows; eliminates manual, labor-intensive saliva pipetting when using a liquid handler
Ease of use
Spill-resistant, single-use funnel and collection tube allow for convenient, noninvasive supervised collection
Optimization for saliva collection and sample accessioning, individually packaged, and 95 kPa-compliant
Helps save time, money, and storage space
Click image to enlarge Figure 1. Improve your diagnostic testing efficiency.
Downstream saliva sample workflows
Watch: Saliva collection with the Thermo Scientific SpeciMAX Dx Stabilized Saliva Collection Kit
Technical specifications of the SpeciMAX Dx Stabilized Saliva Collection Kit
Intended use
The SpeciMAX Dx Stabilized Saliva Collection Kit is intended for the collection and preservation of saliva specimens as an aid in nucleic acid-based Polymerase Chain Reaction (PCR) for In Vitro Diagnostic purposes.
The SpeciMAX Dx Stabilized Saliva Collection Kit is intended for use in the presence of healthcare professionals to collect patient saliva samples.
Included in kit
Spill resistant funnel, pre-filled capped 6 mL tube with stabilization