Search Thermo Fisher Scientific
Enzyme-linked immunosorbent assays (ELISAs) are plate-based assays for detecting and quantifying a specific protein in a complex mixture. The detection and quantification of target-specific protein in a sandwich ELISA is accomplished by using highly specific antibodies that immobilizes the target protein (antigen) to the plate and indirectly detects the presence of the target protein. This type of capture assay is called a sandwich assay because the analyte being measured is bound between two primary antibodies, each detecting a different epitope of the antigen—the capture antibody and the detection antibody. There are many different types of components, such as substrates, plates, and other reagents, that the choices can sometimes be overwhelming. We’ve made general suggestions in the protocols below, and you can also visit our ELISA builder online selection guide that asks a few simple questions about your specific ELISA and then recommends all the components you’ll need, from plates to stop solution.
Search available antibody pairs and ELISA kits Browse available ELISA reagents
Find protocols below for a standard sandwich ELISA using a 96-well plate for the detection techniques—colorimetric (chromogenic), chemiluminescent, and fluorescent detection.
Overall procedure
This protocol represents an example colorimetric sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
If alkaline phosphatase (AP) is to be used instead of HRP for the enzyme conjugate, an AP-specific substrate must be used. Substitute TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (Thermo Scientific 1-Step PNPP Substrate Solution, Cat. No. 37621). Incubate at room temperature for 15–30 minutes. Substitute the stop solution with 50 µL 2 N NaOH to stop the reaction. Measure absorbance of each well at 405 nm.
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
This protocol represents an example of a chemiluminescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A luminol-based substrate is used for detection.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
If alkaline phosphatase (AP) is to be used instead of HRP for the enzyme conjugate, an AP-specific substrate must be used. Substitute TMB substrate solution in step 15 with p-nitrophenyl phosphate (PNPP) (Thermo Scientific 1-Step PNPP Substrate Solution, Cat. No. 37621). Incubate at room temperature for 15–30 minutes. Substitute the stop solution with 50 µL 2 N NaOH to stop the reaction. Measure absorbance of each well at 405 nm.
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
This protocol represents an example of a fluorescent sandwich ELISA using direct detection with an HRP-conjugated antibody or indirect detection with a biotinylated antibody and streptavidin-HRP. A fluorogenic peroxidase substrate is used for detection.
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
|
With a fluorophore-conjugated detection antibody, no enzyme conjugate or substrate is required. The plate fluorescence units can be measured directly after step 11. The working concentration of labeled antibody or protein is typically 2–4 μg/mL.
When immobilizing the antigen-containing sample directly to the plate, there is no need for a capture antibody. Different concentrations of the sample should be prepared in coating buffer and identical volumes added directly to the plate. The rest of the protocol should be performed as previously described using a detection antibody and enzyme conjugate plus substrate.
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
The following tables provide recommended ranges for different ELISA components. Concentrations are guidelines only; for best results optimize each component individually.
Recommended starting concentration ranges for coating and detection antibodies for ELISA optimization. The use of non-purified antibodies will work but may result in higher background. It is generally recommended to use affinity-purified antibodies for optimal signal-to-noise ratio.
Source | Coating antibody | Detection antibody |
Polyclonal serum | 5–15 μg/mL | 1–10 μg/mL |
Crude ascites | 5–15 μg/mL | 1–10 μg/mL |
Affinity-purified polyclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
Affinity-purified monoclonal antibody | 1–12 μg/mL | 0.5–5 μg/mL |
Recommended detection antibody concentration for ELISA in different systems. Check the user guide for the substrate as it may recommend a more defined concentration range for the enzyme conjugate.
Enzyme | System | Concentration |
---|---|---|
HRP | Colorimetric | 20–200 ng/mL |
Chemifluorescent | 25–50 ng/mL | |
Chemiluminescent | 10–100 ng/mL | |
AP | Colorimetric | 100–200 ng/mL |
Chemiluminescent | 40–200 ng/mL |
When using a non-biotinylated detection antibody followed by an enzyme-labeled secondary antibody, there will be slightly less amplification of enzyme signal compared to using a biotinylated detection antibody with streptavidin-HRP. Therefore, it may be necessary to use a slightly higher concentration of secondary antibody-enzyme conjugate than one would normally use for a streptavidin-enzyme conjugate.
For Research Use Only. Not for use in diagnostic procedures.