Search Thermo Fisher Scientific
Water quality is critical to the health of our communities and environment. Our process analytical systems and equipment help you tackle your water application challenges head on. From drinking water safety to wastewater treatment processes, you can rely on our solutions to deliver value and help you achieve your water analysis goals.
Monitor water quality in a broad range of industries: power generation, wastewater treatment, drinking water, food and beverage, chemical and petrochemical, and pulp and paper.
Early calcium detection maximizes process effectiveness. Producing the highest quality of feedwater is important, whether it's for high-purity steam for power generation or for the best-tasting bottled water for consumers. Monitoring and controlling the level of calcium hardness is a big contributing factor to water quality.
Managing corrosion in steam generating systems remains a constant challenge, costing the power industry billions of dollars each year. Measuring ammonia levels in real time helps improve boiler reliability and maximizes power production efficiency.
Eliminate any threats to the safety and efficiency of your steam turbine components. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification and peace of mind. Our products are your best defense against the costly effects of corrosion, providing rapid chloride detection.
Being able to closely monitor chlorine levels is critical for facilities in the municipal drinking water, wastewater treatment, and food and beverage markets. The Thermo Scientific™ Orion™ Chlorine XP Water Quality Analyzer accurately monitors chlorine levels without the burden of frequent maintenance, cleaning, and high reagent usage.
Proven measurement systems for the most demanding environments. Measure conductivity accurately and reliably with Thermo Scientific™ industrial conductivity sensors designed to withstand the environment. Our industrial line of durable, high-performance conductivity probes and monitors is designed for a wide range of conductivity applications, from ultrapure water to final water treatment.
Measure dissolved gaseous oxygen accurately and reliably with industrial sensors designed to withstand the environment. Our industrial line of durable, high-performance Thermo Scientific™ dissolved oxygen probes and monitors is designed for a wide range of conductivity applications, from ultrapure water to final water treatment.
Although fluoride occurs naturally in all types of water, it is typically added to drinking water to maintain dental health. To be effective, the level of fluoride in drinking water needs to be strictly controlled. To protect children from dental fluorosis, the U.S. Environmental Protection Agency (EPA) has set an enforceable drinking water standard for fluoride of 4mg/L and a secondary fluoride standard of 2mg/L.
Our products are designed to help you save time and money while achieving fast, accurate measurements in a variety of samples. With minimal maintenance and a longer lifetime, our line of online pH sensors offers superior performance, reliability, and longevity for a variety of applications, including pure water.
Experience peace-of-mind knowing that your equipment is being protected by controlled dosing of phosphates in your water. Our phosphate analyzer and accessories offer continuous and online phosphate monitoring, critical components in the water quality monitoring process.
Online water measurements have become increasingly important in meeting strict regulatory requirements for contamination levels in water. Selecting top-quality, proven process analyzers ensures that your manufacturing facility or plant is protected at all times.
Continuous, online measurement of reactive silica is key to protecting mission-critical systems. Therefore, we designed an online silica analyzer that helps you protect your equipment from the costly effects of corrosion. The optimized design of our silica analyzer drastically reduces the reagent consumption rate, thus reducing the overall cost of ownership.
Sodium monitoring is a critical measurement in the steam and power generation industry. Being able to continually analyze and monitor the level of sodium in your process water helps ensure the plant operates at maximum efficiency. Our line of proven sodium analyzers are amongst the most reliable solutions in the industry.
Turbidity is an important parameter in many manufacturing operations. It may indicate the presence of bacteria, pathogens or particles in drinking water applications. In industrial applications, turbidity is a parameter to measure the effectiveness of water treatment. Our turbidity sensors and accessories help you save time, space, and money, while achieving your turbidity monitoring needs with confidence.