Search Thermo Fisher Scientific
Find a variety of formats and sizes of atmosphere generation supplies to help you maximize incubation space and organism growth at the same time. All of our environmental solutions are designed to deliver the anaerobic, microaerophilic or CO2 conditions you need for microorganisms to thrive.
Our comprehensive AGS product range includes a choice of jars and containers as well as simple, clean and easy-to-use AnaeroGen™, CampyGen™ and CO2Gen™ sachets which create an optimal environment for growth, without the need for the addition of water, catalyst or hydrogen.
Atmosphere Generation Systems are versatile systems available in a standard format (boxes and jars) or compact plastic pouch format. This system can be tailored to your needs and can accommodate small or large numbers of plates. The system is suitable for the transportation, culture, selective isolation, and susceptibility testing of non-aerobic organisms.
Our comprehensive AGS product range includes a choice of jars and containers as well as simple, clean and easy-to-use AnaeroGen™, CampyGen™ and CO2Gen™ sachets which create an optimal environment for growth, without the need for the addition of water, catalyst or hydrogen.
Atmosphere Generation Systems are versatile systems available in a standard format (boxes and jars) or compact plastic pouch format. This system can be tailored to your needs and can accommodate small or large numbers of plates. The system is suitable for the transportation, culture, selective isolation, and susceptibility testing of non-aerobic organisms.
Whether you are starting your atmospheric generation journey, expanding your catalog, or looking for replacement parts, use our comprehensive list to find the accessories you need.