Search Thermo Fisher Scientific
We offer an unmatched portfolio of Thermo Scientific Nunc Roller Bottles. The choice is yours with options available in both polystyrene (PS) and polyethylene terephthalate (PETG), smooth and expanded surface formats, enabling you to find the optimal substrate for culturing your cells.
Assured performance and compliance
Superior manufacturing
All Thermo Scientific Nunc Roller Bottles are manufactured in ISO 13485-2010–registered facilities and ISO Class 7 clean rooms
Minimize validation of cell culture processes
Save time and optimization steps during scale-up from plates to flasks to roller bottles by choosing a family of products that utilize Polystyrene (PS) materials. PS Roller Bottles are available with a smooth surface with an industry-standard growth area of 850 cm2 or a pleated surface of 1,450 cm2 that increases the growth area by over 40%. The unique vertical pleats make emptying easier and provide less product retention. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet 10-6 SAL.
Every lot is cell culture performance–tested and certified nonpyrogenic at a level of <0.1 EU/mL per USP <85>. Nunc PS Roller Bottles have easy on/off closures that help prevent mis-threading, while providing a secure seal.
Increase cell growth and product yield without increasing labor
Thermo Scientific Nunc Polyethylene Terephthalate Glycol (PETG) Roller Bottles are molded in shatter-resistant PETG and are available in both smooth and pleated surfaces. Growth areas ranging from 1,050 cm² to 4,200 cm² help enable increased cell expansion and product yield without investing in additional equipment or increased labor. PETG can be frozen to –40°C for freeze-thaw release protocols. Lot numbers are located on every bottle for full traceability. Nunc PETG Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Every lot is cell culture performance–tested and certified nonpyrogenic at a level of <0.5 EU/mL per USP <85>.
Assured performance and compliance
Superior manufacturing
All Thermo Scientific Nunc Roller Bottles are manufactured in ISO 13485-2010–registered facilities and ISO Class 7 clean rooms
Minimize validation of cell culture processes
Save time and optimization steps during scale-up from plates to flasks to roller bottles by choosing a family of products that utilize Polystyrene (PS) materials. PS Roller Bottles are available with a smooth surface with an industry-standard growth area of 850 cm2 or a pleated surface of 1,450 cm2 that increases the growth area by over 40%. The unique vertical pleats make emptying easier and provide less product retention. Nunc PS Roller Bottles are sterilized by gamma irradiation and meet 10-6 SAL.
Every lot is cell culture performance–tested and certified nonpyrogenic at a level of <0.1 EU/mL per USP <85>. Nunc PS Roller Bottles have easy on/off closures that help prevent mis-threading, while providing a secure seal.
Increase cell growth and product yield without increasing labor
Thermo Scientific Nunc Polyethylene Terephthalate Glycol (PETG) Roller Bottles are molded in shatter-resistant PETG and are available in both smooth and pleated surfaces. Growth areas ranging from 1,050 cm² to 4,200 cm² help enable increased cell expansion and product yield without investing in additional equipment or increased labor. PETG can be frozen to –40°C for freeze-thaw release protocols. Lot numbers are located on every bottle for full traceability. Nunc PETG Roller Bottles are sterilized by gamma irradiation and meet sterility assurance level (SAL) 10-6.
Every lot is cell culture performance–tested and certified nonpyrogenic at a level of <0.5 EU/mL per USP <85>.
Brochure:Nunc Roller Bottles
Application Note: Rapid, serum-free, high-yield antibody production in Nunc PETG Roller Bottles
Application Note:Nunc Roller Bottles excel with ST, Vero, and MDBK cells
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing: