Search Thermo Fisher Scientific
We offer an array of dissolved oxygen measurement instrumentation and accessories designed to meet your standards for performance and price. Our dissolved oxygen meters, probes, solutions, and accessories are designed to offer maximum repeatability and accuracy in your dissolved oxygen readings.
Browse our complete portfolio or let one of our experts help you make a selection.
Air can be regarded as having a constant percentage (approximately 20.9%) of oxygen. Wherever air comes in contact with water, the oxygen in the air will dissolve in the water. The amount of oxygen that dissolves in the water depends on many factors: whether there is adequate time and adequate mixing to fully saturate the water, the water temperature, the air pressure, the salt content of the water, and whether there are substances in the water which consume the oxygen. Since the oxygen content is important for many biological and chemical processes, measurement of the amount of oxygen actually dissolved in a water sample is of great importance.
Applications for dissolved oxygen measurement include processes where the amount of oxygen affects a reaction rate or process efficiency, or indicates an environmental condition. Some important applications include wastewater treatment, wine production, bio-reactions, and environmental water monitoring. Thermo Scientific Orion DO instruments have the flexibility to use polargraphic and optical technology for DO measurement needs meeting regulatory requirements. Membrane oxygen electrodes are the most widely used and accepted method for real-time measurements of oxygen in water. Learn more about how Thermo Scientific instruments can help you meet your regulatory needs.
A bottle of wine’s oxygen content has a great effect on its quality, stability, and longevity. This is why monitoring and controlling the oxygen incorporation at different stages of the wine-making and bottling process is becoming a growing concern for wineries. Although oxygen is a part of the wine’s natural aging process, adverse levels can cause discoloration to white wines and flavor degradation to both white and red varietals. To ensure the consumer is getting the highest-quality product, measuring the concentration of molecular oxygen of wine after bottling is very important.
Using an Orion Star A223 RDO/DO portable meter with an RDO optical dissolved oxygen sensor enables wineries to continually produce high-quality wines. Because the RDO sensor allows the wine to be measured directly in the bottle, dissolved oxygen measurements can be made with speed and accuracy. The speed, accuracy, and precision of the RDO sensor is equivalent or superior to current DO measurement techniques.
Download the application note on measuring dissolved oxygen in wine
DO solubility in saturated water at any one temperature, barometric pressure, and salinity has been measured and recorded by scientists over the years. The information has then been used in programming modern recording instruments. Water holds less oxygen at low barometric pressure (high altitude) and at high salinity.
Dissolved oxygen levels can be measured at altitudes of 4,500 meters or above sea level utilizing advanced instrumentation with particular design features. The saturated dissolved oxygen (DO) level of a water sample is dependent on a number of factors, including:
The oxygen partial pressure of the sample as measured by the DO sensor is converted into concentration terms (mg/L) by using the solubility value of oxygen at the specific atmospheric pressure, temperature, and salinity.
Advanced instrumentation, like Thermo Scientific™ dissolved oxygen meters, are designed with components that measure these factors and use them to determine readings. Thermo Scientific™ Orion™ Versa Star™, Thermo Scientific™ Orion Star™ A200 series and Orion Star A300 series dissolved oxygen meters have an internal barometer that is used to measure the exact barometric pressure at the point of measurement. No matter what your altitude, you’ll have confidence that your readings are reliable.
Meeting regulatory requirements often goes hand in hand with dissolved oxygen measurements, and it’s critical that you have instruments and electrodes that allow you to stay complaint with these industry standards. Below is a list of international standards for regulatory compliance requirements for a variety of industries.
International standards for regulatory compliance requirements:
Parameter | EPA | SM | ASTM | ISO | China MEP | USP | EP | AOAC | Other |
---|---|---|---|---|---|---|---|---|---|
pH | EPA 150.1 | 4500-H+(B) | D1293 (A,B) | 10523 | GB/T 6920 | <791> | 2.2.3 | 973.41 | FDA - 21 CFR 114.90 |
pH of Soil/Solid Waste | SW-846 9040C & 9045D | D4972 | 10390 | GB/T15555 | 994.16 - 994.18 | USDA NRCS | |||
Conductivity | EPA 120.1; SW-846 9050A | 2510 (B) | D1125 (A) | 7888 | n/a | <645> | 2.2.38 | 973.40 | PSS-78 (salinity) |
Dissolved Oxygen | 4500-O (G) | D888 (B, C) | 17289; 5814 | HJ506 | |||||
BOD | 5210 (B) | 5815 | HJ505 | 973.44 | |||||
Fluoride | SW-846 9214 | 4500-F (C ) | D1179 (B) | 10359 | GB7484 | ||||
Ammonia | 4500-NH3 (D, E) | D1426 (B) | 6778 | ||||||
Nitrate | Orion 601; SW-846 9210A | 4500-NO3 (D) | |||||||
Sulfide | SW-846 9215 | 4500-S2 (G) | D4658 | ||||||
Cyanide | SW-846 9213 | 4500-CN (F) | D2036 (A) | ||||||
Chloride | SW-846 9212 | 4500-Cl (D) | D512 (C) | GB11896 | |||||
Bromide | SW-846 9211 |
SM: Standard Methods for the Examination of Water and Wastewater. APHA, AWWA, WEF. | |
ASTM: American Society for the Testing and Materials | |
ISO: International Organization for Standardization | |
USP: United States Pharmacopeia | |
EP: European Pharmacopeia | |
China MEP: China Ministry of Environmental Protection | |
AOAC: Association of Official Analytical Chemists | |
NRCS: Natural Resources Conservation Service | |
PSS-78: Practical Salinity Scale of 1978 | |
FDA: U.S. Food and Drug Administration | |
USDA: United Stated Department of Agriculture |
We offer a range of dissolved oxygen (DO) measurement instrumentation and accessories designed to meet your standards for performance and price. Our DO meters, DO probes, and standards are designed to offer maximum repeatability and accuracy in your DO readings.
The accuracy and reliability of your dissolved oxygen measurements depend on the instrumentation you use. Learn more about how you can identify the best match based on your required features, performance, specifications, and budget
|
We offer probes, storage solutions, cleaning solutions, kits, and filling solutions for your DO measurement needs.
|
Knowledge Base: Certificates of Analysis and SDS for Orion, ROSS, AQUAfast, AquaMate, and AquaSensors products.
Orion Meters Software and Upgrades
Thermo Scientific Orion benchtop and portable meters are designed to help deliver accurate, consistent results, day after day. Boost productivity by upgrading your software today.