Search Thermo Fisher Scientific
Thermo Scientific Custom Peptide synthesis service offers custom peptides with a broad range of peptide modifications to help meet a variety of research needs. Peptides between 2 and 110 amino acids in length, a range of scales (1 mg–1 kg+) and purity (crude to >98%) are available.
Peptide length | 2-110* amino acids, L- or D-isoforms |
Manufacturing scale | 1 mg–1 kg+ ** |
Purity options | From crude to >98% pure |
Quality control | MS for crude peptides, MS and analytical HPLC for all other grades of purity |
Production time | 2–3 weeks |
Modifications | Extensive list of modifications and labels, including heavy peptides (isotope-labeled) |
Conjugation | Custom conjugation available (e.g., carrier protein-peptide conjugates and multiple antigen peptides, MAPs) |
* Not all peptides will be available in lengths of 110 due to sequence composition.
** Greater amounts available upon request.
Thermo Scientific Custom Peptide Synthesis Service offers numerous options for purity levels, modifications, and formats for your synthetic peptide order, giving you the flexibility to meet your research needs. Peptide synthesis is performed using the latest Fmoc solid-phase technology and peptides are purified by reversed-phase HPLC. Peptide sequence is confirmed by MS. Peptides with guaranteed purity are additionally characterized using analytical HPLC to determine the final purity.
We offer a wide range of N-terminal, C-terminal, and other modifications with our standard peptide service. Custom conjugation services (6−25 amino acids) are available, including conjugation to carrier proteins (KLH, BSA, OVA, or Thermo Scientific Imject Blue Carrier Protein) and MAPs.
Type | Name | Abbreviation or alternative name | Position within sequence | Graphical representation of available positions |
---|---|---|---|---|
Misc | 1-Adamantanecarbonyl - | 1-Adamantanecarbonyl-NH | N-term | ■□□□...□□□□ |
Misc | 2,4,6-Trinitrophenyl - TNP | TNP-NH | N-term | ■□□□...□□□□ |
Misc | 2-Furoyl - | 2-Furoyl-NH | N-term | ■□□□...□□□□ |
Misc | 2-Naphtoic acid - | Naphtyl-NH | N-term | ■□□□...□□□□ |
Misc | 3-(Maleimido)propionyl - | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | 3,5-Diiodo-Tyrosine - | [Y(I)2] | Internal | ■■■■...■■■■ |
Misc | 3-Chloro-Tyrosine - | [Y(Cl)] | Internal | ■■■■...■■■■ |
Misc | 3-nitro-2-pyridylthio-L-Cys - | [C(Npys)] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Indole - | [3-Nitro-Indole] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Pyrrol - | [3-Nitro-Pyrrol] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Tyrosine - | [Y(NO2)] | Internal | ■■■■...■■■■ |
Misc | 4-Methylthio-2-oxobutanoic ac. - | [H3C-S-CH2-CH2-CO-CO-NH] | N-term | ■□□□...□□□□ |
Misc | 4-nitrobenzo-2-oxa-1,3-diazole - NBD | NBD-NH | N-term | ■□□□...□□□□ |
Misc | 4-Pentynoic acid - | 4-Pentynoic acid-NH | N-term | ■□□□...□□□□ |
Misc | 4-Phenylazophenol - O-PAP | CO-PAP | C-term | □□□□...□□□■ |
Misc | 6-Bromo-D/L-Tryptophan - | [D/L-W(Br)] | Internal | ■■■■...■■■■ |
Misc | 6-Cloro-Tryptophan - | [W(Cl)] | Internal | ■■■■...■■■■ |
Misc | Acetylation - | Acetyl-NH | N-term | ■□□□...□□□□ |
Misc | Aldehyde - | CHO | C-term | □□□□...□□□■ |
Misc | Amidation - | CONH2 | C-term | □□□□...□□□■ |
Misc | Amino nitrophenyl prop. acid - | [Amino nitrophenyl prop. acid] | Internal | ■■■■...■■■■ |
Misc | Amino-nitrophenyl propion. ac. - ANP-linker | [ANP] | Internal | ■■■■...■■■■ |
Misc | Aminooxyacetyl - | H2N-O-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Benzoyl - Bz | Benzoyl-NH | N-term | ■□□□...□□□□ |
Misc | Benzyloxycarbonyl - Cbz, Z | Z-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - | Biotin-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - Ethylenediamin-Biotin | CO-NH-CH2-CH2-NH-Biotin | C-term | □□□□...□□□■ |
Misc | Biotin-SS-linker - | Biotin-SS-NH | N-term | ■□□□...□□□□ |
Misc | Bis(aminoethyl)ethylene glycol - | CO-NH-(CH2-CH2-O)2-CH2-CH2-NH2 | C-term | □□□□...□□□■ |
Misc | Brom acetic acid - | BrCH2CO-NH | N-term | ■□□□...□□□□ |
Misc | Chlormethylketon - CMK | CMK | C-term | □□□□...□□□■ |
Misc | Cys(4-Methoxytrityl) - Cys(MMT) | [C(Mmt)] | Internal | ■■■■...■■■■ |
Misc | Cys(Acetamidomethyl) - Cys(Acm) | [C(Acm)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carbamidomethyl) - Cys(CAM) / Cys(CH2CONH2) | [C(Cam)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carboxymethyl) - Cys(CMC) / Cys(CH2COOH) | [C(Cmc)] | Internal | ■■■■...■■■■ |
Misc | Cys(di(palmitoyloxy)-propyl) - Cys(Pam2) | [C(Pam2)] | Internal | ■■■■...■■■■ |
Misc | Cys(Farnesyl) - | [C(Farnesyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(HRP) - Cys(horseradish peroxidase) | [C(HRP)] | Internal | ■■■■...■■■■ |
Misc | Cys(MTSL) - | [C(MTSL)] | Internal | ■■■■...■■■■ |
Misc | Cys(Myristyl) - Cys(CH2(CH2)12CH3) | [C(Myristyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(Palmitoyl) - | [C(Palm)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Butyl) - | [C(S-But)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Ethyl) - | [C(S-Et)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isobutyl) - | [C(S-Isobut)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isopropyl) - | [C(S-Isoprop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Methyl) - | [C(S-Me)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Pentyl) - | [C(S-Pentyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Propyl) - | [C(S-Prop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-tert-Butyl) - | [C(S-tert-But] | Internal | ■■■■...■■■■ |
Misc | Cysteamide - | Cysteamide | C-term | □□□□...□□□■ |
Misc | Decanoyl - | Decanoyl-NH | N-term | ■□□□...□□□□ |
Misc | Desthiobiotin - | Desthiobiotin-NH | N-term | ■□□□...□□□□ |
Misc | Digoxigenin - | Digoxigenin-NH | N-term | ■□□□...□□□□ |
Misc | DOTA - | [DOTA] | N-term | ■□□□...□□□□ |
Misc | Ethylamide - N-Ethyl | CONH(C2H5) | C-term | □□□□...□□□■ |
Misc | Ethylester - O-Ethyl | COOEt | C-term | □□□□...□□□■ |
Misc | Fmoc - | Fmoc-NH | N-term | ■□□□...□□□□ |
Misc | Formyl - | Formyl-NH | N-term | ■□□□...□□□□ |
Misc | Glu(Arg) - Arg at side chain of Glu | [Glu(Arg)] | Internal | ■■■■...■■■■ |
Misc | Homoserine lactone - | [Homoser lactone] | C-term | □□□□...□□□■ |
Misc | HRP - | CO-NH -HRP | C-term | □□□□...□□□■ |
Misc | HRP - | HRP-NH | N-term | ■□□□...□□□□ |
Misc | Hydrazide - | CO-NH -NH2 | C-term | □□□□...□□□■ |
Misc | Hydrazine-succinyl - | H2N-NH-CO-CH2-CH2-CO-NH- | N-term | ■□□□...□□□□ |
Misc | Iodoacetyl - | IH2C-CO- | N-term | ■□□□...□□□□ |
Misc | Lys(2-Aminooxy acetic acid) - Lys(Aoa) | [K(Aoa)] | Internal | ■■■■...■■■■ |
Misc | Lys(Acetyl) - | [K(Ac)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-Biotin) - | [K(Ahx-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-SS-Biotin) - | [K(Ahx-SS-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Biotin) - | [K(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Boc) - | [K(Boc)] | Internal | ■■■■...■■■■ |
Misc | Lys(Butyryl) - | [K(Butyryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Desthiobiotin) - | [K(Desthiobiotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Digoxigenin] - | [K(Digoxigenin)] | Internal | ■■■■...■■■■ |
Misc | Lys(DST-PEG 2-Biotin) - | [K(DST-PEG 2-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ferrocene) - | [K(Ferrocene)] | Internal | ■■■■...■■■■ |
Misc | Lys(Formyl) - | [K(Formyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(GG) - Ubiquitin | [K(GG)] | Internal | ■■■■...■■■■ |
Misc | Lys(GGRLR) - | [K(GGRLR)] | Internal | ■■■■...■■■■ |
Misc | Lys(Glutaryl) - | [K(Glutaryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Lipoyl) - | [K(Lipoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(NIP) - | [K(NIP)] | Internal | ■■■■...■■■■ |
Misc | Lys(Palmitoyl) - | [K(Palmitoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Propionyl) - | [K(Propionyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Pyrofolate) - | [K(Pyrofolate)] | Internal | ■■■■...■■■■ |
Misc | Lys(Stearyl) - | [K(Stearyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Succinyl) - | [K(Succinyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(TFA) - Lysine(Trifluoroacetyl) | [K(TFA)] | Internal | ■■■■...■■■■ |
Misc | Maleimide - via SMCC | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | Methanethiosulfonate - MTS | MTS-NH | N-term | ■□□□...□□□□ |
Misc | Methoxysuccinyl - | H3C-O-CO-CH2-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Methylamide - N-Methyl | CO-NH(CH3) | C-term | □□□□...□□□■ |
Misc | Methylester - O-Methyl | COOMe | C-term | □□□□...□□□■ |
Misc | Myristoyl - | Myristoyl-NH | N-term | ■□□□...□□□□ |
Misc | N-Hydroxysuccinimid ester - NHS | CO-NHS | C-term | □□□□...□□□■ |
Misc | Orn(Biotin) - | [Orn(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Palmitoyl - | Palmitoyl-NH | N-term | ■□□□...□□□□ |
Misc | para-Bromo-D-Phenylalanine - | [D-F(4-Br)] | Internal | ■■■■...■■■■ |
Misc | para-Chloro-Phenylalanine - | [F(4-Cl)] | Internal | ■■■■...■■■■ |
Misc | para-Iodo-Phenylalanine - | [F(I)] | Internal | ■■■■...■■■■ |
Misc | para-Nitro-Phenylalanine - | [F(NO2)] | Internal | ■■■■...■■■■ |
Misc | Phenylacetic acid - Paa | Paa-NH | N-term | ■□□□...□□□□ |
Misc | Photo-Leucine - | [Photo-L] | Internal | ■■■■...■■■■ |
Misc | Photo-Methionine - | [Photo-M] | Internal | ■■■■...■■■■ |
Misc | p-tosyl-Gly - | [p-tosyl-G] | N-term | ■□□□...□□□□ |
Misc | S-Acetylmercaptosuccinyl - SAMSA | SAMSA-NH | N-term | ■□□□...□□□□ |
Misc | Ser(Acetyl) - | [S(Ac)] | Internal | ■■■■...■■■■ |
Misc | Ser(Octanoyl) - | [S(Oct)] | Internal | ■■■■...■■■■ |
Misc | Stearyl - | Stearyl-NH | N-term | ■□□□...□□□□ |
Misc | Succinyl - | Succinyl-NH | N-term | ■□□□...□□□□ |
Misc | t-Butylester - | COOtBu | C-term | □□□□...□□□■ |
Misc | tert. Butoxycarbonyl - Boc | Boc-NH | N-term | ■□□□...□□□□ |
Misc | Thiobenzyl - | CO-S(Bzl) | C-term | □□□□...□□□■ |
Misc | Thiopropionyl - Tpa | Tpa-NH | N-term | ■□□□...□□□□ |
Misc | trans-Cinnamoyl - | Cinnamoyl-NH | N-term | ■□□□...□□□□ |
Misc | Tyr(Palmitoyl) - | [Y(Palm)] | Internal | ■■■■...■■■■ |
Dye | (2,4-Dinitrophenyl)-Glycine - (2,4-DNP)-Gly | DNP-NH-G | N-term | ■□□□...□□□□ |
Dye | 2,4-Dinitrophenyl - DNP | CO-DNP | C-term | □□□□...□□□■ |
Dye | 2,4-Dinitrophenyl - DNP | DNP-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Carboxyfluorescein - 5/6-FLUO | 5/6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Tetramethylrhodamine - 5/6-TAMRA | 5/6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 5-Carboxyfluorescein - 5-FLUO | 5-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5-Tetramethylrhodamine - 5-TAMRA | 5-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 6-Carboxyfluorescein - 6-FLUO | 6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 6-Tetramethylrhodamine - 6-TAMRA | 6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 7-Amino-4-methylcoumarin - AMC | CO-AMC | C-term | □□□□...□□□■ |
Dye | Ala(2-Bacd) - benzo[b]acridin-12(5H)-on-2-yl | [Ala(2-Bacd)] | Internal | ■■■■...■■■■ |
Dye | Amino-trifluoromethyl-coumarin - AFC | CO-AFC | C-term | □□□□...□□□■ |
Dye | ATTO 647N - | ATTO 647N-NH | N-term | ■□□□...□□□□ |
Dye | Cy7 - | Cy7-NH | N-term | ■□□□...□□□□ |
Dye | Cys(5/6-TAMRA) - | [C(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(5-TAMRA) - | [C(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(6-TAMRA) - | [C(6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(acetamido-fluorescein) - | [C(acetamido-fluorescein)] | Internal | ■■■■...■■■■ |
Dye | Cys(AEDANS) - | [C(AEDANS)] | Internal | ■■■■...■■■■ |
Dye | Cys(Allophycocyanine) - | [C(Allophycocyanine)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 555) - | [C(DY 555)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 647P1) - | [C(DY 647P1)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 649) - | [C(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Cys(DYQ660) - | [Cys(DYQ660)] | Internal | ■■■■...■■■■ |
Dye | Dabcyl - | Dabcyl-NH | N-term | ■□□□...□□□□ |
Dye | Dabsyl - | Dabsyl-NH | N-term | ■□□□...□□□□ |
Dye | Dansyl - | CO-Dansyl | C-term | □□□□...□□□■ |
Dye | Dansyl - | Dansyl-NH | N-term | ■□□□...□□□□ |
Dye | DAP(DNP) - DNP at Diaminopropionic acid | [DAP(DNP)] | Internal | ■■■■...■■■■ |
Dye | DAP(DY 647) - DY 647 at Diaminopropionic ac. | [DPA(DY-647)] | Internal | ■■■■...■■■■ |
Dye | DAP(DyLight 488) - DyLight 488 at Diaminoprop. ac | [DPA(DY 488)] | Internal | ■■■■...■■■■ |
Dye | DAP(NMA) - NMA at Diaminopropionic acid | [DAP(NMA)] | Internal | ■■■■...■■■■ |
Dye | Dinitrophenyl-Ethylenediamine - EDDnp | CO-EDDnp | C-term | □□□□...□□□■ |
Dye | D-Lys(5/6-Fluorescein) - | [D-K(5/6-Fluorescein)] | Internal | ■■■■...■■■■ |
Dye | D-Lys(5/6-TAMRA) - | [D-Lys(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | DY 495 - | DY 495-NH | N-term | ■□□□...□□□□ |
Dye | DY 505 - | DY 505-NH | N-term | ■□□□...□□□□ |
Dye | DY 530 - | DY 530-NH | N-term | ■□□□...□□□□ |
Dye | DY 547 - | DY 547-NH | N-term | ■□□□...□□□□ |
Dye | DY 549 - | DY 549-NH | N-term | ■□□□...□□□□ |
Dye | DY 630 - | DY630-NH | N-term | ■□□□...□□□□ |
Dye | DY 633 - | DY 633-NH | N-term | ■□□□...□□□□ |
Dye | DY 635 - | DY 635-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - | DY 647-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - aminomodified | CO-DY 647 | C-term | □□□□...□□□■ |
Dye | DY 649 - | DY 649-NH | N-term | ■□□□...□□□□ |
Dye | DY 649P1 - aminomodified | CO-DY 649P1 | N-term | ■□□□...□□□□ |
Dye | DY 650 - | DY650-NH | Internal | ■■■■...■■■■ |
Dye | DY 680 - | DY 680-NH | N-term | ■□□□...□□□□ |
Dye | DY 682 - | DY 682-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | DY 750-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | CO-DY 750 | C-term | □□□□...□□□■ |
Dye | DY 751 - | DY 751-NH | N-term | ■□□□...□□□□ |
Dye | DY 800 - | DY 800-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 488 - | DY 488-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 504Q - | DyLight 504Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 543Q - | DyLight 543Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 554-R1 - | DyLight 554-R1-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 683Q - | DyLight 683Q-NH | N-term | ■□□□...□□□□ |
Dye | DYQ 660 - | DYQ 660-NH | N-term | ■□□□...□□□□ |
Dye | EDANS - | CO-EDANS | C-term | □□□□...□□□■ |
Dye | Glu(EDANS) - | [E(EDANS)] | Internal | ■■■■...■■■■ |
Dye | His(DNP) - | [H(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(2-(N-methylamino)benzoyl) - Lys(NMA) | [Lys(NMA)] | Internal | ■■■■...■■■■ |
Dye | Lys(2,4-DNP) - | [K(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-Fluorescein) - | [K(5/6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-TAMRA) - | [K(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-Fluorescein) - | [K(5-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-TAMRA) - | [K(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(6-Fluorescein) - | [K(6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(Ahx-(5-Fluorescein)) - | [K(Ahx-(5-Fluo))] | Internal | ■■■■...■■■■ |
Dye | Lys(ATTO 655) - | [K(ATTO 655)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dabcyl) - | [K(Dabcyl)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dde) - | [Lys(Dde)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 405) - | [K(DY 405)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 495) - | [K(DY 495)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 530) - | [K(DY 530)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 547) - | [K(DY 547)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 590) - | [K(DY 590)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 630) - | [K(DY 630)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 635) - | [K(DY 635)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 647) - | [K(DY 647)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 650) - | [Lys(DY 650)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 800) - | [K(DY 800)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 488) - | [K(DY 488)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 633) - Lys(DY633) | [K(DyLight 633)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 649) - | [K(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Lys(DYQ 3) - | [K(DYQ 3)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOC) - (7-hydroxycoumarin-3-yl)formyl | [Lys(HOC)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOMCA) - | [Lys(HOMCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MCA) - | [K(MCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MOC) - (7-methoxycoumarin-3-yl)formyl | Lys(MOC) | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine B) - | [K(Rhodamine B)] | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine Green) - | [K(Rhodamine Green)] | Internal | ■■■■...■■■■ |
Dye | Methoxycoumarin acetic acid - MCA | CO-MCA | C-term | □□□□...□□□■ |
Dye | Methoxycoumarin acetic acid - MCA | MCA-NH | N-term | ■□□□...□□□□ |
Dye | Ornithine(Coumarin343) - | [Orn(C343)] | Internal | ■■■■...■■■■ |
Dye | para-Nitroanilide - pNA | CO-pNA | C-term | □□□□...□□□■ |
Dye | para-Nitrophenylester - | COONp | C-term | □□□□...□□□■ |
Dye | QSY21 - | QSY21-NH | N-term | ■□□□...□□□□ |
Dye | Rhodamine 110 - | CO-Rhod 110 | C-term | □□□□...□□□■ |
Dye | Rhodamine B - | Rhodamin B-NH | N-term | ■□□□...□□□□ |
Dye | R-Phycoerythrin - | CO- R-Phycoerythrin | C-term | □□□□...□□□■ |
Dye | Tide Quencher 2 - TQ2 | TQ2-NH | N-term | ■□□□...□□□□ |
Linkers | 12-Aminododecanoic acid - Ado | [Ado] | Internal | ■■■■...■■■■ |
Linkers | 5-Aminopentanoic acid - Aminovaleric acid (Ava) | [Ava] | Internal | ■■■■...■■■■ |
Linkers | 6-Aminohexanoic acid (Ahx) - Aminocaproic acid (Aca) | [Ahx] | Internal | ■■■■...■■■■ |
Linkers | 8-Aminooctanoic acid - Aoc | [Aoc] | Internal | ■■■■...■■■■ |
Linkers | Cys(PEG 40000) - | [C(PEG 40000)] | Internal | ■■■■...■■■■ |
Linkers | Methyl-PEG12 - MS(PEG)12 | MS(PEG)12-NH | N-term | ■□□□...□□□□ |
Linkers | Methyl-PEG24 - MS(PEG)24 | MS(PEG)24-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 12 (40 atoms) - Amino-PEG-acid | [PEG 12] | Internal | ■■■■...■■■■ |
Linkers | PEG 2 (9 atoms) - 8-Amino-3,6-dioxaoctanoic acid | [PEG 2] | Internal | ■■■■...■■■■ |
Linkers | PEG 20,000 - | PEG 20,000-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 2000 - Amino-PEG-acid | [PEG 2000] | Internal | ■■■■...■■■■ |
Linkers | PEG 3 (13 atoms) - Amino-trioxadodecanoic acid | [PEG 3] | Internal | ■■■■...■■■■ |
Linkers | PEG 4 (16 atoms) - Amino-PEG-acid | [PEG 4] | Internal | ■■■■...■■■■ |
Linkers | PEG 5 (19 atoms) - Amino-PEG-acid | [PEG 5] | Internal | ■■■■...■■■■ |
Linkers | PEG 5000 - | CO-PEG 5000 | C-term | □□□□...□□□■ |
Linkers | PEG 5000 - Amino-PEG-acid | [PEG 5000] | Internal | ■■■■...■■■■ |
Linkers | PEG 6 (22 atoms) - Amino-PEG-acid | [PEG 6] | Internal | ■■■■...■■■■ |
Linkers | PEG 8 (28 atoms) - Amino-PEG-acid | [PEG 8] | Internal | ■■■■...■■■■ |
Linkers | PEG-4 Azide - | N3-PEG4-NH | N-term | ■□□□...□□□□ |
Special AAs | 2-(4'-Pentenyl)alanine - | [A(pentenyl)] | Internal | ■■■■...■■■■ |
Special AAs | 2,3-Diaminopropionic acid - DAP | [DAP] | Internal | ■■■■...■■■■ |
Special AAs | 2-Amino-4-thiazolinic acid - | [2-Amino-4-thiazolinyl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Indanyl-Glycine - Igl | [Igl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Methyl-D-Tryptophan - | [D-W(2-Me)] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-Alanine - 2-Nal | [2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-D-Alanine - D-2-Nal | [D-2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 3,3-Diphenyl-Alanine - | [A(F2)] | Internal | ■■■■...■■■■ |
Special AAs | 3,4-Dihydroxy-Phenylalanine - | [F(OH)2] | Internal | ■■■■...■■■■ |
Special AAs | 5,5-Dimethyl-Proline - | [P(Me2)] | Internal | ■■■■...■■■■ |
Special AAs | 5-Hydroxy-Tryptophan - | [W(OH)] | Internal | ■■■■...■■■■ |
Special AAs | Allyl-Glycine - 2-Aminopent-4-enoic acid | [G(allyl)] | Internal | ■■■■...■■■■ |
Special AAs | alpha-Allylalanine - | [Allyl-A] | Internal | ■■■■...■■■■ |
Special AAs | alpha-aminoadipic acid - Aad | [Aad] | Internal | ■■■■...■■■■ |
Special AAs | alpha-t-butylglycine - | [G(tBu)] | Internal | ■■■■...■■■■ |
Special AAs | Aminobenzoic acid - Abz | [Abz] | Internal | ■■■■...■■■■ |
Special AAs | Aminobutyric acid - Abu | [Abu] | Internal | ■■■■...■■■■ |
Special AAs | Aminocyclohexyl carbox. acid - Ac6c | [Ac6c] | Internal | ■■■■...■■■■ |
Special AAs | Aminoisobutyric acid - Aib | 0 | Internal | ■■■■...■■■■ |
Special AAs | Asp beta-methyl ester - Asp(OMe) | [D(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Asp(Isopropylamide) - | [D(Isopropylamide)] | Internal | ■■■■...■■■■ |
Special AAs | Azetidine-2-carboxylic acid - Aze | [Aze] | Internal | ■■■■...■■■■ |
Special AAs | Canavanine - Cav | [Cav] | Internal | ■■■■...■■■■ |
Special AAs | Citrulline - | [Cit] | Internal | ■■■■...■■■■ |
Special AAs | Cyclopentyl-Glycine - | [G(cyclopentyl)] | Internal | ■■■■...■■■■ |
Special AAs | Cysteic acid - Cya | [Cya] | Internal | ■■■■...■■■■ |
Special AAs | Cysteine sulfonate - | [C(SO3H)] | Internal | ■■■■...■■■■ |
Special AAs | D-2-Indanyl-Glycine - D-Igl | [D-Igl] | Internal | ■■■■...■■■■ |
Special AAs | D-4-Hydroxyphenly-Glycine - D-Hpg | [D-Hpg] | Internal | ■■■■...■■■■ |
Special AAs | D-Alanine - | *A | Internal | ■■■■...■■■■ |
Special AAs | D-Amino Acid - | [D-aa] | Internal | ■■■■...■■■■ |
Special AAs | D-Arginine - | *R | Internal | ■■■■...■■■■ |
Special AAs | D-Asparagine - | *N | Internal | ■■■■...■■■■ |
Special AAs | D-Aspartic acid - | *D | Internal | ■■■■...■■■■ |
Special AAs | D-Cysteine - | *C | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamic acid - | *E | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamine - | *Q | Internal | ■■■■...■■■■ |
Special AAs | D-Histidine - | *H | Internal | ■■■■...■■■■ |
Special AAs | D-Homocysteine - | [D-Homocys] | Internal | ■■■■...■■■■ |
Special AAs | D-Homoserine - | [D-Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Diaminotrioxatridecan suc. ac. - TTDS | [TTDS] | Internal | ■■■■...■■■■ |
Special AAs | D-Isoleucine - | *I | Internal | ■■■■...■■■■ |
Special AAs | D-Leucine - | *L | Internal | ■■■■...■■■■ |
Special AAs | D-Lysine - | *K | Internal | ■■■■...■■■■ |
Special AAs | D-Methionine - | *M | Internal | ■■■■...■■■■ |
Special AAs | D-Ornithine - | [D-Orn] | Internal | ■■■■...■■■■ |
Special AAs | D-Phenylalanine - | *F | Internal | ■■■■...■■■■ |
Special AAs | D-Proline - | *P | Internal | ■■■■...■■■■ |
Special AAs | D-Serine - | *S | Internal | ■■■■...■■■■ |
Special AAs | D-Threonine - | *T | Internal | ■■■■...■■■■ |
Special AAs | D-Tryptophan - | *W | Internal | ■■■■...■■■■ |
Special AAs | D-Tyrosine - | *Y | Internal | ■■■■...■■■■ |
Special AAs | D-Valine - | *V | Internal | ■■■■...■■■■ |
Special AAs | Formylglycine - | [G(CHO)] | Internal | ■■■■...■■■■ |
Special AAs | gamma-Carboxyglutamic acid - Gla | [Gla] | Internal | ■■■■...■■■■ |
Special AAs | Gamma-Glu, amidated - Gla, amidation at side chain | [Gla amide] | Internal | ■■■■...■■■■ |
Special AAs | Glu gamma-methyl ester - Glu(OMe) | [E(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Homo-Citrulline - | [Homocit] | Internal | ■■■■...■■■■ |
Special AAs | Homophenylalanine - | [HomoF] | Internal | ■■■■...■■■■ |
Special AAs | Homoserine - | [Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Hydroxy-Proline - Hyp | [Hyp] | Internal | ■■■■...■■■■ |
Special AAs | Hyposine - Hpu | [Hpu] | Internal | ■■■■...■■■■ |
Special AAs | Isoaspartic acid - | 0 | Internal | ■■■■...■■■■ |
Special AAs | Kynurenine - Kyn | [Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptoacetic acid - Thioglycolic acid | [Mercaptoacetyl] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptopropionic acid - Mpa | [Mpa] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfone - | [M(O2)] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfoxide - | [M(O)] | Internal | ■■■■...■■■■ |
Special AAs | N-Formylkynurenine - | [Formyl-Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Norleucine - | [Nle] | Internal | ■■■■...■■■■ |
Special AAs | Norvaline - 2-Aminopentanoic acid, Nva | [Nva] | Internal | ■■■■...■■■■ |
Special AAs | Octahydroindole carbox. acid - | [Oic] | Internal | ■■■■...■■■■ |
Special AAs | Ornitine - | [Orn] | Internal | ■■■■...■■■■ |
Special AAs | para-Acetylphenylalanine - pAF | [para-Ac-Phe] | Internal | ■■■■...■■■■ |
Special AAs | Phenylglycine - Phg | [Phg] | Internal | ■■■■...■■■■ |
Special AAs | Pyroglutamic acid - Pyr | [Pyr] | Internal | ■■■■...■■■■ |
Special AAs | Selenocysteine - Sec | [Sec] | Internal | ■■■■...■■■■ |
Special AAs | β-Alanine - | [β-A] | Internal | ■■■■...■■■■ |
Special AAs | β-Cyclohexyl-Alanine - Cha | [Cha] | Internal | ■■■■...■■■■ |
Special AAs | β-cyclohexyl-D-Alanine - D-Cha | [D-Cha] | Internal | ■■■■...■■■■ |
Special AAs | Tetrahydroisquinol.carb.acid - Tic | [Tic] | Internal | ■■■■...■■■■ |
Special AAs | Thiazolidine-4-carboxylic acid - Thz | [Thz] | Internal | ■■■■...■■■■ |
Special AAs | γ-Glutamic acid - | [γ-E] | Internal | ■■■■...■■■■ |
PTM | 1-Methyl-Tryptophan - | [W(1-Me)] | Internal | ■■■■...■■■■ |
PTM | 2,6-Dimethyl-Tyrosine - | [Y(Me2)] | Internal | ■■■■...■■■■ |
PTM | 2-Methylproline - | [P(2-Me)] | Internal | ■■■■...■■■■ |
PTM | 2-Methyl-Tryptophan - | [2-Me-Trp] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (asym.) - | [R(Me)2-asym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (sym.) - | [R(Me)2-sym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Cysteine - Penicillamine | [Pen] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Lysine - | [K(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Phenylalanine - | [F(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Valine - | [V(Me)2] | Internal | ■■■■...■■■■ |
PTM | Methyl-Arginine - | [R(Me)] | Internal | ■■■■...■■■■ |
PTM | Methyl-Lysine - | [K(Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Alanine - | [Ala(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Glycine - | [G(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Leucine - | [L(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Phenylalanine - | [F(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Tyrosine - | [Y(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Valine - | [V(N-Me)] | Internal | ■■■■...■■■■ |
PTM | Phosphono-Phenylalanine - | [F(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Serine - | [S(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Threonin - | [T(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Tyrosin - | [Y(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | β-GalNAc-Threonine - | [T(β-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Asparagine - | [N(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Serine - | [S(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Threonine - | [T(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Serine - | [S(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Threonine - | [T(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Tyrosine - | [Y(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Arginine - | [R(Me3)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Lysine - | [K(Me)3] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Serine - | [S(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Threonine - | [T(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Serine - | [S(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Threonine - | [T(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Aspartic acid - | [D(α-Me)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Glutamic acid - | [E(α-Me)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (13C3;15N) - | [A(13C3; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (D3) - | [A(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6) - | [R(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6;15N4) - | [R(13C6; 15N4)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asn (13C4;15N2) - | [N(13C4; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asp (13C4;15N - | [D(13C4; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gln (13C5;15N2) - | [Q(13C5;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Glu (13C5;15N) - | [E(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gly (13C2;15N) - | [G(13C2; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy His (15N3) - | [His(15N3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ile (13C6;15N) - | [I(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C1) - | [L(13C1)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6) - | [L(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6;15N) - | [L(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D10) - | [L(D10)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D3) - | [L(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6) - | [K(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6;15N2) - | [K(13C6; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Phe (13C9;15N) - | [F(13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Pro (13C5;15N) - | [P(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (13C3;15N) - | [S (13C3;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (D3) - | [S(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Thr(13C4;15N) - | [T(13C4;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Trp (13C11;15N2) - | [W (13C11;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Tyr (13C9;15N) - | [Y (13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5) - | [V(13C5)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5;15N) - | [V(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | L-tyrosine-phenyl-d4 - deuterated Tyr | [Tyr(D4)] | Internal | ■■■■...■■■■ |
Isotopic | TMT6-126 - | TMT6-126-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-127 - | TMT6-127-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-128 - | TMT6-128-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-129 - | TMT6-129-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-130 - | TMT6-130-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-131 - | TMT6-131-NH | N-term | ■□□□...□□□□ |
Immunization | Blue Carrier Protein - | CO-NH -Blue Carrier Protein | C-term | □□□□...□□□■ |
Immunization | BSA - | CO-BSA | C-term | □□□□...□□□■ |
Immunization | BSA via Glutaraldehyde - | BSA-NH | N-term | ■□□□...□□□□ |
Immunization | Cys(BSA) - | [C(BSA)] | Internal | ■■■■...■■■■ |
Immunization | Cys(KLH) - | [C(KLH)] | Internal | ■■■■...■■■■ |
Immunization | Cys(OVA) - | [C(OVA)] | Internal | ■■■■...■■■■ |
Immunization | KLH - | CO-KLH | C-term | □□□□...□□□■ |
Immunization | KLH via Glutaraldehyde - | KLH-NH | N-term | ■□□□...□□□□ |
Immunization | Lys(BSA) - | [K(BSA] | Internal | ■■■■...■■■■ |
Immunization | Lys(KLH) - | [K(KLH] | Internal | ■■■■...■■■■ |
Immunization | MAP 16 - Crude 8-20 aa, at C-term. only | MAP 16 | C-term | □□□□...□□□■ |
Immunization | MAP 4 - Crude 8-20 aa, at C-term. only | MAP 4 | C-term | □□□□...□□□■ |
Immunization | MAP 8 - Crude 8-20 aa, at C-term. only | MAP 8 | C-term | □□□□...□□□■ |
Immunization | OVA - | CO-OVA | C-term | □□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | 3-Azido-Alanine - Aza | [Aza] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | 5-Azidopentanoyl - 5-Azidovalerianoyl | 5-Azidopentanoyl-NH | N-term | ■□□□...□□□□ |
Cyclization/Dimerization/Special Synthesis | 6-Azido-Norleucine - Nle(N3) | [Nle(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azidohomoalanine - Aha/ Dab(N3) | [Aha] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azido-Lysine - | [K(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (C-C), disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (D-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (E-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (K-C-terminus) - side chain of K to C-terminus | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (NH-CO), homodetic - | [ ] | N-term | ■□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | Double disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | D-Propargylglycine - D-Pra | [D-Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Hetero-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Homo-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Mixed amino acids - | [X] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | para-Azido-Phenylalanine - | [F(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylalanine - alpha-Me-L-Ala(Pentynyl)-OH | [Propargylala] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylglycine - Pra | [Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Asymmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Symmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Site directed disulfide bridge - Cyclisation (C-C) | [Cys cyclo(S-S)] | Internal | ■■■■...■■■■ |
A. Our standard peptides are delivered lyophilized with trifluoroacetate (TFA) as a counterion and packaged in glass vials, stored under argon to protect from oxygen. For additional formats, such as delivery in low-binding plastics or 2-D barcoded plates, or in-solution with different solvents, please inquire.
A. The detailed spectra are included on the Certificate of Analysis for your records. Additional quality controls such as amino acid analysis (AAA) and further nonstandard analysis are available on request.
A. We recommend preparing single-use aliquots and storing the products immediately at –20°C, upon receipt. In this manner, the lyophilized peptides can be stable for several years. Solubilized peptides should be used immediately because they are unstable (the lower the concentration, the more unstable the peptides are). Any remaining peptides in solution should be re-lyophilized for longer storage.
A. Always wear gloves when working with peptides to avoid contamination (e.g., enzymatic, bacterial, etc.). As many peptides are light-sensitive, they should also be protected from direct light. Avoid repeated freeze-thaw cycles.
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design
A. All the information required to get an accurate quote is included in the Quote Request form.
A. Peptides are complex biomolecules, and each peptide has unique chemical and physical properties because of the unique amino acid composition. Although some peptides are easy to dissolve in aqueous solutions, peptides are often insoluble, especially when they contain long stretches of hydrophobic amino acids.
Because of the unique solubility of each peptide, we recommend first testing the solubilization of each peptide with a small amount of product.
Peptide length | 2-110* amino acids, L- or D-isoforms |
Manufacturing scale | 1 mg–1 kg+ ** |
Purity options | From crude to >98% pure |
Quality control | MS for crude peptides, MS and analytical HPLC for all other grades of purity |
Production time | 2–3 weeks |
Modifications | Extensive list of modifications and labels, including heavy peptides (isotope-labeled) |
Conjugation | Custom conjugation available (e.g., carrier protein-peptide conjugates and multiple antigen peptides, MAPs) |
* Not all peptides will be available in lengths of 110 due to sequence composition.
** Greater amounts available upon request.
Thermo Scientific Custom Peptide Synthesis Service offers numerous options for purity levels, modifications, and formats for your synthetic peptide order, giving you the flexibility to meet your research needs. Peptide synthesis is performed using the latest Fmoc solid-phase technology and peptides are purified by reversed-phase HPLC. Peptide sequence is confirmed by MS. Peptides with guaranteed purity are additionally characterized using analytical HPLC to determine the final purity.
We offer a wide range of N-terminal, C-terminal, and other modifications with our standard peptide service. Custom conjugation services (6−25 amino acids) are available, including conjugation to carrier proteins (KLH, BSA, OVA, or Thermo Scientific Imject Blue Carrier Protein) and MAPs.
Type | Name | Abbreviation or alternative name | Position within sequence | Graphical representation of available positions |
---|---|---|---|---|
Misc | 1-Adamantanecarbonyl - | 1-Adamantanecarbonyl-NH | N-term | ■□□□...□□□□ |
Misc | 2,4,6-Trinitrophenyl - TNP | TNP-NH | N-term | ■□□□...□□□□ |
Misc | 2-Furoyl - | 2-Furoyl-NH | N-term | ■□□□...□□□□ |
Misc | 2-Naphtoic acid - | Naphtyl-NH | N-term | ■□□□...□□□□ |
Misc | 3-(Maleimido)propionyl - | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | 3,5-Diiodo-Tyrosine - | [Y(I)2] | Internal | ■■■■...■■■■ |
Misc | 3-Chloro-Tyrosine - | [Y(Cl)] | Internal | ■■■■...■■■■ |
Misc | 3-nitro-2-pyridylthio-L-Cys - | [C(Npys)] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Indole - | [3-Nitro-Indole] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Pyrrol - | [3-Nitro-Pyrrol] | Internal | ■■■■...■■■■ |
Misc | 3-Nitro-Tyrosine - | [Y(NO2)] | Internal | ■■■■...■■■■ |
Misc | 4-Methylthio-2-oxobutanoic ac. - | [H3C-S-CH2-CH2-CO-CO-NH] | N-term | ■□□□...□□□□ |
Misc | 4-nitrobenzo-2-oxa-1,3-diazole - NBD | NBD-NH | N-term | ■□□□...□□□□ |
Misc | 4-Pentynoic acid - | 4-Pentynoic acid-NH | N-term | ■□□□...□□□□ |
Misc | 4-Phenylazophenol - O-PAP | CO-PAP | C-term | □□□□...□□□■ |
Misc | 6-Bromo-D/L-Tryptophan - | [D/L-W(Br)] | Internal | ■■■■...■■■■ |
Misc | 6-Cloro-Tryptophan - | [W(Cl)] | Internal | ■■■■...■■■■ |
Misc | Acetylation - | Acetyl-NH | N-term | ■□□□...□□□□ |
Misc | Aldehyde - | CHO | C-term | □□□□...□□□■ |
Misc | Amidation - | CONH2 | C-term | □□□□...□□□■ |
Misc | Amino nitrophenyl prop. acid - | [Amino nitrophenyl prop. acid] | Internal | ■■■■...■■■■ |
Misc | Amino-nitrophenyl propion. ac. - ANP-linker | [ANP] | Internal | ■■■■...■■■■ |
Misc | Aminooxyacetyl - | H2N-O-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Benzoyl - Bz | Benzoyl-NH | N-term | ■□□□...□□□□ |
Misc | Benzyloxycarbonyl - Cbz, Z | Z-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - | Biotin-NH | N-term | ■□□□...□□□□ |
Misc | Biotin - Ethylenediamin-Biotin | CO-NH-CH2-CH2-NH-Biotin | C-term | □□□□...□□□■ |
Misc | Biotin-SS-linker - | Biotin-SS-NH | N-term | ■□□□...□□□□ |
Misc | Bis(aminoethyl)ethylene glycol - | CO-NH-(CH2-CH2-O)2-CH2-CH2-NH2 | C-term | □□□□...□□□■ |
Misc | Brom acetic acid - | BrCH2CO-NH | N-term | ■□□□...□□□□ |
Misc | Chlormethylketon - CMK | CMK | C-term | □□□□...□□□■ |
Misc | Cys(4-Methoxytrityl) - Cys(MMT) | [C(Mmt)] | Internal | ■■■■...■■■■ |
Misc | Cys(Acetamidomethyl) - Cys(Acm) | [C(Acm)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carbamidomethyl) - Cys(CAM) / Cys(CH2CONH2) | [C(Cam)] | Internal | ■■■■...■■■■ |
Misc | Cys(Carboxymethyl) - Cys(CMC) / Cys(CH2COOH) | [C(Cmc)] | Internal | ■■■■...■■■■ |
Misc | Cys(di(palmitoyloxy)-propyl) - Cys(Pam2) | [C(Pam2)] | Internal | ■■■■...■■■■ |
Misc | Cys(Farnesyl) - | [C(Farnesyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(HRP) - Cys(horseradish peroxidase) | [C(HRP)] | Internal | ■■■■...■■■■ |
Misc | Cys(MTSL) - | [C(MTSL)] | Internal | ■■■■...■■■■ |
Misc | Cys(Myristyl) - Cys(CH2(CH2)12CH3) | [C(Myristyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(Palmitoyl) - | [C(Palm)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Butyl) - | [C(S-But)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Ethyl) - | [C(S-Et)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isobutyl) - | [C(S-Isobut)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Isopropyl) - | [C(S-Isoprop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Methyl) - | [C(S-Me)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Pentyl) - | [C(S-Pentyl)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-Propyl) - | [C(S-Prop)] | Internal | ■■■■...■■■■ |
Misc | Cys(S-tert-Butyl) - | [C(S-tert-But] | Internal | ■■■■...■■■■ |
Misc | Cysteamide - | Cysteamide | C-term | □□□□...□□□■ |
Misc | Decanoyl - | Decanoyl-NH | N-term | ■□□□...□□□□ |
Misc | Desthiobiotin - | Desthiobiotin-NH | N-term | ■□□□...□□□□ |
Misc | Digoxigenin - | Digoxigenin-NH | N-term | ■□□□...□□□□ |
Misc | DOTA - | [DOTA] | N-term | ■□□□...□□□□ |
Misc | Ethylamide - N-Ethyl | CONH(C2H5) | C-term | □□□□...□□□■ |
Misc | Ethylester - O-Ethyl | COOEt | C-term | □□□□...□□□■ |
Misc | Fmoc - | Fmoc-NH | N-term | ■□□□...□□□□ |
Misc | Formyl - | Formyl-NH | N-term | ■□□□...□□□□ |
Misc | Glu(Arg) - Arg at side chain of Glu | [Glu(Arg)] | Internal | ■■■■...■■■■ |
Misc | Homoserine lactone - | [Homoser lactone] | C-term | □□□□...□□□■ |
Misc | HRP - | CO-NH -HRP | C-term | □□□□...□□□■ |
Misc | HRP - | HRP-NH | N-term | ■□□□...□□□□ |
Misc | Hydrazide - | CO-NH -NH2 | C-term | □□□□...□□□■ |
Misc | Hydrazine-succinyl - | H2N-NH-CO-CH2-CH2-CO-NH- | N-term | ■□□□...□□□□ |
Misc | Iodoacetyl - | IH2C-CO- | N-term | ■□□□...□□□□ |
Misc | Lys(2-Aminooxy acetic acid) - Lys(Aoa) | [K(Aoa)] | Internal | ■■■■...■■■■ |
Misc | Lys(Acetyl) - | [K(Ac)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-Biotin) - | [K(Ahx-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ahx-SS-Biotin) - | [K(Ahx-SS-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Biotin) - | [K(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Boc) - | [K(Boc)] | Internal | ■■■■...■■■■ |
Misc | Lys(Butyryl) - | [K(Butyryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Desthiobiotin) - | [K(Desthiobiotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Digoxigenin] - | [K(Digoxigenin)] | Internal | ■■■■...■■■■ |
Misc | Lys(DST-PEG 2-Biotin) - | [K(DST-PEG 2-Biotin)] | Internal | ■■■■...■■■■ |
Misc | Lys(Ferrocene) - | [K(Ferrocene)] | Internal | ■■■■...■■■■ |
Misc | Lys(Formyl) - | [K(Formyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(GG) - Ubiquitin | [K(GG)] | Internal | ■■■■...■■■■ |
Misc | Lys(GGRLR) - | [K(GGRLR)] | Internal | ■■■■...■■■■ |
Misc | Lys(Glutaryl) - | [K(Glutaryl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Lipoyl) - | [K(Lipoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(NIP) - | [K(NIP)] | Internal | ■■■■...■■■■ |
Misc | Lys(Palmitoyl) - | [K(Palmitoyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Propionyl) - | [K(Propionyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Pyrofolate) - | [K(Pyrofolate)] | Internal | ■■■■...■■■■ |
Misc | Lys(Stearyl) - | [K(Stearyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(Succinyl) - | [K(Succinyl)] | Internal | ■■■■...■■■■ |
Misc | Lys(TFA) - Lysine(Trifluoroacetyl) | [K(TFA)] | Internal | ■■■■...■■■■ |
Misc | Maleimide - via SMCC | Maleimide-NH | N-term | ■□□□...□□□□ |
Misc | Methanethiosulfonate - MTS | MTS-NH | N-term | ■□□□...□□□□ |
Misc | Methoxysuccinyl - | H3C-O-CO-CH2-CH2-CO-NH | N-term | ■□□□...□□□□ |
Misc | Methylamide - N-Methyl | CO-NH(CH3) | C-term | □□□□...□□□■ |
Misc | Methylester - O-Methyl | COOMe | C-term | □□□□...□□□■ |
Misc | Myristoyl - | Myristoyl-NH | N-term | ■□□□...□□□□ |
Misc | N-Hydroxysuccinimid ester - NHS | CO-NHS | C-term | □□□□...□□□■ |
Misc | Orn(Biotin) - | [Orn(Biotin)] | Internal | ■■■■...■■■■ |
Misc | Palmitoyl - | Palmitoyl-NH | N-term | ■□□□...□□□□ |
Misc | para-Bromo-D-Phenylalanine - | [D-F(4-Br)] | Internal | ■■■■...■■■■ |
Misc | para-Chloro-Phenylalanine - | [F(4-Cl)] | Internal | ■■■■...■■■■ |
Misc | para-Iodo-Phenylalanine - | [F(I)] | Internal | ■■■■...■■■■ |
Misc | para-Nitro-Phenylalanine - | [F(NO2)] | Internal | ■■■■...■■■■ |
Misc | Phenylacetic acid - Paa | Paa-NH | N-term | ■□□□...□□□□ |
Misc | Photo-Leucine - | [Photo-L] | Internal | ■■■■...■■■■ |
Misc | Photo-Methionine - | [Photo-M] | Internal | ■■■■...■■■■ |
Misc | p-tosyl-Gly - | [p-tosyl-G] | N-term | ■□□□...□□□□ |
Misc | S-Acetylmercaptosuccinyl - SAMSA | SAMSA-NH | N-term | ■□□□...□□□□ |
Misc | Ser(Acetyl) - | [S(Ac)] | Internal | ■■■■...■■■■ |
Misc | Ser(Octanoyl) - | [S(Oct)] | Internal | ■■■■...■■■■ |
Misc | Stearyl - | Stearyl-NH | N-term | ■□□□...□□□□ |
Misc | Succinyl - | Succinyl-NH | N-term | ■□□□...□□□□ |
Misc | t-Butylester - | COOtBu | C-term | □□□□...□□□■ |
Misc | tert. Butoxycarbonyl - Boc | Boc-NH | N-term | ■□□□...□□□□ |
Misc | Thiobenzyl - | CO-S(Bzl) | C-term | □□□□...□□□■ |
Misc | Thiopropionyl - Tpa | Tpa-NH | N-term | ■□□□...□□□□ |
Misc | trans-Cinnamoyl - | Cinnamoyl-NH | N-term | ■□□□...□□□□ |
Misc | Tyr(Palmitoyl) - | [Y(Palm)] | Internal | ■■■■...■■■■ |
Dye | (2,4-Dinitrophenyl)-Glycine - (2,4-DNP)-Gly | DNP-NH-G | N-term | ■□□□...□□□□ |
Dye | 2,4-Dinitrophenyl - DNP | CO-DNP | C-term | □□□□...□□□■ |
Dye | 2,4-Dinitrophenyl - DNP | DNP-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Carboxyfluorescein - 5/6-FLUO | 5/6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5/6-Tetramethylrhodamine - 5/6-TAMRA | 5/6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 5-Carboxyfluorescein - 5-FLUO | 5-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 5-Tetramethylrhodamine - 5-TAMRA | 5-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 6-Carboxyfluorescein - 6-FLUO | 6-Fluorescein-NH | N-term | ■□□□...□□□□ |
Dye | 6-Tetramethylrhodamine - 6-TAMRA | 6-TAMRA-NH | N-term | ■□□□...□□□□ |
Dye | 7-Amino-4-methylcoumarin - AMC | CO-AMC | C-term | □□□□...□□□■ |
Dye | Ala(2-Bacd) - benzo[b]acridin-12(5H)-on-2-yl | [Ala(2-Bacd)] | Internal | ■■■■...■■■■ |
Dye | Amino-trifluoromethyl-coumarin - AFC | CO-AFC | C-term | □□□□...□□□■ |
Dye | ATTO 647N - | ATTO 647N-NH | N-term | ■□□□...□□□□ |
Dye | Cy7 - | Cy7-NH | N-term | ■□□□...□□□□ |
Dye | Cys(5/6-TAMRA) - | [C(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(5-TAMRA) - | [C(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(6-TAMRA) - | [C(6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Cys(acetamido-fluorescein) - | [C(acetamido-fluorescein)] | Internal | ■■■■...■■■■ |
Dye | Cys(AEDANS) - | [C(AEDANS)] | Internal | ■■■■...■■■■ |
Dye | Cys(Allophycocyanine) - | [C(Allophycocyanine)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 555) - | [C(DY 555)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 647P1) - | [C(DY 647P1)] | Internal | ■■■■...■■■■ |
Dye | Cys(DY 649) - | [C(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Cys(DYQ660) - | [Cys(DYQ660)] | Internal | ■■■■...■■■■ |
Dye | Dabcyl - | Dabcyl-NH | N-term | ■□□□...□□□□ |
Dye | Dabsyl - | Dabsyl-NH | N-term | ■□□□...□□□□ |
Dye | Dansyl - | CO-Dansyl | C-term | □□□□...□□□■ |
Dye | Dansyl - | Dansyl-NH | N-term | ■□□□...□□□□ |
Dye | DAP(DNP) - DNP at Diaminopropionic acid | [DAP(DNP)] | Internal | ■■■■...■■■■ |
Dye | DAP(DY 647) - DY 647 at Diaminopropionic ac. | [DPA(DY-647)] | Internal | ■■■■...■■■■ |
Dye | DAP(DyLight 488) - DyLight 488 at Diaminoprop. ac | [DPA(DY 488)] | Internal | ■■■■...■■■■ |
Dye | DAP(NMA) - NMA at Diaminopropionic acid | [DAP(NMA)] | Internal | ■■■■...■■■■ |
Dye | Dinitrophenyl-Ethylenediamine - EDDnp | CO-EDDnp | C-term | □□□□...□□□■ |
Dye | D-Lys(5/6-Fluorescein) - | [D-K(5/6-Fluorescein)] | Internal | ■■■■...■■■■ |
Dye | D-Lys(5/6-TAMRA) - | [D-Lys(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | DY 495 - | DY 495-NH | N-term | ■□□□...□□□□ |
Dye | DY 505 - | DY 505-NH | N-term | ■□□□...□□□□ |
Dye | DY 530 - | DY 530-NH | N-term | ■□□□...□□□□ |
Dye | DY 547 - | DY 547-NH | N-term | ■□□□...□□□□ |
Dye | DY 549 - | DY 549-NH | N-term | ■□□□...□□□□ |
Dye | DY 630 - | DY630-NH | N-term | ■□□□...□□□□ |
Dye | DY 633 - | DY 633-NH | N-term | ■□□□...□□□□ |
Dye | DY 635 - | DY 635-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - | DY 647-NH | N-term | ■□□□...□□□□ |
Dye | DY 647 - aminomodified | CO-DY 647 | C-term | □□□□...□□□■ |
Dye | DY 649 - | DY 649-NH | N-term | ■□□□...□□□□ |
Dye | DY 649P1 - aminomodified | CO-DY 649P1 | N-term | ■□□□...□□□□ |
Dye | DY 650 - | DY650-NH | Internal | ■■■■...■■■■ |
Dye | DY 680 - | DY 680-NH | N-term | ■□□□...□□□□ |
Dye | DY 682 - | DY 682-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | DY 750-NH | N-term | ■□□□...□□□□ |
Dye | DY 750 - | CO-DY 750 | C-term | □□□□...□□□■ |
Dye | DY 751 - | DY 751-NH | N-term | ■□□□...□□□□ |
Dye | DY 800 - | DY 800-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 488 - | DY 488-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 504Q - | DyLight 504Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 543Q - | DyLight 543Q-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 554-R1 - | DyLight 554-R1-NH | N-term | ■□□□...□□□□ |
Dye | DyLight 683Q - | DyLight 683Q-NH | N-term | ■□□□...□□□□ |
Dye | DYQ 660 - | DYQ 660-NH | N-term | ■□□□...□□□□ |
Dye | EDANS - | CO-EDANS | C-term | □□□□...□□□■ |
Dye | Glu(EDANS) - | [E(EDANS)] | Internal | ■■■■...■■■■ |
Dye | His(DNP) - | [H(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(2-(N-methylamino)benzoyl) - Lys(NMA) | [Lys(NMA)] | Internal | ■■■■...■■■■ |
Dye | Lys(2,4-DNP) - | [K(DNP)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-Fluorescein) - | [K(5/6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5/6-TAMRA) - | [K(5/6-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-Fluorescein) - | [K(5-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(5-TAMRA) - | [K(5-TAMRA)] | Internal | ■■■■...■■■■ |
Dye | Lys(6-Fluorescein) - | [K(6-Fluo)] | Internal | ■■■■...■■■■ |
Dye | Lys(Ahx-(5-Fluorescein)) - | [K(Ahx-(5-Fluo))] | Internal | ■■■■...■■■■ |
Dye | Lys(ATTO 655) - | [K(ATTO 655)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dabcyl) - | [K(Dabcyl)] | Internal | ■■■■...■■■■ |
Dye | Lys(Dde) - | [Lys(Dde)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 405) - | [K(DY 405)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 495) - | [K(DY 495)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 530) - | [K(DY 530)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 547) - | [K(DY 547)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 590) - | [K(DY 590)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 630) - | [K(DY 630)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 635) - | [K(DY 635)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 647) - | [K(DY 647)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 650) - | [Lys(DY 650)] | Internal | ■■■■...■■■■ |
Dye | Lys(DY 800) - | [K(DY 800)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 488) - | [K(DY 488)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 633) - Lys(DY633) | [K(DyLight 633)] | Internal | ■■■■...■■■■ |
Dye | Lys(DyLight 649) - | [K(DY 649)] | Internal | ■■■■...■■■■ |
Dye | Lys(DYQ 3) - | [K(DYQ 3)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOC) - (7-hydroxycoumarin-3-yl)formyl | [Lys(HOC)] | Internal | ■■■■...■■■■ |
Dye | Lys(HOMCA) - | [Lys(HOMCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MCA) - | [K(MCA)] | Internal | ■■■■...■■■■ |
Dye | Lys(MOC) - (7-methoxycoumarin-3-yl)formyl | Lys(MOC) | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine B) - | [K(Rhodamine B)] | Internal | ■■■■...■■■■ |
Dye | Lys(Rhodamine Green) - | [K(Rhodamine Green)] | Internal | ■■■■...■■■■ |
Dye | Methoxycoumarin acetic acid - MCA | CO-MCA | C-term | □□□□...□□□■ |
Dye | Methoxycoumarin acetic acid - MCA | MCA-NH | N-term | ■□□□...□□□□ |
Dye | Ornithine(Coumarin343) - | [Orn(C343)] | Internal | ■■■■...■■■■ |
Dye | para-Nitroanilide - pNA | CO-pNA | C-term | □□□□...□□□■ |
Dye | para-Nitrophenylester - | COONp | C-term | □□□□...□□□■ |
Dye | QSY21 - | QSY21-NH | N-term | ■□□□...□□□□ |
Dye | Rhodamine 110 - | CO-Rhod 110 | C-term | □□□□...□□□■ |
Dye | Rhodamine B - | Rhodamin B-NH | N-term | ■□□□...□□□□ |
Dye | R-Phycoerythrin - | CO- R-Phycoerythrin | C-term | □□□□...□□□■ |
Dye | Tide Quencher 2 - TQ2 | TQ2-NH | N-term | ■□□□...□□□□ |
Linkers | 12-Aminododecanoic acid - Ado | [Ado] | Internal | ■■■■...■■■■ |
Linkers | 5-Aminopentanoic acid - Aminovaleric acid (Ava) | [Ava] | Internal | ■■■■...■■■■ |
Linkers | 6-Aminohexanoic acid (Ahx) - Aminocaproic acid (Aca) | [Ahx] | Internal | ■■■■...■■■■ |
Linkers | 8-Aminooctanoic acid - Aoc | [Aoc] | Internal | ■■■■...■■■■ |
Linkers | Cys(PEG 40000) - | [C(PEG 40000)] | Internal | ■■■■...■■■■ |
Linkers | Methyl-PEG12 - MS(PEG)12 | MS(PEG)12-NH | N-term | ■□□□...□□□□ |
Linkers | Methyl-PEG24 - MS(PEG)24 | MS(PEG)24-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 12 (40 atoms) - Amino-PEG-acid | [PEG 12] | Internal | ■■■■...■■■■ |
Linkers | PEG 2 (9 atoms) - 8-Amino-3,6-dioxaoctanoic acid | [PEG 2] | Internal | ■■■■...■■■■ |
Linkers | PEG 20,000 - | PEG 20,000-NH | N-term | ■□□□...□□□□ |
Linkers | PEG 2000 - Amino-PEG-acid | [PEG 2000] | Internal | ■■■■...■■■■ |
Linkers | PEG 3 (13 atoms) - Amino-trioxadodecanoic acid | [PEG 3] | Internal | ■■■■...■■■■ |
Linkers | PEG 4 (16 atoms) - Amino-PEG-acid | [PEG 4] | Internal | ■■■■...■■■■ |
Linkers | PEG 5 (19 atoms) - Amino-PEG-acid | [PEG 5] | Internal | ■■■■...■■■■ |
Linkers | PEG 5000 - | CO-PEG 5000 | C-term | □□□□...□□□■ |
Linkers | PEG 5000 - Amino-PEG-acid | [PEG 5000] | Internal | ■■■■...■■■■ |
Linkers | PEG 6 (22 atoms) - Amino-PEG-acid | [PEG 6] | Internal | ■■■■...■■■■ |
Linkers | PEG 8 (28 atoms) - Amino-PEG-acid | [PEG 8] | Internal | ■■■■...■■■■ |
Linkers | PEG-4 Azide - | N3-PEG4-NH | N-term | ■□□□...□□□□ |
Special AAs | 2-(4'-Pentenyl)alanine - | [A(pentenyl)] | Internal | ■■■■...■■■■ |
Special AAs | 2,3-Diaminopropionic acid - DAP | [DAP] | Internal | ■■■■...■■■■ |
Special AAs | 2-Amino-4-thiazolinic acid - | [2-Amino-4-thiazolinyl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Indanyl-Glycine - Igl | [Igl] | Internal | ■■■■...■■■■ |
Special AAs | 2-Methyl-D-Tryptophan - | [D-W(2-Me)] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-Alanine - 2-Nal | [2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 2-Naphthyl-D-Alanine - D-2-Nal | [D-2-Nal] | Internal | ■■■■...■■■■ |
Special AAs | 3,3-Diphenyl-Alanine - | [A(F2)] | Internal | ■■■■...■■■■ |
Special AAs | 3,4-Dihydroxy-Phenylalanine - | [F(OH)2] | Internal | ■■■■...■■■■ |
Special AAs | 5,5-Dimethyl-Proline - | [P(Me2)] | Internal | ■■■■...■■■■ |
Special AAs | 5-Hydroxy-Tryptophan - | [W(OH)] | Internal | ■■■■...■■■■ |
Special AAs | Allyl-Glycine - 2-Aminopent-4-enoic acid | [G(allyl)] | Internal | ■■■■...■■■■ |
Special AAs | alpha-Allylalanine - | [Allyl-A] | Internal | ■■■■...■■■■ |
Special AAs | alpha-aminoadipic acid - Aad | [Aad] | Internal | ■■■■...■■■■ |
Special AAs | alpha-t-butylglycine - | [G(tBu)] | Internal | ■■■■...■■■■ |
Special AAs | Aminobenzoic acid - Abz | [Abz] | Internal | ■■■■...■■■■ |
Special AAs | Aminobutyric acid - Abu | [Abu] | Internal | ■■■■...■■■■ |
Special AAs | Aminocyclohexyl carbox. acid - Ac6c | [Ac6c] | Internal | ■■■■...■■■■ |
Special AAs | Aminoisobutyric acid - Aib | 0 | Internal | ■■■■...■■■■ |
Special AAs | Asp beta-methyl ester - Asp(OMe) | [D(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Asp(Isopropylamide) - | [D(Isopropylamide)] | Internal | ■■■■...■■■■ |
Special AAs | Azetidine-2-carboxylic acid - Aze | [Aze] | Internal | ■■■■...■■■■ |
Special AAs | Canavanine - Cav | [Cav] | Internal | ■■■■...■■■■ |
Special AAs | Citrulline - | [Cit] | Internal | ■■■■...■■■■ |
Special AAs | Cyclopentyl-Glycine - | [G(cyclopentyl)] | Internal | ■■■■...■■■■ |
Special AAs | Cysteic acid - Cya | [Cya] | Internal | ■■■■...■■■■ |
Special AAs | Cysteine sulfonate - | [C(SO3H)] | Internal | ■■■■...■■■■ |
Special AAs | D-2-Indanyl-Glycine - D-Igl | [D-Igl] | Internal | ■■■■...■■■■ |
Special AAs | D-4-Hydroxyphenly-Glycine - D-Hpg | [D-Hpg] | Internal | ■■■■...■■■■ |
Special AAs | D-Alanine - | *A | Internal | ■■■■...■■■■ |
Special AAs | D-Amino Acid - | [D-aa] | Internal | ■■■■...■■■■ |
Special AAs | D-Arginine - | *R | Internal | ■■■■...■■■■ |
Special AAs | D-Asparagine - | *N | Internal | ■■■■...■■■■ |
Special AAs | D-Aspartic acid - | *D | Internal | ■■■■...■■■■ |
Special AAs | D-Cysteine - | *C | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamic acid - | *E | Internal | ■■■■...■■■■ |
Special AAs | D-Glutamine - | *Q | Internal | ■■■■...■■■■ |
Special AAs | D-Histidine - | *H | Internal | ■■■■...■■■■ |
Special AAs | D-Homocysteine - | [D-Homocys] | Internal | ■■■■...■■■■ |
Special AAs | D-Homoserine - | [D-Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Diaminotrioxatridecan suc. ac. - TTDS | [TTDS] | Internal | ■■■■...■■■■ |
Special AAs | D-Isoleucine - | *I | Internal | ■■■■...■■■■ |
Special AAs | D-Leucine - | *L | Internal | ■■■■...■■■■ |
Special AAs | D-Lysine - | *K | Internal | ■■■■...■■■■ |
Special AAs | D-Methionine - | *M | Internal | ■■■■...■■■■ |
Special AAs | D-Ornithine - | [D-Orn] | Internal | ■■■■...■■■■ |
Special AAs | D-Phenylalanine - | *F | Internal | ■■■■...■■■■ |
Special AAs | D-Proline - | *P | Internal | ■■■■...■■■■ |
Special AAs | D-Serine - | *S | Internal | ■■■■...■■■■ |
Special AAs | D-Threonine - | *T | Internal | ■■■■...■■■■ |
Special AAs | D-Tryptophan - | *W | Internal | ■■■■...■■■■ |
Special AAs | D-Tyrosine - | *Y | Internal | ■■■■...■■■■ |
Special AAs | D-Valine - | *V | Internal | ■■■■...■■■■ |
Special AAs | Formylglycine - | [G(CHO)] | Internal | ■■■■...■■■■ |
Special AAs | gamma-Carboxyglutamic acid - Gla | [Gla] | Internal | ■■■■...■■■■ |
Special AAs | Gamma-Glu, amidated - Gla, amidation at side chain | [Gla amide] | Internal | ■■■■...■■■■ |
Special AAs | Glu gamma-methyl ester - Glu(OMe) | [E(OMe)] | Internal | ■■■■...■■■■ |
Special AAs | Homo-Citrulline - | [Homocit] | Internal | ■■■■...■■■■ |
Special AAs | Homophenylalanine - | [HomoF] | Internal | ■■■■...■■■■ |
Special AAs | Homoserine - | [Homoser] | Internal | ■■■■...■■■■ |
Special AAs | Hydroxy-Proline - Hyp | [Hyp] | Internal | ■■■■...■■■■ |
Special AAs | Hyposine - Hpu | [Hpu] | Internal | ■■■■...■■■■ |
Special AAs | Isoaspartic acid - | 0 | Internal | ■■■■...■■■■ |
Special AAs | Kynurenine - Kyn | [Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptoacetic acid - Thioglycolic acid | [Mercaptoacetyl] | Internal | ■■■■...■■■■ |
Special AAs | Mercaptopropionic acid - Mpa | [Mpa] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfone - | [M(O2)] | Internal | ■■■■...■■■■ |
Special AAs | Methionine-sulfoxide - | [M(O)] | Internal | ■■■■...■■■■ |
Special AAs | N-Formylkynurenine - | [Formyl-Kyn] | Internal | ■■■■...■■■■ |
Special AAs | Norleucine - | [Nle] | Internal | ■■■■...■■■■ |
Special AAs | Norvaline - 2-Aminopentanoic acid, Nva | [Nva] | Internal | ■■■■...■■■■ |
Special AAs | Octahydroindole carbox. acid - | [Oic] | Internal | ■■■■...■■■■ |
Special AAs | Ornitine - | [Orn] | Internal | ■■■■...■■■■ |
Special AAs | para-Acetylphenylalanine - pAF | [para-Ac-Phe] | Internal | ■■■■...■■■■ |
Special AAs | Phenylglycine - Phg | [Phg] | Internal | ■■■■...■■■■ |
Special AAs | Pyroglutamic acid - Pyr | [Pyr] | Internal | ■■■■...■■■■ |
Special AAs | Selenocysteine - Sec | [Sec] | Internal | ■■■■...■■■■ |
Special AAs | β-Alanine - | [β-A] | Internal | ■■■■...■■■■ |
Special AAs | β-Cyclohexyl-Alanine - Cha | [Cha] | Internal | ■■■■...■■■■ |
Special AAs | β-cyclohexyl-D-Alanine - D-Cha | [D-Cha] | Internal | ■■■■...■■■■ |
Special AAs | Tetrahydroisquinol.carb.acid - Tic | [Tic] | Internal | ■■■■...■■■■ |
Special AAs | Thiazolidine-4-carboxylic acid - Thz | [Thz] | Internal | ■■■■...■■■■ |
Special AAs | γ-Glutamic acid - | [γ-E] | Internal | ■■■■...■■■■ |
PTM | 1-Methyl-Tryptophan - | [W(1-Me)] | Internal | ■■■■...■■■■ |
PTM | 2,6-Dimethyl-Tyrosine - | [Y(Me2)] | Internal | ■■■■...■■■■ |
PTM | 2-Methylproline - | [P(2-Me)] | Internal | ■■■■...■■■■ |
PTM | 2-Methyl-Tryptophan - | [2-Me-Trp] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (asym.) - | [R(Me)2-asym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Arginine (sym.) - | [R(Me)2-sym] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Cysteine - Penicillamine | [Pen] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Lysine - | [K(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Phenylalanine - | [F(Me)2] | Internal | ■■■■...■■■■ |
PTM | Dimethyl-Valine - | [V(Me)2] | Internal | ■■■■...■■■■ |
PTM | Methyl-Arginine - | [R(Me)] | Internal | ■■■■...■■■■ |
PTM | Methyl-Lysine - | [K(Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Alanine - | [Ala(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Glycine - | [G(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Leucine - | [L(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Phenylalanine - | [F(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Tyrosine - | [Y(N-Me)] | Internal | ■■■■...■■■■ |
PTM | N-Methyl-Valine - | [V(N-Me)] | Internal | ■■■■...■■■■ |
PTM | Phosphono-Phenylalanine - | [F(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Serine - | [S(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Threonin - | [T(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | Phospho-Tyrosin - | [Y(PO3H2)] | Internal | ■■■■...■■■■ |
PTM | β-GalNAc-Threonine - | [T(β-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Asparagine - | [N(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Serine - | [S(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | β-GlcNAc-Threonine - | [T(β-GlcNAc)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Serine - | [S(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Threonine - | [T(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Sulfuryl-Tyrosine - | [Y(SO3H)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Arginine - | [R(Me3)] | Internal | ■■■■...■■■■ |
PTM | Trimethyl-Lysine - | [K(Me)3] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Serine - | [S(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-GalNAc-Threonine - | [T(α-GalNAc)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Serine - | [S(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Mannosyl-Threonine - | [T(α-Mannosyl)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Aspartic acid - | [D(α-Me)] | Internal | ■■■■...■■■■ |
PTM | α-Methyl Glutamic acid - | [E(α-Me)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (13C3;15N) - | [A(13C3; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ala (D3) - | [A(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6) - | [R(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Arg (13C6;15N4) - | [R(13C6; 15N4)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asn (13C4;15N2) - | [N(13C4; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Asp (13C4;15N - | [D(13C4; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gln (13C5;15N2) - | [Q(13C5;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Glu (13C5;15N) - | [E(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Gly (13C2;15N) - | [G(13C2; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy His (15N3) - | [His(15N3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ile (13C6;15N) - | [I(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C1) - | [L(13C1)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6) - | [L(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (13C6;15N) - | [L(13C6; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D10) - | [L(D10)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Leu (D3) - | [L(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6) - | [K(13C6)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Lys (13C6;15N2) - | [K(13C6; 15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Phe (13C9;15N) - | [F(13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Pro (13C5;15N) - | [P(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (13C3;15N) - | [S (13C3;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Ser (D3) - | [S(D3)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Thr(13C4;15N) - | [T(13C4;15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Trp (13C11;15N2) - | [W (13C11;15N2)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Tyr (13C9;15N) - | [Y (13C9; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5) - | [V(13C5)] | Internal | ■■■■...■■■■ |
Isotopic | Heavy Val (13C5;15N) - | [V(13C5; 15N)] | Internal | ■■■■...■■■■ |
Isotopic | L-tyrosine-phenyl-d4 - deuterated Tyr | [Tyr(D4)] | Internal | ■■■■...■■■■ |
Isotopic | TMT6-126 - | TMT6-126-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-127 - | TMT6-127-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-128 - | TMT6-128-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-129 - | TMT6-129-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-130 - | TMT6-130-NH | N-term | ■□□□...□□□□ |
Isotopic | TMT6-131 - | TMT6-131-NH | N-term | ■□□□...□□□□ |
Immunization | Blue Carrier Protein - | CO-NH -Blue Carrier Protein | C-term | □□□□...□□□■ |
Immunization | BSA - | CO-BSA | C-term | □□□□...□□□■ |
Immunization | BSA via Glutaraldehyde - | BSA-NH | N-term | ■□□□...□□□□ |
Immunization | Cys(BSA) - | [C(BSA)] | Internal | ■■■■...■■■■ |
Immunization | Cys(KLH) - | [C(KLH)] | Internal | ■■■■...■■■■ |
Immunization | Cys(OVA) - | [C(OVA)] | Internal | ■■■■...■■■■ |
Immunization | KLH - | CO-KLH | C-term | □□□□...□□□■ |
Immunization | KLH via Glutaraldehyde - | KLH-NH | N-term | ■□□□...□□□□ |
Immunization | Lys(BSA) - | [K(BSA] | Internal | ■■■■...■■■■ |
Immunization | Lys(KLH) - | [K(KLH] | Internal | ■■■■...■■■■ |
Immunization | MAP 16 - Crude 8-20 aa, at C-term. only | MAP 16 | C-term | □□□□...□□□■ |
Immunization | MAP 4 - Crude 8-20 aa, at C-term. only | MAP 4 | C-term | □□□□...□□□■ |
Immunization | MAP 8 - Crude 8-20 aa, at C-term. only | MAP 8 | C-term | □□□□...□□□■ |
Immunization | OVA - | CO-OVA | C-term | □□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | 3-Azido-Alanine - Aza | [Aza] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | 5-Azidopentanoyl - 5-Azidovalerianoyl | 5-Azidopentanoyl-NH | N-term | ■□□□...□□□□ |
Cyclization/Dimerization/Special Synthesis | 6-Azido-Norleucine - Nle(N3) | [Nle(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azidohomoalanine - Aha/ Dab(N3) | [Aha] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Azido-Lysine - | [K(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (C-C), disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (D-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (E-K), amide bond - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (K-C-terminus) - side chain of K to C-terminus | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Cyclisation (NH-CO), homodetic - | [ ] | N-term | ■□□□...□□□■ |
Cyclization/Dimerization/Special Synthesis | Double disulfide - | [C cyclo(S-S)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | D-Propargylglycine - D-Pra | [D-Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Hetero-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Homo-Dimer - | [ ] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Mixed amino acids - | [X] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | para-Azido-Phenylalanine - | [F(N3)] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylalanine - alpha-Me-L-Ala(Pentynyl)-OH | [Propargylala] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Propargylglycine - Pra | [Pra] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Asymmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Side chain coupling to Lys - Symmetric Sequence | [K] | Internal | ■■■■...■■■■ |
Cyclization/Dimerization/Special Synthesis | Site directed disulfide bridge - Cyclisation (C-C) | [Cys cyclo(S-S)] | Internal | ■■■■...■■■■ |
A. Our standard peptides are delivered lyophilized with trifluoroacetate (TFA) as a counterion and packaged in glass vials, stored under argon to protect from oxygen. For additional formats, such as delivery in low-binding plastics or 2-D barcoded plates, or in-solution with different solvents, please inquire.
A. The detailed spectra are included on the Certificate of Analysis for your records. Additional quality controls such as amino acid analysis (AAA) and further nonstandard analysis are available on request.
A. We recommend preparing single-use aliquots and storing the products immediately at –20°C, upon receipt. In this manner, the lyophilized peptides can be stable for several years. Solubilized peptides should be used immediately because they are unstable (the lower the concentration, the more unstable the peptides are). Any remaining peptides in solution should be re-lyophilized for longer storage.
A. Always wear gloves when working with peptides to avoid contamination (e.g., enzymatic, bacterial, etc.). As many peptides are light-sensitive, they should also be protected from direct light. Avoid repeated freeze-thaw cycles.
A. The ability to synthesize the peptide and its solubility, stability, and overall charge are discussed in the Protein Biology Learning Center under the Peptide Design
A. All the information required to get an accurate quote is included in the Quote Request form.
A. Peptides are complex biomolecules, and each peptide has unique chemical and physical properties because of the unique amino acid composition. Although some peptides are easy to dissolve in aqueous solutions, peptides are often insoluble, especially when they contain long stretches of hydrophobic amino acids.
Because of the unique solubility of each peptide, we recommend first testing the solubilization of each peptide with a small amount of product.
Contact us using these email addresses, or download and send the quote form.
Europe, Middle East, and Africa: europeservicespeptides@thermofisher.com
Rest of World (incl. N. America): peps@thermofisher.com