Search Thermo Fisher Scientific
by Suzanne M. Smith, M.S.; Kay K. Opperman, Ph.D.; Barbara J. Kaboord, Ph.D.; Rizwan Farooqui, Ph.D. - 04/05/11
The Ras superfamily of small GTPases serve as molecular switches to control diverse eukaryotic cellular behaviors, including cell growth, differentiation and motility. Consequently, small GTPases are involved in several disease states such as cancer and metabolic disorders.1,2 GTPases are active when bound to guanosine triphosphate (GTP) and inactive when the triphosphate is hydrolyzed to guanosine diphosphate (GDP). The Thermo Scientific Pierce Active GTPase Pull-down and Detection Kits enable GTPase activation studies by preferentially enriching their active form. These kits contain a GST-protein binding-domain (PBD or RBD) fusion that is selective for active Rho, Ras, Rac1, Cdc42, Rap1, Arf1 or Arf6 (Table 1).
This pull-down method is based on the affinity of known downstream effector proteins for the active forms of specific GTPases. The respective protein-binding domain (PBD) of these downstream effectors is expressed as a GST-fusion protein (Table 1). When immobilized on glutathione agarose resin, the PBD will bind active, GTP-bound GTPase from a cell lysate (Figure 1). The pulled-down active GTPase is detected via Western blotting. As a control, cell lysates can be treated with GTPgS, which is a non-hydrolyzable analog of GTP. This method traps all GTPases in the active form and results in high GTPase enrichment. As a negative control, cell lysates are treated with an excess of GDP to shift the majority of GTPase to the inactive state.
Each active GTPase kit includes a GST fusion of the protein-binding domain
GTPase | Downstream effector binding domain | Cellular function |
---|---|---|
Rho | GST-Rhotekin-RBD | Filopodia, lamellipodia formation, and stress fibers |
Ras | GST-Raf1-RBD | Cell proliferation and differentiation |
Rac1 | GST-Pak1-PBD | Filopodia, lamellipodia formation, and stress fibers |
Cdc42 | ||
Rap1 | GST-RalGDS-RBD | Cell proliferation and differentiation |
Arf1 | GST-GGA3-PBD-PBD | Assembly of coat proteins onto budding vesicles on trans-golgi network and endosomes |
Arf6 | Membrane traffic, actin remodeling and structural organization at the cell surface | |
Cellular functions identified in this table are based on cited references 3-7. |
Figure 1. Thermo Scientific Active GTPase Pull-down and Detection Kit protocol summary.
To determine the specificity and function of the GTPase pull-down and detection kits, NIH 3T3 cell lysate was incubated with either GTPγS or GDP to activate or inactivate endogenous GTPases, respectively. The specific GST-PBD or -RBD was used to pull down active Rho, Ras, Rac1, Cdc42, Rap1, Arf1 or Arf6. A strong signal is detected in the GTPγS-treated lysate; however, minimal or no signal is detected in the GDP-treated lysate (Figure 2). These results illustrate the specificity of the PBD for active GTPases.
Figure 2. Specific detection of active Rho, Ras, Rac1, Cdc42, Rap1, Arf1 and Arf6 by western blotting. NIH 3T3 cell lysate treated with GTPγS or GDP was incubated with the appropriate GST binding domain and immobilized glutathione resin. Eluted samples and a portion of the lysate were analyzed by Western blot using GTPase-specific antibodies.
The pull-down of endogenous active small GTPases after growth factor or serum stimulations was highly effective in a variety of cell types derived from different species (Figure 3). Changes in the GTPase activities can be detected in time-course studies and differ with cell type and treatment. Because total GTPase levels in each lysate are constant, the amount of GTPase pulled down in each assay reflects activation rather than changes in GTPase expression levels. The activity profiles detected are similar to those reported in the literature.8-11
Figure 3. Specific, induced changes in the level of endogenously activated GTPases from a variety of cell types are easily monitored by the pull-down assay. In each panel, the top western blot shows the level of active GTPase isolated by pull-down assay; the lower western blot shows the total amount of expressed GTPase in the lysate. Densitometry was performed on the western blots and plotted graphically for each system. Panel A: Rho activity in HeLa (human) cells stimulated with EGF. Panel B: Ras activity in NIH 3T3 (murine) cells stimulated with PDGF. Panel C: Rac1 activity in NS1 (rodent) cells stimulated with NGF. Panel D: Arf1 activity in MDCK (canine) cells stimulated with HGF. Panel E: Arf6 activity in C2C12 (murine) cells stimulated with serum.
These results demonstrate the effectiveness of the GTPase pull-down and detection kits for monitoring sensitive changes in activity using time-dependent activity assays.
HeLa cells were grown in Dulbecco’s modified eagle medium (DMEM) supplemented with 10% fetal bovine serum (FBS) to ~70% confluency and then starved in 1% FBS medium for 24 hours before stimulation with 100ng/mL of epidermal growth factor (EGF) for the indicated times. NIH 3T3 cells were grown in DMEM supplemented with 10% FBS to ~70% confluency and starved in 0.1% FBS medium for 24 hours. Platelet-derived growth factor (PDGF) was added at 50ng/mL for the indicated times. NS1 cells were grown in RPMI supplemented with 10% FBS to ~70% confluency and nerve growth factor (NGF, 50ng/mL) was added for the indicated times. MDCK cells were grown in EMEM supplemented with 10% FBS to ~70% confluency and starved in serum-free medium for 48 hours before stimulation with 50ng/mL of hepatocyte growth factor (HGF) at indicated times. C2C12 cells were grown in DMEM supplemented with 10% FBS to ~70% confluency and were starved in serum-free medium for 48 hours before adding 10% serum at the indicated times.
NIH 3T3 cells were lysed on the culture plate with 1mL lysis/binding/wash buffer. The clarified cell lysate (500μg) was treated with either GTPγS (positive control) or GDP (negative control). The treated lysates (or 1mg of the endogenous time-course lysates) were incubated with 400μg GST-Rhotekin-RBD (for active Rho), 80μg GST-Raf1-RBD (for active Ras), 20μg GST-Pak1-PBD (for active Rac1 or Cdc42), 20μg GST-RalGDS-RBD (for active Rap1) or 100μg GST-GGA3-PBD (for active Arf1 or Arf6). Half of each elution was analyzed by SDS-PAGE and detected by Western blot using the specific GTPase primary antibody.
The Thermo Scientific Active Rac1 Pull-Down and Detection Kit is a complete kit for the selective enrichment and detection of GTP-bound Rac1 GTPase through specific protein interaction with the Pak1 protein-binding domain.
The Active Rac1 Pull-Down and Detection Kit includes purified GST-Pak1 protein-binding domain (PBD), glutathione agarose resin, positive and negative controls (GTPγS and GDP, respectively), lysis/binding/wash buffer, anti-Rac1 primary antibody, SDS sample buffer, spin columns and collection tubes. The kit was validated using lysates from NIH3T3 cells, a cell line that is known to have robust Rac1 activity.
Lean more about Thermo Scientific Active Rac1 Pull-Down and Detection Kit