Search Thermo Fisher Scientific
The Ion Reporter data analysis process helps ensure you get high quality data by prioritizing and annotating variants. It completes with a report that simplifies your bioinformatics path to discovery by enabling you to focus on finding the biological meaning of your data.
Ion Reporter Software removes the barriers of NGS data analysis, enabling you to focus on the meaning of your data. Streamlined analysis with integrated run planning from Torrent Suite Software automates the primary analysis, data transfer, and secondary analysis in Ion Reporter Software.
Ion Reporter Software analysis and variant calling helps you quickly focus on variants of interest supported by full annotation from >25 public and proprietary databases.
Ion Reporter Software is designed with preconfigured and customizable workflows for a variety of research applications from inherited disease to reproductive health to oncology. Workflows are flexible to enable custom optimization and can also be locked to enable controlled use without depending on the advanced skills of a bioinformatician.
The Ion Reporter Server System pairs two individual components, Ion Reporter Software and a powerful computer server, into one integrated solution right in your own building for the ultimate in secure computational resources.
Ion chip | File footprint size | Storage capacity (# runs) |
---|---|---|
Ion 314 Chip | 1 GB | 20,000 |
Ion 316 Chip | 2 GB | 10,000 |
Ion 318 Chip | 5 GB | 4,000 |
Ion 510 Chip | 5 GB | 4,000 |
Ion 520 Chip | 10 GB | 2,000 |
Ion 530 Chip | 20 GB | 1,000 |
Ion 540 Chip | 45 GB | 444 |
Ion PI Chip | 50 GB | 400 |
Number of Ion Torrent sequencer run BAM files that can be stored on the Ion Reporter Server.