Search Thermo Fisher Scientific
于津浦:BRCA为核心的多基因检测在乳腺癌/卵巢癌遗传风险评估中的应用
天津医科大学肿瘤医院肿瘤分子诊断中心的于津浦教授发表了《BRCA为核心的多基因检测在乳腺癌/卵巢癌遗传风险评估中的应用》的主题演讲。
于教授介绍BRCA1/2 基因是抑癌基因,主要参与DNA损伤的修复和转录的调控,有助于维持基因组的完整性,也是双链DNA断裂的同源重组修复途径中的关键成分。BRCA1/2基因突变会导致乳腺癌和卵巢癌的罹患风险增加,并且会以常染色体显性遗传方式传递给下一代。
于教授同时指出,BRCA1/2基因是乳腺癌/卵巢癌最重要的易感基因 ,约5%-10%乳腺癌和15%-22%的卵巢癌是由BRCA1/2基因突变导致的。BRCA1/2 基因致病性突变使女性发生乳腺癌风险提高10倍,发生卵巢癌风险提高10-30倍。因此,检测BRCA1/2基因突变,有助于遗传性乳腺癌/卵巢癌发病风险的预测及早诊,及时预防和干预,改善患者预后,提高疾病治愈率。
目前,在众多的BRCA1/2突变检测方法中,二代测序(NGS)是一种检测基因主要常见突变(包括大重排)的方法。大部分实验室中使用这种方法,可针对BRCA1/2 的全外显子靶向测序,同时检测点突变、小片段插入/缺失以及大片段基因变异。
于教授带领的团队建立了用于乳腺癌易感性筛查系统的BRCA为核心的多基因检测,并自主研发设计乳腺癌6基因测序panel: