辅料CAD:吐温+CCS-Retain AX
A highly sensitive high-performance liquid chromatography-charged aerosol detection method for the quantitative analysis of polysorbate 80 in protein solution
Goal
To describe the development of a highly sensitive charged aerosol detection (CAD) method for the determination of polysorbate 80, also known as Tween™ 80, in biopharmaceutical products.
Introduction
Polysorbates, such as polysorbate 20 and polysorbate 80, are non-ionic surfactants. They are commonly used in biotherapeutic formulations to prevent surface adsorption and stabilize proteins against aggregation induced by stress, such as agitation and shear.1 For quality control purposes, it is important to determine the concentration of polysorbate in the final products. However, the quantitative analysis of polysorbate is challenging—polysorbate is a complex mixture of many different species, which lack natural UV chromophores, and is therefore difficult to analyze by UV detection. Also, chromatographic separation often leads to peaks or peak groupings that consist of many unresolved components and poor peak shapes, thus making accurate and sensitive quantitation problematic.
在线阅读
为什么选择该应用方法?
吐温表面活性剂,使得蛋白更稳定不聚集,是常用的生物药辅料。极弱的紫外吸收,目前药典的方法都比较复杂,需要进行衍生化。
该应用有什么特点?
CAD 质量通用型检测器,是无/弱紫外吸收物质的分析利器。检测原理仅与物质气溶胶颗粒表面的电荷有关。
应用的优势有哪些?
灵敏度达到ng级,相较传统ELSD有一个数量级以上的提升。重现性较ELSD等喷雾型检测器更佳。独特检测原理使其具有响应一致性。
用了该方法可以获得哪些收益?
相较传统方法省去复杂的前处理步骤,更少的引入人为误差——检测效率高,省时省力,结果更加稳定重现性好。