LNP:CAD+ Accucure C30(): 要进行前处理,在于不同组成的脂质体,在C30上优化方案
Characterization of lipid nanoparticle (LNP) composition using UHPLC-CAD
Application benefits
  • Simple UHPLC method for characterization of the lipid components of LNPs
  • Universal response shows relative amounts of each lipid compound in the formulation
  • Low limits of detection even forcholesterol
  • Wide linear range for all lipid compounds
  • Baseline separation ofcomponents
Goal
Development of a simple UHPLC method for the characterization of the lipid content ot LNP formulations.Implementation of the universal CAD detector to quantify large non- chromophoric lipids present in the LNP structure.
在线阅读
为什么选择该应用方法?
LNPs是弱紫外吸收,或无紫外吸收,其含量、杂质等表征是递送体系的核心CQA之一。
该应用有什么特点?
C30 官能团,键合在核壳基质上,我们专有的键合方式和技术);CAD 质量通用型检测器,是无/弱紫外吸收物质的分析利器。检测原理仅与物质气溶胶颗粒表面的电荷有关。
应用的优势有哪些?
保证良好的通用性,可以满足不同种类配方LNP分离要求(AN做了2个配方的LNP)CAD相较传统ELSD在、灵敏度、重现性、响应一致性都有显著提升,不仅适用于LNP的含量检测,还适用于LNP的杂质检测及LNP的表征分析。
用了该方法可以获得哪些收益?
攻克LNP核心工艺开发,保驾护航。