mRNA的纯度:反相和IEC。工艺过程中mRNA纯度的检测,纯化前或纯化后都可以用
Simultaneous reversed-phase ano Ianion-exchange method scouting with a dual syste em for mRNA impurity determination
Application benefits
  • Leverage two different separation chemistries in the same instrument at the same time: ion-exchange(IEX)andion-pairing reversed-phase(IP-RP)
  • Scout up to 10 solvents for each column type to find most suitable eluent condition
  • Accelerate method development
  • Maximize sample knowledge
  • Perform mRNA profiling and detection of post-in vitrotranscription(IVT) purification impurities
  • Use corrosive mobile phases,such as NaCl 1M,with thefully biocompatible flow path of the Thermo ScientificVanquishDuo UHPLC system
Goal
Determine the most suitable conditions for the detection of post-transcriptiona impurities in mRNA with a time-effective scouting approach
在线阅读
为什么选择该应用方法?
mRNA纯度表征方法:如反相、离子交换等,同时需要两套仪器进行检测。
该应用有什么特点?
DNA pac RP:极具代表性的核酸专用柱,独特的键合方式; DNA Pac 200 RS(Check AN):阴离子交换-核酸专用柱,专有的包裹技术,和官能团键合方法;Vanquish Duo 具有双针进样器、双泵,可以实现一套仪器同时进行两种工作流程。
应用的优势有哪些?
DANpac RP:从小到10-20 nt 寡核苷酸,达到几千nt的核酸都适用;DNA Pac 200 RS(Check AN): 无孔基质对于核酸的碱基对个数没有要求,适用范围更宽。提升分析效率,节省了实验室空间,减少仪器成本。
用了该方法可以获得哪些收益?
专用的反向和离子交换模式,双维度监控mRNA纯化,其对于工艺优化筛选和质量监控尤为适用。