Search Thermo Fisher Scientific
A major challenge for healthcare providers and geneticists is to learn how SARS-CoV-2, the coronavirus that causes the COVID-19 disease, interacts with the human genome and to stratify populations in order to understand disease susceptibility, severity, and outcomes.
We have launched our new Axiom Human Genotyping SARS-CoV-2 Research Array.
Axiom genotyping arrays and associated modules for human genotyping research can be used in various aspects of SARS-CoV-2 infection research:
Category | Number of Markers* |
SARS-CoV-2 Susceptibility Research Variant Module | >135,000 |
SARS-CoV-2 putative receptor variants | >1,200 |
Signaling pathway variants | >130,000 |
Variants on X-Chromosome | >60,000 |
SARS-CoV-2 Severity Research Module – Underlying conditions implicated in severity of SARS-CoV-2 infections | >16,000 |
Blood phenotypes and blood groups | >1,200 |
Diabetes | >800 |
Cardiovascular disease | >1,300 |
Stroke | >50 |
Lung function and COPD | >9,000 |
BMI | >1,000 |
CKD/RAAS | >450 |
Asthma | >500 |
SARS-CoV-2 Immune Response and Therapeutic Treatment Research Variants Module | >24,000 |
ADME and protein markers for druggable targets | >6,000 |
Immunoglobulin and miRNA markers for vaccine research | >12,000 |
HLA/KIR markers for immune response research after vaccination | >9,000 |
Multi-ethnic Genome Wide Association Study (GWAS) Grid | >725,000 |
ACMG 59 Variants | >6,000 |
Functional coding-region variants including eQTL and loss-of-function variants | >24,000 |
A marker may appear in more than one category.
For Research Use Only. Not for use in diagnostic procedures.