Search Thermo Fisher Scientific
TaqMan® array cards and plates contain TaqMan® Gene Expression Assays in 96-well TaqMan® Array plate, 384-well TaqMan® Array card, and OpenArray® formats. These formats are suited for gene expression screening and validation applications that require analysis of tens to thousands of targets. Choose from a broad selection of pre-configured content or design your own custom card or plate.
Not sure which format is right for you?
Visit the TaqMan® Gene Expression Assays & Arrays Selection Guide >
View the Instrument Compatibility Guide >
TaqMan® array cards and plates are pre-configured with the most relevant TaqMan® Gene Expression Assays for specific biological pathways, processes or diseases. Click below to browse our offering of pre-configured array cards and plates.
By Disease
By Pathway
By Biological Process
Endogenous Controls
If your specific assays are not available as pre-configured TaqMan® Arrays, you can configure your own custom TaqMan® Arrays using the links below, or take advantage of the Custom Plating Service. Contact your local Life Technologies Sales Representative for further assistance.
Custom TaqMan® Array cards and plates are flexible, affordable, and convenient for gene expression analysis screening of biological samples. TaqMan® Assays can be configured into 384-well TaqMan® Array microfluidic cards, or 96-well plates, using our easy ordering interface.
Need a custom TaqMan Array solution that includes customized TaqMan® Gene Expression Assays, TaqMan SNP Genotyping Assays, TaqMan Copy Number Assays, TaqMan miRNA Assays, or combination assays? The TaqMan® Custom Plating service provides TaqMan® Assays in 96- or 384-well plates, or 384-well microfluidic cards configured to your specifications.
Product Format/ Instrument | StepOne Plus™ | StepOne™ | 7500 | 7500 Fast | 7900 | ViiA7™ | QuantStudio™ |
96-well Plate, Standard | No | No | 96-well Block | No | 96-well Block | 96-well Block | 96-well Block |
96-well Plate, Fast | 96-well Fast Block | No | No | 96-well Fast Block | 96-well Fast Block | 96-well Fast Block | 96-well Fast Block |
384-well Plate | No | No | No | No | 384-well Block | 384-well Block | 384-well Block |
Microfluidic Cards | No | No | No | No | TaqMan® Array Card Block | TaqMan® Array Card Block | TaqMan® Array Card Block |
OpenArray® | No | No | No | No | No | No | OpenArray® Block |
Regulatory Status: For Research Use Only. Not for human or animal therapeutic or diagnostic use.