Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The CTS DynaCellect Magnetic Separation System is a closed, automated isolation, activation, and bead removal system for cell therapy development and manufacturing. Together with fit-for-purpose consumables, it delivers high cell purity, recovery, and viability. As a standalone instrument or as part of a workflow, the operator-independent system will help you isolate the right cells and minimize failures in manufacturing while helping provide increased robustness and precision.
The Gibco CTS DynaCellect Magnetic Separation System has been recognized as a 2023 Winner of the following industry awards:
Designed to be used with magnetic beads, the CTS DynaCellect Magnetic Separation System consists of an intuitive, programmable interface, integrated magnet-rocker, and fluidics panel that will help you achieve exceptional cell recovery, a wide range of reaction volumes, optimal efficiency, and a cell purity rate of greater than 95%, while maintaining cell viability. Together with process flexibility, sterile single-use consumables, and an available software upgrade for 21 CFR Part 11 compliance, this system is designed to help you easily scale from development to clinical and commercial manufacturing.
The DynaCellect Magnetic Separation System is designed to deliver exceptional isolation efficiency, purity, and cell recovery while maintaining cell viability.
The DynaCellect Magnetic Separation System and fit-for-purpose consumables are GMP-manufactured, automated, and work in a closed system for cell therapy manufacturing.
Lock down protocols for commercial manufacturing, and add electronic record-keeping software to enable FDA 21 CFR Part 11 compliance.
See media and reagents that are compatible with the DynaCellect system.
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals