Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Single point hotplate stirrers are available in basic, standard and advanced models, offering various top plate materials and multiple sizes for all applications in the general lab.
Find the hot plate stirrer to meet your needs using our hot plate stirrer selection guide ›
Cimarec Basic Stirring Hotplates | ||
---|---|---|
Combine performance and economy with the sleek, rugged design of Thermo Scientific Cimarec Basic Stirring Hotplates, ideal for general-purpose stirring at speeds up to 2500rpm. | ||
Part no. | SP194715 | SP195025 |
Plate material | Ceramic | Ceramic |
Plate shape | Square | Square |
Plate size | 4.25 x 4.25 in. (10.8 x 10.8cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) |
Voltage | 120V | 120V |
Speed | 250 to 2500rpm | 250 to 2500rpm |
Temperature Range | 150° to 538°C | 150° to 538°C |
RT2 Basic Hotplate Stirrer | ||
---|---|---|
Rely on safety and consistent stirring speed with the Thermo Scientific RT2 Basic hotplate stirrer. Benefit from exceptionally strong magnet coupling power and a smooth-start stirring mechanism. Experience fast heat-up time and an excellent heat transfer rate thanks to the tightly integrated structure of the heater and a ceramic-coated aluminum alloy top plate which provides excellent temperature uniformity and corrosion resistance while allowing for optimal monitoring of color changes in samples or media during operation. | ||
Part no. | 88880003 | 88880004 |
Plate material | Ceramic-coated aluminum | Ceramic-coated aluminum |
Plate shape | Round | Round |
Plate size | 5.5 in. (140mm) dia. | 5.5 in. (140mm) dia. |
Voltage | 230V | 120V |
Speed | 50 to 2,000rpm | 50 to 2,000rpm |
Temperature Range | 50° to 350°C | 50° to 350°C |
Capacity | 20L | 20L |
5.5 in. (140mm) dia. |
Cimarec+ Stirring Hotplates Series | ||||
---|---|---|---|---|
Our Thermo Scientific Cimarec+ series digital stirring hotplates offer precise stirring control, exceptional safety and temperature performance for your routine protocols. The Cimarec+ Series are available in three sizes, providing flexibility from microscale chemistries to production operations. | ||||
Part no. | ||||
Plate material | Ceramic | Ceramic | Aluminum | Ceramic |
Plate shape | Square | Square | Square | Square |
Plate size | 4.25 x 4.25 in. (10.8 x 10.8cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
Speed | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm |
Max Temperature | 540°C | 540°C | 300°C | 400°C |
Capacity | 1L | 4L | 4L | 6L |
SuperNuova+ Stirring Hotplates Series | ||||||
---|---|---|---|---|---|---|
Our Thermo Scientific SuperNuova+ stirring hotplate series provides reliable performance, dependable safety and simple operation. Units are available in two sizes, including a multi-position model with optimized controls and settings for applications that demand advanced precision. | ||||||
Part no. | SP88857190 | SP88857195 | SP88857194 | SP88857198 | SP88850190 | SP88850195 |
Plate material | Ceramic | Ceramic | Aluminum | Aluminum | Ceramic | Ceramic |
Plate shape | Square | Square | Square | Square | Square | Square |
Plate size | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 7.25 x 7.25 in. (18.4 x 18.4cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) | 10.25 x 10.25 in. (26.0 x 26.0cm) |
Voltage | 100-120V | 230V | 100-120V | 230V | 100-120V | 230V |
Speed | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm | 50 to 1,500rpm |
Max Temperature | 450°C | 450°C | 300°C | 300°C | 450°C | 450°C |
Capacity | 4L | 4L | 4L | 4L | 6L | 6L |
RT2 Advanced Hotplate Stirrer | ||
---|---|---|
It’s all about precise control without sacrificing your budget with the sleek, low profile Thermo Scientific RT2 Advanced Digital Hotplate Stirrer which offers digital temperature and speed control with 0.1°C temperature resolution and stir speed range of 30 to 2000rpm. Choose your heat-up rate and experience an excellent heat transfer due to the tightly integrated structure of the heater and a ceramic-coated aluminum alloy top plate that also provides excellent uniformity and corrosion resistance while allowing for optimal monitoring of color changes in samples or media during operation. | ||
Part no. | 88880006 | 88880005 |
Plate material | Ceramic-coated aluminum | Ceramic-coated aluminum |
Plate shape | Round | Round |
Plate size | 5.5 in. (140mm) dia. | 5.5 in. (140mm) dia. |
Voltage | 120V | 230V |
Speed | 30 to 2,000rpm | 30 to 2,000rpm |
Temperature Range | 50° to 350°C | 50° to 350°C |
Capacity | 20L | 20L |
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.