Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Optimize your experiments to get the best results. We’ve compiled a detailed knowledge base of the top tips and tricks to meet your analyses needs.
View the relevant questions below:
Having problems with your experiment? Visit
No. However, it is a good idea to calibrate all scan settings at least once a year. In this way, if the zoom scan setting is used in the future, it can be reasonably easy to calibrate at that time.
Scan Mode | LCQ Fleet Scan Rate (amu/s) | LTQ & LXQ Scan Rate (amu/s) | LTQ Velos Scan Rate (amu/s) | LTQ Velos Pro Scan Rate (amu/s) |
---|---|---|---|---|
Rapid | NA | NA | NA | 66.000 |
Normal | 12.500 | 16.700 | 33.300 | 33.300 |
Enhanced | 5.000 | 5.000 | 5.000 | 5.000 |
Zoom | 1.100 | 1.100 | 1.100 | 1.100 |
Ultra Zoom | NA | 27 | 27 | 27 |
Turbo | 125.000 | 125.000 | 125.000 | 125.000 |
The tune file contains four items: Source parameters, ion optic parameters, AGC targets, and scan time settings.
Need more information? Contact us ›
For Research Use Only. Not for use in diagnostic procedures.