Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
The Applied Biosystems VeritiPro Thermal Cycler delivers proven reliability with advanced temperature control technology and connectivity. Take advantage of next-level PCR optimization with the precision offered by VeriFlex Block technology. Connect to the cloud-enabled VeritiPro Thermal Cycler remotely. Conveniently design and securely upload your methods, monitor runs, and check instrument availability from any mobile device or desktop computer with Connect.
The VeriFlex Block technology inside VeritiPro Thermal Cycler allows you to set block temperatures your way for PCR optimization experiments or reactions with specific temperature requirements, such as enzyme studies and restriction digests.
Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.
The Applied Biosystems Veriti Thermal Cycler set a new standard for innovation when it launched in 2007. It was the first thermal cycler to employ an LCD touchscreen interface, multiple temperature zone block, and web-enabled remote-control software. Building on these innnovations, the VeritiPro Thermal Cycler upgrade features the latest block technology and connectivity options. These advancements provide higher ramp rates, quieter runs, and more intuitive Applied Biosystems VeriFlex temperature optimization setup while continuing to offer the same high-end performance.
The VeriFlex Block technology inside VeritiPro Thermal Cycler allows you to set block temperatures your way for PCR optimization experiments or reactions with specific temperature requirements, such as enzyme studies and restriction digests.
Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall
Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for any experiment.
The Applied Biosystems Veriti Thermal Cycler set a new standard for innovation when it launched in 2007. It was the first thermal cycler to employ an LCD touchscreen interface, multiple temperature zone block, and web-enabled remote-control software. Building on these innnovations, the VeritiPro Thermal Cycler upgrade features the latest block technology and connectivity options. These advancements provide higher ramp rates, quieter runs, and more intuitive Applied Biosystems VeriFlex temperature optimization setup while continuing to offer the same high-end performance.