Search Thermo Fisher Scientific
Our array of custom antibody services includes a customizable option for guinea pig immunization, serum collection, and titer analysis for guinea pig antibody production, plus several extension possibilities.
This guinea pig immunization schedule uses two Hartley Pigmented guinea pigs for each immunogen. Animals are housed individually in separate cages. Injections are subcutaneous (SQ) as emulsions in Complete Freund's Adjuvant (CFA) or Incomplete Freund's Adjuvant (IFA); alternative adjuvants can be used upon request. Customizations to the protocol are available. For example, you can choose the number of guinea pigs needed and you can adjust the length of your protocol beyond the 80-day immunization schedule to meet your research goals. Please contact us to discuss the options in detail. Antigen preparation, such as peptide synthesis and/or immunogen conjugation, occurs before Day 0. Protocol Days are approximate (± 2 days).
Procedure | Protocol day | Description |
---|---|---|
Control serum collection | Day 0 | Pre-immune bleed (~1 mL per guinea pig) |
Primary injection | Day 1 | Immunize with 100 µg antigen in CFA, SQ 4 sites |
1st booster | Day 21 | Boost with 50 µg antigen in IFA, SQ 4 sites |
2nd booster | Day 42 | Boost with 50 µg antigen in IFA, SQ 4 sites |
Test bleed | Day 50 | Test-bleed (~1 mL per guinea pig) |
ELISA and shipping | Day 60 | ELISA titration of pre-immune and test bleed; (results available online) |
3rd booster | Day 62 | Boost with 50 µg antigen in IFA, SQ 4 sites |
Decision | Day 64 | Verify disposition of animals; decide to continue or terminate |
Instruction due date | Day 80 | Terminal bleed (exsanguination, yielding 10–15 mL per guinea pig) and termination of animals if instructions not received |
Deliverables: 8 vials crude antibody sera