Search Thermo Fisher Scientific
The Invitrogen Vivid Colors Fluorescent Protein Vectors are our next-generation cloning and expression tools featuring Aequoreavictoria-derived fluorescent proteins (FP) for simple, non-invasive detection of recombinant proteins. With Invitrogen Gateway and TOPO cloning vector options, cloning, expression, and viewing has never been so easy!
Expression of the Invitrogen Vivid Colors EmGFP and YFP fusion proteins can be detected by fluorescence microscopy or flow cytometry using standard FITC filter sets. For optimal visibility, it is recommended that you use a filter set optimized for detection within the excitation and emission ranges for each fluorescent protein (see table below).
Recommended filters | ||||||
---|---|---|---|---|---|---|
Protein | Color | Excitation | Emission | Mutations | Omega | Chroma |
EmGFP | Green | 487 | 509 | S65T, S72A, N149K, M153T, I167T | XF100 | |
YFP | Yellow | 514 | 527 | S65G, S72A, K79R, T203Y | XF1042 | 41028 |
CFP | Cyan | 452 | 505 | K26R, Y66W, N146I, M153T, V163A, N164H | XF114 | 31044 |
BFP | Blue | 308-383 | 440-447 | F64L, Y66H, Y145F, V163A , N198S | XF10 | 31021 |
Cycle 3 GFP | Green | 395478 | 507 | F64L, F100S, M154T, V164A | XF76 | |
These Invitrogen Vivid Colors vectors are for generation of N-terminal or C-terminal fluorescent protein fusions to your gene-of-interest (GOI). Emerald Green (EmGFP) and Yellow (YFP) Fluorescent Protein versions are available. To create a Vivid Colors fusion with protein, you can employ either Gateway or TOPO cloning technology. Invitrogen Gateway Destination Vectors require the use of an entry clone, while the TOPO vectors require only a PCR product of your GOI (Invitrogen TA Cloning). Regardless of the cloning techniques you select, the Invitrogen Ultimate ORF collection is a comprehensive and convenient source of full-length cDNA clones for creating fusion proteins.
To express your gene with alternative promoters, or with other selectable markers for stable-cell generation, consider the Invitrogen pTracer vectors series. These vectors express the Cycle 3 Green Fluorescent Protein (Cycle 3 GFP) which has spectral excitation and emission wavelengths identical to wild-type GFP. Although nearly as bright as the Vivid Colors FPs, you’ll need to use the proper filter set to observe the fully-enhanced brightness.
Highly specific antibodies to Aequorea victoria fluorescent proteins are available, including polyclonal anti-GFP (rabbit serum) and anti-GFP (rabbit IgG fraction), for western blot detection. These antibodies have been tested and are shown to react specifically and strongly to both Vivid Colors EmGFP and YFP. A number of different Invitrogen Molecular Probes anti-GFP antibody conjugates is available, offering you a choice of multiple detection technologies you can employ.
Anti-GFP; Rabbit polyclonal antiserum (A6455)
Anti-GFP; Rabbit polyclonal IgG fraction (A11122)
Anti-GFP; Invitrogen ABfinity recombinant rabbit monoclonal antibody (G10362)
Gene of Interest | Mammalian | |||||
---|---|---|---|---|---|---|
Description | Reporter | Expr. Format | Cloning | Promoter | Tags | Selection |
Vivid Colors pcDNA 6.2/C-EmGFP-DEST | Emerald GFP | C-terminal FP fusion | Gateway | CMV | bsd | |
Vivid Colors pcDNA 6.2/N-EmGFP-DEST | Emerald GFP | N-terminal FP fusion | Gateway | CMV | N-term.V5 | bsd |
Vivid Colors pcDNA 6.2/C-YFP-DEST | Yellow | C-terminal FP fusion | Gateway | CMV | bsd | |
Vivid Colors pcDNA 6.2/N-YFP-DEST | Yellow | N-terminal FP fusion | Gateway | CMV | N-term.V5 | bsd |
Vivid Colors pcDNA 6.2/EmGFP-Bsd/V5-DEST | Emerald GFP | native | Gateway | CMV | GFP-bsd fusion | |
Vivid Colors pcDNA 6.2/C-EmGFP-GW/TOPO Mam. Expr. Vector | Emerald GFP | C-terminal FP fusion | TOPO | CMV | bsd | |
Vivid Colors pcDNA 6.2/N-EmGFP-GW/TOPO Mam. Expr. Vector | Emerald GFP | N-terminal FP fusion | TOPO | CMV | N-term.V5 | bsd |
Vivid Colors pcDNA 6.2/C-YFP-GW/TOPO Mam. Expr. Vector | Yellow | C-terminal FP fusion | TOPO | CMV | bsd | |
Vivid Colors pcDNA 6.2/N-YFP-GW/TOPO Mam. Expr. Vector | Yellow | N-terminal FP fusion | TOPO | CMV | N-term.V5 | bsd |
NT-GFP Fusion TOPO TA Expression Kit | Cycle 3 GFP | N-terminal FP fusion | TOPO | CMV | G418 | |
CT-GFP Fusion TOPO TA Expression Kit | Cycle 3 GFP | C-terminal FP fusion | TOPO | CMV | G418 | |
pcDNA-DEST47 Gateway Vector | Cycle 3 GFP | C-terminal FP fusion | Gateway | CMV | G418 | |
pcDNA-DEST53 Gateway Vector | Cycle 3 GFP | N-terminal FP fusion | Gateway | CMV | G418 | |
pBlue TOPO TA Expression Kit | beta-gal | n/a | TOPO | any | n/a | neo |
pGlow TOPO TA Expression Kit | Emerald GFP | n/a | TOPO | any | n/a | neo |
pTracer-CMV/Bsd | Cycle 3 GFP | native | R.E. | CMV | GFP-bsd fusion | |
pTracer-CMV/Bsd Kit | Cycle 3 GFP | native | R.E. | CMV | GFP-bsd fusion | |
pTracer-EF/Bsd A, B, & C | Cycle 3 GFP | native + tag | R.E. | EF-1 alpha | C-term.V5, 6xHis | GFP-bsd fusion |
pTracer-EF/Bsd Kit | Cycle 3 GFP | native + tag | R.E. | EF-1 alpha | C-term.V5, 6xHis | GFP-bsd fusion |
pTracer-CMV2 Kit | Cycle 3 GFP | native | R.E. | CMV | GFP-zeo fusion | |
pTracer-CMV2 Vector | Cycle 3 GFP | native | R.E. | CMV | GFP-zeo fusion | |
pTracer-SV40 Kit | Cycle 3 GFP | native | R.E. | SV40 | GFP-zeo fusion | |
pTracer-SV40 Vector | Cycle 3 GFP | native | R.E. | SV40 | GFP-zeo fusion | |
pTracer-EF Kit | Cycle 3 GFP | native + tag | R.E. | EF-1 alpha | C-term.V5, 6xHis | GFP-zeo fusion |
pTracer-EF A, B, & C Vector | Cycle 3 GFP | native + tag | R.E. | EF-1 alpha | C-term.V5, 6xHis | GFP-zeo fusion |
pRSET/CFP | Cyan | (n/a: bacterial expression clone) | 6xHis, Xpress, EK | n/a | ||
pRSET/BFP | Blue | (n/a: bacterial expression clone) | 6xHis, Xpress, EK | n/a | ||
pLenti6.2-GW/EmGFP Expression Control Vector | Emerald GFP | (GFP control only; no cloning site) | CMV | n/a | bsd |