Search Thermo Fisher Scientific
Applied Biosystems genetic analyzers are a trusted standard for Sanger sequencing and fragment analysis by capillary electrophoresis (CE). Our instruments add simplicity, scalability, and speed to your workflow, enabling you to analyze your most difficult samples with complete confidence. Versatile and innovative, our systems have been designed with you in mind to meet the demands of your genetic analysis needs, whether you are running a few samples or need larger scale options.
SeqStudio Genetic Analyzer | SeqStudio Flex Series Genetic Analyzer | Applied Biosystems 3500 Series Genetic Analyzer | Applied Biosystems 3730 Series Genetic Analyzer | |
Request a quote | Request a quote | Request a quote | Request a quote | |
Lowest sample throughput, easy-to-use, flexible system | Medium sample throughput, easy-to-use, flexible, connected system | Medium sample throughput, basic and automated operation system | Maximum throughput, scalability, and flexibility | |
Number of capillaries | 4 | 8 or 24 | 8 or 24 | 48 or 96 |
Capillary array length (cm) | 28 | 36, 50 | 36, 50 | 36, 50 |
Sample capacity | 1 plate; 96-well plate; 8-strip tube (compatible) | 4 plates; 96- or 384-well plates; 8-strip tube (compatible) | 2 plates; 96- or 384-well plates; 8-strip tube (compatible) | 16 plates; 96- or 384-well plates |
Polymer type | POP-1, integrated into click-in cartridge | POP-6, POP-7, POP-4 | POP-6, POP-7, POP-4 | POP-7 conformational analysis polymer |
Continual plate loading | Not applicable | Yes | No | No |
Sample re-prioritization | No | Yes | Yes | No |
Radio frequency ID | Yes | Yes | Yes | No |
Configuration | Integrated computer with touchscreen; optional desktop | Integrated computer with touchscreen; optional desktop | External desktop required | External desktop required |
Voice command capabilities | No | Yes | No | No |
Remote monitoring and data sharing | Yes | Yes | No | No |
Integrated remote troubleshooting tools | No | Yes | No | No |
Connectivity | USB, ethernet ports, and Wi-Fi dongle | USB, ethernet ports, and Wi-Fi dongle | Ethernet port | Ethernet port |
SeqStudio Genetic Analyzer for HID ›
SeqStudio Flex Genetic Analyzer HID ›
CE-IVD and IVDR compliant
3500xL Dx Genetic Analyzer ›
The Applied Biosystems SeqStudio Series systems deliver the high quality and reliable performance of Sanger sequencing and fragment analysis that scientists need, while leveraging design improvements and technological advances for increased flexibility, easier operation, enhanced connectivity, and remote serviceability.
With a wide range of applications from simple, targeted sequencing to identification of the latest SARS variant of concern, SeqStudio Series instruments help you to achieve freedom and peace of mind, providing extra time in your day to do what you do best: answering the big scientific questions.
Learn more about SeqStudio Family enabled applications. Download the SeqStudio Family Application Guide ›
For a low-scale, cost-effective option: Discover SeqStudio ›
For a mid-scale, efficient and flexible option: Discover SeqStudio Flex ›
Need high-throughput solution? Find the right analyzer for your testing needs ›
Get bite-sized answers to your everyday Sanger sequencing and fragment analysis questions
Learn about the history of sequencing and how to pick the right platform for your research needs