Search Thermo Fisher Scientific
Having difficulties with your experiment?
We are dedicated to your success. Get back on track. View our expert recommendations for commonly encountered problem scenarios.
View the relevant questions below:
Beginning your experiment?
Visit our
Primary cells are not immortalized and undergo a limited number of population doublings. Each cell type has a different maximum population doubling number that is found on the certificate of analysis for the assigned lot number. For information on “Population Doubling” vs. “Passage Number” refer to the “Getting Started” page.
Be sure to use the Coating Matrix Kit if you are using the Animal Origin–Free (AOF) supplementation for your culture. There are no attachment factors in the AOF supplements, and Coating Matrix Kit is required for cells to adhere properly.
Yes, we strongly recommended that you contact our Molecular Biology scientists at techsupport@thermofisher.com to help you choose the best transfection reagent option, as primary cells tend to be very sensitive.
It is important to store the cells, once received, in the vapor phase of liquid nitrogen. The vials are not leak proof, and submerging them into the liquid phase can allow liquid nitrogen to leak into the vial and affect the viability of the cells.
These are senescent cells, and this is normal for adult keratinocyte culture. You always have a population of cells that are old and no longer proliferate. However younger cells, which are small in size should keep proliferating. When a culture gets older, you see more and more large cells, and the culture will eventually stop growing. With the right care an attention, the culture should yield at least 25 population doublings.
It is very important to do a viability count prior to plating and to follow the recommended seeding density. The flask number mentioned in some of the protocols is a guideline so that you are aware of how many vessels should be anticipated from each vial. Cell counts on the vial are the minimum guaranteed, but there are usually more supplied to make sure that you receive the amount of cells that we promise. That being said, it is important to count the cells to make sure you have the correct seeding density and so that you can monitor the number of population doublings that your cells go through.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Improper thawing technique |
|
Sub-optimal thawing medium | Use HTM Medium during thawing to remove cryoprotectant |
Rough handling of hepatocytes during counting |
|
Improper counting technique |
|
Cells left out too long | Plate cells immediately after counting |
For additional information, please refer to the ADME/Tox Support Center.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Improper thawing technique |
|
Sub-optimal thawing medium | Use HTM Medium during thawing to remove cryoprotectant |
Incorrect centrifugation speed | Check thawing protocol for proper centrifugation speed and time (varies by species; human is 100 x g for 10 min at RT) |
Rough handling of hepatocytes during counting |
|
Improper counting technique |
|
For additional information, please refer to the ADME/Tox Support Center.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Not enough time for cells to attach |
|
Poor-quality substratum | Use Gibco Collagen I-Coated Plates |
Hepatocyte lot not characterized as plateable |
|
For additional information, please refer to the ADME/Tox Support Center.
Please see our recommendations above for:
Possible cause | Recommendation |
Seeding density too low | Check lot-specific characterization specification sheet for appropriate seeding density (human cells) Observe cells under microscope for appropriate seeding prior to incubation |
Insufficient dispersion of hepatocytes during plating | Disperse cells evenly by moving plate slowly in a figure-eight and back and forth pattern in incubator |
Insufficient plating volume used for well format | Refer to literature or technical support for suggested plating volumes |
Low attachment efficiency | Please see our recommendations above for: “I’m getting low attachment efficiency with my hepatocytes. What should I do?” |
Some animal lots are not >80% confluent | Check lot-specific characterization specification sheet for appropriate seeding density. Note: Some animal species create chains or islands of cells rather than being 100% confluent. |
For additional information, please refer to the ADME/Tox Support Center.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Seeding density too high |
|
Insufficient dispersion of hepatocytes during plating |
|
Improper plating volume used for well format | Refer to literature or technical support for suggested plating volumes |
For additional information, please refer to the ADME/Tox Support Center.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Hepatocyte lot not characterized as plateable | Check lot specifications to ensure it is qualified for plating |
Sub-optimal culture medium |
|
Cells were cultured for too long | In general, plateable cryopreserved hepatocytes should not be cultured for more than five days |
For additional information, please refer to the ADME/Tox Support Center.
Please see the following causes and recommendations:
Possible cause | Recommendation |
Hepatocyte lot not transporter-qualified | Check lot specifications to ensure it is transporter-qualified |
Sub-optimal culture medium |
|
Not enough time for bile canaliculi to form | In general, at least 4–5 days in culture is required for bile canalicular network formation |
For additional information, please refer to the ADME/Tox Support Center.
First of all, we recommend comparing results to those reported on our lot-specific characterization specification sheet (human cells) and also referring to our enzyme induction protocol. Here are other potential causes and recommendations:
Possible Cause | Recommendation |
Sub-optimal monolayer confluency
| Please see our recommendations above for ‘I have a sub-optimal monolayer confluency for my hepatocytes. What should I do?’ |
Poor monolayer integrity
| Please see our recommendations below for ‘With my hepatocytes, I’m seeing rounding up of the cells, cellular debris, and/or holes in the monolayer, indicating dying cells. What should I do?’ |
Inappropriate positive control | Check positive control to ensure suitability |
Incorrect concentration of positive control | Use the correct concentration of positive control |
For additional information, please refer to the ADME/Tox Support Center.
This could be due to the toxicity of the test compound. Here are other potential causes and recommendations:
Possible cause | Recommendation |
Sub-optimal culture medium |
|
Hepatocyte lot not characterized as plateable | Check lot specifications to ensure it is qualified for plating |
Cells were cultured for too long | In general, plateable cryopreserved hepatocytes should not be cultured for more than five days |
For additional information, please refer to the ADME/Tox Support Center.
There are two possible causes for this issue:
These cells are very fragile. We recommend that you follow the procedure in the manual and use the correct medium. Fast thawing is the key for healthy culture. There are several critical points to consider:
There are several possibilities causing the failure of neural induction: