Search Thermo Fisher Scientific
A range of hotplates from basic to specialty, to suit any lab’s needs. Choose from ceramic or aluminum surface types and multiple plate shapes and sizes. Our uniform-heating hotplates deliver a host of capabilities that deliver reproducible results, including temperature stability, durability, and remote-control access for safety and convenience.
Find the hotplate to meet your needs using our hot plate selection guide ›
Basic | Standard | Advanced | |
---|---|---|---|
User Interface | Analog | Digital | Digital |
Timer | ✓ | ||
Save Settings | ✓ | ||
Hot Top Warning Light (Safety Feature) | ✓ | ✓ | |
Optional Accessories | * | ✓ | ✓ |
Quick Heat-up Time to Max Temp | ✓ | ||
Accuracy of temp setting | Within ±1 degree | Within ±1 degree | |
Round Top Option | ✓ | ✓ | ✓ |
Square Top Option | ✓ | ✓ | ✓ |
Ceramic Surface | ✓ | ✓ | ✓ |
Aluminum Surface | ✓ | ✓ | ✓ |
Products | Cimarec Basic Hotplates |