Search Thermo Fisher Scientific
Invitrogen Novex Value Tris-Glycine gels are affordable polyacrylamide gels that provide good separation and resolution of your proteins for traditional Laemmli protein electrophoresis. These gels are compatible with the Invitrogen Mini gel tank.
Features of Novex Value Tris-Glycine gels include:
Superior band quality with Novex Value Tris-Glycine Mini Gels. Protein ladders, purified proteins, and E. coli lysate were loaded on a Novex Value Tris-Glycine 4–20% gradient gel and Supplier B’s 4–20% gradient gel. Better protein band resolution in lysate samples and sharper, better-resolved high–molecular weight and low-abundance protein bands in purified protein samples are observed on Novex Value Tris-Glycine Mini Gels. Our gel also shows superior Coomassie staining/destaining qualities compared to Supplier B’s gel. Lanes 1, 5, 10: 5 μL Thermo Scientific™ PageRuler™ Unstained Protein Ladder; lanes 2, 6, 9: 5 μL Invitrogen™ Mark12™ Unstained Standard; lane 3: 10 μg E. coli lysate (10 μL sample volume); lane 4: 6 μg BSA (10 μL sample volume); lane 7: 6 μg h-IgG (10 μL sample volume); lane 8: 20 μg E. coli lysate (20 μL sample volume).
Novex Value Tris-Glycine Gel | Novex™ WedgeWell™ Tris-Glycine | |
---|---|---|
Band shape and sharpness | Good | Great |
Staining / destaining | Great | Great |
Transfer efficiency | Good | Great |
Run time | 46 min (225V) | 50 min (225V) |
Buffer system | Novex™ Tris-Glycine buffer | Novex Tris-Glycine buffer |
Well type | Standard | WedgeWell |
Applications |
|