PEPTIDE-Custom-Peptides-250
Thermo Scientific™ Custom Peptide Synthesis Service offers a broad range of peptide modifications to help meet a variety of research needs. Peptides between 6 and 70 amino acids in length are synthesized using the latest FMOC solid-phase technology and purified using HPLC. A range of scales (0.1 mg–1 g) and purity (crude to 98%) are available. All peptides come with mass spectrometric (MALDI MS) analysis (crude) and MALDI MS and HPLC (purified).
  • High quality—synthesized using pure materials and packaged using ArgonGuard
  • Versatility—over 400 N-, C-terminal, and internal peptide modifications available
  • Validation—all peptides are analyzed by MS alone or combined with analytical HPLC
  • Enhanced services—IMP service provides traceability and documentation
Custom peptide modifications selection guide
Featured peptide categories
A selection guide to seperate options for the over 400 N- and C-terminal and internal peptide modifications to help you produce the custom peptides that you need to propel your research forward.
Our HeavyPeptide AQUA Custom Synthesis Service provides isotopically labelled, AQUA-grade peptides for the relative and absolute quantitation of proteins at very low concentrations in complex protein mixtures.
Our 2-step approach to accelerate targeted assay development and absolute quantitation by selected reaction monitoring (SRM) within a controlled budget.
Our service combines keyhole limpet hemocyanin (KLH), the most immunogenic and widely-used carrier protein for the preparation of peptide antigens, with high-quality peptides at multiple scale and purity options.