Search Thermo Fisher Scientific
Since 2011, Ion Torrent next-generation sequencing (NGS) has provided complete solutions for clinical research of human diseases with leading Ion AmpliSeq technology.
Now, Ion AmpliSeq On-Demand Panels are available for the Ion Torrent Genexus System. Together, they provide a powerful targeted NGS workflow that enables any lab to go from specimen to variant report in a single day*, providing accessibility, efficiency, and convenience at a speed never possible before.
This highly flexible system lets you process samples—even just one—cost-effectively, as they come in. The hands-off, set-up-and-go workflow makes NGS accessible, even if your human genetics lab is new to the technology.
Ion Torrent NGS reduces complexities that come with other sequencing methods
Dr. Jose Costa, Affiliate Researcher at IPATIMUP, discusses how Ion Torrent NGS enables clinical research labs to overcome common challenges of bringing NGS to their lab.
See how IPATIMUP is developing targeted NGS assays for the Genexus system and Ion GeneStudio S5 system to further clinical research into Noonan syndrome and other RASopathies. Read blog post >
Benefits of Ion AmpliSeq On-Demand Panels on the Genexus System for inherited disease clinical research:
The Ion Torrent Genexus System integrates and automates nucleic acid extraction and purification, library preparation, sequencing, and analysis under a single software ecosystem. This all-in-one solution boosts your lab’s overall efficiency by reducing the number of instruments and consumables required in the lab and frees up your time for more technical applications.
With just 10 minutes of hands-on time and two touchpoints, all users can get up and running quickly with significantly less training, making NGS accessible even if your lab is new to the technology.
Ion AmpliSeq On-Demand Panels enables flexible design and iteration of custom gene panels for clinical research of human diseases—conveniently and with confidence.
Our powerful content selection engine automates optimal gene selection based on the degree of gene/disease association, informed by public databases like MeSH.
Over 5,000 genes available in the selection tool are optimized and pretested, with wet-lab performance data, allowing you to proceed with confidence in your panel design and clinical research outcomes.
Easily configure your panel from an optimized catalog of over 5,000 wet-lab tested genes most relevant in clinical research of inherited diseases like hereditary cancers, primary immunodeficiency, hearing loss, muscular dystrophy, Noonan syndrome and many more (Figure 1).
There’s no time to waste. With just 10 minutes of hands-on time and two user touchpoints, our solution for clinical research of hereditary cancer enables single-day turnaround time from biological specimen to variants*.
Leverage the power of automation, rapid turnaround time, and easy NGS panel design with the winning combination of Genexus System and Ion AmpliSeq On-Demand Panels—an optimal pairing for studying hereditary cancers.
In this application note, we demonstrate the DNA-to-variant report NGS workflow on the Genexus Integrated Sequencer using two Ion AmpliSeq On-Demand panels for clinical research of hereditary cancer.
Tumor type | Genes |
---|---|
Breast | BRCA1, BRCA2 |
Basal cell carcinoma | PTCH1, PTCH2, SUFU |
Colorectal | APC, BMPR1A, EPCAM, MLH1, MLH3, MSH2, MSH6, MUTYH, PMS2, TGFBR2 |
Endocrine gland | CDKN1B, MEN1, NTRK1, RET |
Kidney | DIS3L2, POU6F2, VHL, WT1 |
Connective and soft tissue | EXT1, EXT2, LIFR, PTH1R, PTPN11 |
Nervous system | LZTR1, NF1, NF2, SMARCB1 |
Ovarian | BRCA1, BRCA2 |
Skin | MLH1, MSH2 |
If you don’t see all of your genes of interest listed within our catalog, you can still design an Ion AmpliSeq On-Demand Panel with additional genes spiked-in.
Or create a fully custom gene panels with Ion AmpliSeq Made-to-Order Panels.
Contact your sales representative for ordering support.
Please fill out the project request form to inquire about ordering the Genexus Integrated Sequencer for inherited disease clinical research applications.
Blog Post: Automated targeted next-generation sequencing (NGS) critical in RASopathies clinical research
Blog Post: Targeted panels or exome—which is the right NGS approach for inherited disease research?
App Note: Identification of relevant genetic variants in hereditary cancer research samples