MicroScreen Urine Drug and Alcohol Screening Cups
Explore our drug screening solutions
Drug and Alcohol testing with MicroScreen 2 and MicroScreen Split Cups
Make your workplace safer with the MicroScreen 2 and Microscreen Split urine drug and alcohol screening cups. Designed for use across a diverse range of industries such as mining, transportation, manufacturing, healthcare, construction, agriculture and marine operations. MicroScreen 2 and MicroScreen Split cups help quickly identify risks and support the prevention of workplace injuries caused by the use of illicit drugs, alcohol and dangerous substances.
  Informational: This product is not available for purchase by the general public
Detectable drugs of abuse
MicroScreen 2 for urine drug screening
The MicroScreen™ 2 urine drug testing cup provides preliminary screening for six drugs of abuse:
  • Amphetamines
  • Methamphetamines
  • Benzodiazepines
  • Opiates
  • Cocaine
  • THC (Cannabis)
  The MicroScreen 2 is independently validated  (AS/NZS4308:2008) for workplace and forensic use as well as for clinical use. ARTG (Ref: 370490) 
MicroScreen 2 Plus for urine drug and alcohol screening
The MicroScreen™ 2 Plus urine drug and alcohol screening cup provides preliminary testing for eight drugs of abuse:
  • Amphetamines
  • Methamphetamines
  • Benzodiazepines
  • Opiates
  • Cocaine
  • THC (Cannabis)
  • Oxycodone
  • Ethyl glucuronide (marker of recent alcohol ingestion)
  The MicroScreen 2 Plus is independently validated  (AS/NZS4308:2008) for workplace testing and forensic use only
MicroScreen Split Cup for urine drug screening
The MicroScreen Split urine drug testing cup provides preliminary screening for six drugs of abuse:
  • Amphetamines
  • Methamphetamines
  • Benzodiazepines
  • Opiates
  • Cocaine
  • THC (Cannabis)
  The MicroScreen Split Cup is independently validated  (AS/NZS4308:2008) for workplace and forensic use only
Why choose the MicroScreen urine drug screening cups?
  • Accurate and reliable: MicroScreen 2 urine drug testing cups are manufactured to comply with the Australia and New Zealand Drug Standards (AS/NZS4308:2008)
  • Designed to minimise the risk of tampering:
    o   Unlike all-in-one devices, the donor does not handle the testing panel component of the MicroScreen 2 device, significantly reducing the potential risk for tampering
    o    MicroScreen 2 also features in-built temperature and adulteration panels to provide an indication of sample tampering
    o   Both the testing panel and collection cup are supplied with tamper-evident packaging
  • No manual handling of specimen means there is reduced potential for contamination
  • Rapid results: Results ready in 5-10 minutes
  • Certificate of compliance, available upon request
  • Convenient: The MicroScreen Split Cup’s unique key mechanism provides a simple activation of the test procedure along with control for when the test starts.
  • Secure and leak-proof: Included robust no-leak rubber seal with a click-down lid the Microscreen Split cup ensures sample integrity during testing and transport.
  • Accurate and reliableMicroScreen Split Cup is verified by AS/NZS4308 test strips. Includes temperature verification and an inbuilt adulteration panel for confirming urine sample integrity.
  • Rapid results & efficient samplingFast negative results with minimal urine required. Easy 3-step process: Collect urine, push the key and read the results.
Tilt vs Split Cup samples for workplace testing – understanding the differences
Tilt Cup Drug Testing Cup
Split Cup Drug Testing Cup
Tamper proof design Separate test lid and cup. Unlike all-in-one devices, the donor does not handle the testing panel component of the MicroScreen 2 device, significantly reducing the potential risk for tampering. Protective label hides the test panel and adulteration strips from the donor.
Test activation Elegant tilt design that’s easy for collectors to control when the test starts.  Key activated. Test starts when the collector is ready (not when the subject gives the specimen) and is activated with a simple key mechanism.
Lid Tamper proof secure screw lid provided in separate packaging. Robust no-leak rubber seal with a click-down lid. 
Simplicity and user-friendly
Compact box with a smaller footprint than other cups in the market allowing for easy transportation and storage.
Rapid results with minimal urine required. (Results ready in 5-10 minutes)
Rapid results with minimal urine required. (Results ready in 5-10 minutes)
3-step process: Collect urine, push the key and read the results.
Validation Independently validated  (AS/NZS4308:2008) for workplace and forensic use. The MicroScreen2 Cup is also included on the ARTG (Ref: 370490) for clinical use. Independently validated (AS/NZS4308:2008) for workplace and forensic use. 
How to use the MicroScreen urine drug testing cups
Step-by-step MicroScreen 2 test procedure
How to test in 4 easy steps
Step 1: Client receives MicroScreen 2 cup component and collects sample
Step 2: Record temperature and verify it is within range (32 - 38℃)
Step 3: Attach the test panel lid to the cup with the collected sample, ensuring correct alignment to prevent cross-threading and leakage
Step 4: Tilt to run. Place cup on its side and read results after 5 minutes
Reading Microscreen 2 results
Wait 5 minutes before reading positive results.
Do not read results past 10 minutes.
Negative
A negative test is indicated by the presence of a test line below each designated drug. Note: Line intensities will vary. Any line, even faint, = NEG.
Positive
The ABSENCE of a test line (T) for a designated drug = POS.
Invalid
An invalid test is indicated when the control lines (C) are completely missing in one or more of the test windows. In the event of an invalid screen, a second screen should be performed.
Step-by-step MicroScreen Split test procedure
Step 1: Client receives MicroScreen Split cup and collects sample. Collector records the temperature and verify it is within range (32 - 38℃)
Step 2: Place cup on a flat surface, insert key and push in
Step 3: Peel off the label to view results. 
Interpret adulteration strips between 3-5 minutes. Refer to enclosed colour chart for interpretation. Do not read results past 10 minutes
Analyte performance
Accuracy
Drug test   GC-MS
(<-50% C/0)
GC-MS
(-50% C/0 to C/0)
GC-MS
(C/0 to +50% C/0)
GC-MS
(>+50% C/0)
% Agreement
with GC/MS
AMP/300 (+) 0 1 11 52 100%
(-) 40 9 0 0 98%
BZO/200 (+) 0 3 13 43 100%
(-) 40 10 0 0 94.30%
COC/300 (+) 0 0 10 35 100%
(-) 40 12 0 0 100%
ETG/500 (+) 0 1 15 65 97.65%
(-) 141 23 5 0 99.40%
MET/300 (+) 0 0 9 43 98.10%
(-) 40 7 1 0 100%
OPI/300 (+) 0 1 10 46 100%
(-) 40 8 0 0 98%
OXY/100 (+) 0 3 7 33 100%
(-) 40 1 0 0 93.20%
THC/50 (+) 0 1 11 44 100%
(-) 40 7 0 0 97.90%
Precision
AMP (ng/ml) 0 150 450
(+/-) 0/30 0/30 30/0
BZO (ng/ml) 0 100 300
(+/-) 0/30 0/30 30/0
COC (ng/ml) 0 150 450
(+/-) 0/30 0/30 30/0
EtG (ng/ml) 0 250 750
(+/-) 0/30 0/30 30/0
MET (ng/ml) 0 150 450
(+/-) 0/30 0/30 30/0
OPI (ng/ml) 0 150 450
(+/-) 0/30 0/30 30/0
OXY (ng/ml) 0 50 75
(+/-) 0/30 0/30 30/0
THC (ng/ml) 0 25 75
(+/-) 0/30 0/30 30/0
A drug-free urine pool was spiked with drugs at the listed concentrations. The results are summarised below.
Drug Concentration Cut-off Range
AMP
300
BZO
200
COC
300
OPI
300
MET
300
THC
50
-
+
-
+
-
+
-
+
-
+
-
+
0% Cut-off
30
0
30
0
30
0
30
0
30
0
30
0
-50% Cut-off
30
0
30
0
30
0
30
0
30
0
30
0
-25% Cut-off
27
3
27
3
26
4
26
4
27
3
26
4
Cut-off
15
15
16
14
13
17
15
15
16
14
14
16
+25% Cut-off
4
26
3
27
3
27
3
27
3
27
3
27
+50% Cut-off
0
30
0
30
0
30
0
30
0
30
0
30
+300% Cut-off
0
30
0
30
0
30
0
30
0
30
0
30
Application highlight - mining sector
Screen for safety in mining
The safety implications of drug and alcohol misuse in the mining setting elevates workplace risk on a mine site. Drug and alcohol screening is essential to the health and safety of the Australian mining workplace.
Featured drugs of abuse testing resources
Watch drug testing overview video
Learn more about Microscreen 2