Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Our sterilisation consumables range from products used in gamma irradiation and dry heat sterilisation, indicator tapes, indicator strips, test kits, seal validation, and more.
Our indicator tapes and strips allow for the reliable monitoring of critical parameters of the sterilisation cycle in accordance with ISO 11140-1. Our products feature clearly contrasting colours before and after sterilisation for safe use. Interpretation of colour changes helps to detect malfunction or mistakes in loading of a steriliser. Permanent end-point colour may be used for recordkeeping.